Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 124983135-124983718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF217
Entrez Gene ID
154214
Gene Name
ring finger protein 217
Gene Aliases
C6orf172, IBRDC1, OSTL, dJ84N20.1
UniGene
Hs.12876
Gene Chromosome Location
Chr.6: 124962407-125092634 on Build GRCh38
Transcripts
NM_001286398.2 NM_152553.4 NR_104440.2 NR_136734.1 XM_011535494.2 XM_011535496.2 XM_011535500.2

SNP Detail

SNP ID Chromosome Location
rs897540890 Chr.6: 124983174-124983174 on Build GRCh38
rs773430149 Chr.6: 124983609-124983609 on Build GRCh38
rs967030950 Chr.6: 124983497-124983497 on Build GRCh38
rs1006712986 Chr.6: 124983401-124983401 on Build GRCh38
rs1019922490 Chr.6: 124983482-124983482 on Build GRCh38
rs1030195852 Chr.6: 124983563-124983563 on Build GRCh38
rs941940189 Chr.6: 124983614-124983614 on Build GRCh38
rs1023133065 Chr.6: 124983353-124983353 on Build GRCh38
rs376452948 Chr.6: 124983464-124983464 on Build GRCh38
rs768817643 Chr.6: 124983197-124983197 on Build GRCh38
rs1051828024 Chr.6: 124983675-124983675 on Build GRCh38
rs1036294269 Chr.6: 124983200-124983200 on Build GRCh38
rs933316085 Chr.6: 124983640-124983640 on Build GRCh38
rs1000289984 Chr.6: 124983446-124983446 on Build GRCh38
rs1030371536 Chr.6: 124983273-124983273 on Build GRCh38
rs981220159 Chr.6: 124983509-124983509 on Build GRCh38
rs897869570 Chr.6: 124983630-124983630 on Build GRCh38
rs890988912 Chr.6: 124983308-124983308 on Build GRCh38
rs889182357 Chr.6: 124983681-124983681 on Build GRCh38
rs114103798 Chr.6: 124983600-124983600 on Build GRCh38
rs937527927 Chr.6: 124983551-124983551 on Build GRCh38
rs997517770 Chr.6: 124983241-124983241 on Build GRCh38
rs1012574503 Chr.6: 124983322-124983322 on Build GRCh38
rs1018625265 Chr.6: 124983421-124983421 on Build GRCh38
rs895475104 Chr.6: 124983283-124983283 on Build GRCh38
rs576340063 Chr.6: 124983487-124983487 on Build GRCh38
rs545384694 Chr.6: 124983523-124983523 on Build GRCh38
rs904790532 Chr.6: 124983398-124983398 on Build GRCh38
rs897924988 Chr.6: 124983225-124983225 on Build GRCh38
rs140904347 Chr.6: 124983625-124983625 on Build GRCh38
rs919084279 Chr.6: 124983627-124983627 on Build GRCh38
rs779587363 Chr.6: 124983474-124983474 on Build GRCh38
rs954070802 Chr.6: 124983585-124983585 on Build GRCh38
rs760850050 Chr.6: 124983682-124983682 on Build GRCh38
rs910438261 Chr.6: 124983605-124983605 on Build GRCh38
rs942046216 Chr.6: 124983173-124983173 on Build GRCh38
rs957280685 Chr.6: 124983455-124983455 on Build GRCh38
rs909470048 Chr.6: 124983606-124983606 on Build GRCh38
rs1007148596 Chr.6: 124983683-124983683 on Build GRCh38
rs562635280 Chr.6: 124983350-124983350 on Build GRCh38
rs1018073257 Chr.6: 124983695-124983695 on Build GRCh38

View Full Product Details