Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14457620-14458167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC6A6
Entrez Gene ID
6533
Gene Name
solute carrier family 6 member 6
Gene Aliases
TAUT
UniGene
Hs.529488
Gene Chromosome Location
Chr.3: 14402576-14489349 on Build GRCh38
Transcripts
NM_001134367.3 NM_003043.5 NR_103507.2 XM_006713307.2 XM_011534030.2 XM_011534031.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6468520 c.675T>G p.L225L Chr.3: 14458025-14458025 on Build GRCh38
4504743 c.673C>T p.L225F Chr.3: 14458023-14458023 on Build GRCh38
4848413 c.666C>G p.L222L Chr.3: 14458016-14458016 on Build GRCh38
1664239 c.625G>T p.G209* Chr.3: 14457975-14457975 on Build GRCh38
303239 c.610C>T p.L204L Chr.3: 14457960-14457960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767645569 Chr.3: 14457957-14457957 on Build GRCh38
rs202158519 Chr.3: 14457921-14457921 on Build GRCh38
rs986656067 Chr.3: 14457894-14457894 on Build GRCh38
rs759805633 Chr.3: 14458026-14458026 on Build GRCh38
rs538138027 Chr.3: 14457971-14457971 on Build GRCh38
rs776275164 Chr.3: 14457915-14457915 on Build GRCh38
rs775653728 Chr.3: 14458064-14458064 on Build GRCh38
rs887166175 Chr.3: 14457669-14457669 on Build GRCh38
rs765690879 Chr.3: 14458098-14458098 on Build GRCh38
rs1056494406 Chr.3: 14457658-14457658 on Build GRCh38
rs375554283 Chr.3: 14458127-14458127 on Build GRCh38
rs757701813 Chr.3: 14458072-14458072 on Build GRCh38
rs760251422 Chr.3: 14457883-14457883 on Build GRCh38
rs377494450 Chr.3: 14457994-14457994 on Build GRCh38
rs533640088 Chr.3: 14458102-14458102 on Build GRCh38
rs201094151 Chr.3: 14458065-14458065 on Build GRCh38
rs754717139 Chr.3: 14458105-14458105 on Build GRCh38
rs752754187 Chr.3: 14457959-14457959 on Build GRCh38
rs747629906 Chr.3: 14457981-14457981 on Build GRCh38
rs767698700 Chr.3: 14458031-14458031 on Build GRCh38
rs554207043 Chr.3: 14458112-14458112 on Build GRCh38
rs754374495 Chr.3: 14458069-14458069 on Build GRCh38
rs200442035 Chr.3: 14458038-14458038 on Build GRCh38
rs1056543361 Chr.3: 14458089-14458089 on Build GRCh38
rs955273737 Chr.3: 14457892-14457893 on Build GRCh38
rs143655034 Chr.3: 14457962-14457962 on Build GRCh38
rs764103908 Chr.3: 14458068-14458068 on Build GRCh38
rs762835002 Chr.3: 14457931-14457931 on Build GRCh38
rs774009626 Chr.3: 14458012-14458012 on Build GRCh38
rs374763222 Chr.3: 14457845-14457845 on Build GRCh38
rs1031043943 Chr.3: 14458142-14458142 on Build GRCh38
rs143191441 Chr.3: 14457844-14457844 on Build GRCh38
rs545799997 Chr.3: 14458094-14458094 on Build GRCh38
rs770761525 Chr.3: 14458011-14458011 on Build GRCh38
rs879459373 Chr.3: 14457765-14457765 on Build GRCh38
rs980718220 Chr.3: 14458093-14458093 on Build GRCh38
rs895407355 Chr.3: 14457761-14457761 on Build GRCh38
rs997114795 Chr.3: 14457882-14457882 on Build GRCh38
rs746743487 Chr.3: 14457972-14457972 on Build GRCh38
rs755694932 Chr.3: 14458117-14458117 on Build GRCh38
rs775906052 Chr.3: 14457914-14457920 on Build GRCh38
rs529797674 Chr.3: 14457788-14457788 on Build GRCh38
rs765906100 Chr.3: 14457965-14457965 on Build GRCh38
rs769832662 Chr.3: 14457917-14457917 on Build GRCh38
rs1022551839 Chr.3: 14457796-14457796 on Build GRCh38
rs768409026 Chr.3: 14457977-14457977 on Build GRCh38
rs560928917 Chr.3: 14457764-14457764 on Build GRCh38
rs761230581 Chr.3: 14457935-14457936 on Build GRCh38
rs769387760 Chr.3: 14457992-14457992 on Build GRCh38
rs1003728967 Chr.3: 14457655-14457655 on Build GRCh38
rs1035575403 Chr.3: 14457745-14457745 on Build GRCh38
rs749118284 Chr.3: 14458010-14458010 on Build GRCh38
rs774245573 Chr.3: 14458131-14458131 on Build GRCh38
rs138163519 Chr.3: 14457952-14457952 on Build GRCh38
rs566272127 Chr.3: 14458096-14458096 on Build GRCh38
rs200030714 Chr.3: 14457909-14457909 on Build GRCh38
rs751818003 Chr.3: 14457942-14457942 on Build GRCh38
rs754250557 Chr.3: 14457966-14457966 on Build GRCh38
rs373791222 Chr.3: 14457914-14457914 on Build GRCh38
rs761898015 Chr.3: 14457916-14457916 on Build GRCh38
rs940107066 Chr.3: 14458146-14458146 on Build GRCh38
rs575166633 Chr.3: 14457741-14457741 on Build GRCh38
rs766035129 Chr.3: 14457933-14457933 on Build GRCh38
rs778286920 Chr.3: 14457964-14457964 on Build GRCh38
rs780896452 Chr.3: 14457980-14457980 on Build GRCh38
rs200206592 Chr.3: 14458106-14458106 on Build GRCh38
rs968253214 Chr.3: 14458067-14458067 on Build GRCh38
rs371021588 Chr.3: 14458124-14458124 on Build GRCh38
rs1004366392 Chr.3: 14457834-14457834 on Build GRCh38

View Full Product Details