Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 65523921-65524438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LAS1L
Entrez Gene ID
81887
Gene Name
LAS1 like, ribosome biogenesis factor
Gene Aliases
Las1-like, dJ475B7.2
UniGene
Hs.522675
Gene Chromosome Location
Chr.X: 65512582-65534806 on Build GRCh38
Transcripts
NM_001170649.1 NM_001170650.1 NM_031206.4 XM_005262301.1 XM_005262304.1 XM_005262305.3 XM_005262306.3 XM_011531045.1 XM_011531046.1 XM_017029877.1 XM_017029878.1 XM_017029879.1 XM_017029880.1

SNP Detail

SNP ID Chromosome Location
rs1048902938 Chr.X: 65524322-65524322 on Build GRCh38
rs766096009 Chr.X: 65523952-65523952 on Build GRCh38
rs373363607 Chr.X: 65524047-65524047 on Build GRCh38
rs369880834 Chr.X: 65524035-65524035 on Build GRCh38
rs771153611 Chr.X: 65524288-65524288 on Build GRCh38
rs186130633 Chr.X: 65524242-65524242 on Build GRCh38
rs755816378 Chr.X: 65524195-65524195 on Build GRCh38
rs952101815 Chr.X: 65524385-65524385 on Build GRCh38
rs768620249 Chr.X: 65524015-65524015 on Build GRCh38
rs375513406 Chr.X: 65524010-65524010 on Build GRCh38
rs759857942 Chr.X: 65524105-65524105 on Build GRCh38
rs768280616 Chr.X: 65524299-65524300 on Build GRCh38
rs749134583 Chr.X: 65524014-65524014 on Build GRCh38
rs770283190 Chr.X: 65524286-65524287 on Build GRCh38
rs779625300 Chr.X: 65524222-65524222 on Build GRCh38
rs369291009 Chr.X: 65524258-65524258 on Build GRCh38
rs940708873 Chr.X: 65523972-65523972 on Build GRCh38
rs141099046 Chr.X: 65524147-65524147 on Build GRCh38
rs373278066 Chr.X: 65524171-65524171 on Build GRCh38
rs919155075 Chr.X: 65524172-65524172 on Build GRCh38
rs759932688 Chr.X: 65524301-65524301 on Build GRCh38
rs962344354 Chr.X: 65524360-65524360 on Build GRCh38
rs887612234 Chr.X: 65524383-65524383 on Build GRCh38
rs113353326 Chr.X: 65524053-65524053 on Build GRCh38
rs201135811 Chr.X: 65524307-65524307 on Build GRCh38
rs188766357 Chr.X: 65524087-65524087 on Build GRCh38
rs772288401 Chr.X: 65524042-65524042 on Build GRCh38
rs565077547 Chr.X: 65524123-65524123 on Build GRCh38
rs776784036 Chr.X: 65524290-65524290 on Build GRCh38
rs190949399 Chr.X: 65524384-65524384 on Build GRCh38
rs748222445 Chr.X: 65524041-65524041 on Build GRCh38
rs753261127 Chr.X: 65524120-65524120 on Build GRCh38
rs763483309 Chr.X: 65524313-65524313 on Build GRCh38
rs780681664 Chr.X: 65524287-65524287 on Build GRCh38
rs986187652 Chr.X: 65523996-65523996 on Build GRCh38
rs1035253086 Chr.X: 65524314-65524315 on Build GRCh38
rs1003637820 Chr.X: 65524310-65524310 on Build GRCh38
rs920573023 Chr.X: 65524029-65524030 on Build GRCh38
rs748098548 Chr.X: 65524274-65524274 on Build GRCh38
rs749847829 Chr.X: 65524300-65524301 on Build GRCh38
rs777030562 Chr.X: 65524088-65524088 on Build GRCh38
rs766968964 Chr.X: 65524314-65524314 on Build GRCh38
rs772235476 Chr.X: 65524275-65524275 on Build GRCh38
rs1054870452 Chr.X: 65524408-65524408 on Build GRCh38
rs972711887 Chr.X: 65524378-65524378 on Build GRCh38
rs113072894 Chr.X: 65524054-65524054 on Build GRCh38
rs926144541 Chr.X: 65523950-65523950 on Build GRCh38
rs56185266 Chr.X: 65524306-65524306 on Build GRCh38
rs761006702 Chr.X: 65524086-65524086 on Build GRCh38
rs1057518699 Chr.X: 65524113-65524113 on Build GRCh38
rs180689421 Chr.X: 65524108-65524108 on Build GRCh38
rs774199645 Chr.X: 65524033-65524033 on Build GRCh38
rs758840845 Chr.X: 65524287-65524287 on Build GRCh38
rs778847976 Chr.X: 65524265-65524265 on Build GRCh38

View Full Product Details