Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 108047265-108047858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LEF1
Entrez Gene ID
51176
Gene Name
lymphoid enhancer binding factor 1
Gene Aliases
LEF-1, TCF10, TCF1ALPHA, TCF7L3
UniGene
Hs.743478
Gene Chromosome Location
Chr.4: 108047545-108168956 on Build GRCh38
Transcripts
NM_001130713.2 NM_001130714.2 NM_001166119.1 NM_016269.4 XM_005263046.3 XM_005263047.1 XM_005263048.1 XM_006714233.1

SNP Detail

SNP ID Chromosome Location
rs1018067469 Chr.4: 108047594-108047594 on Build GRCh38
rs565147303 Chr.4: 108047745-108047745 on Build GRCh38
rs572154916 Chr.4: 108047521-108047521 on Build GRCh38
rs990004516 Chr.4: 108047609-108047609 on Build GRCh38
rs555429349 Chr.4: 108047492-108047492 on Build GRCh38
rs543984736 Chr.4: 108047352-108047352 on Build GRCh38
rs927824853 Chr.4: 108047632-108047632 on Build GRCh38
rs1027926971 Chr.4: 108047461-108047461 on Build GRCh38
rs191295179 Chr.4: 108047692-108047692 on Build GRCh38
rs1027976896 Chr.4: 108047558-108047558 on Build GRCh38
rs953665291 Chr.4: 108047574-108047575 on Build GRCh38
rs1012941221 Chr.4: 108047289-108047289 on Build GRCh38
rs577796713 Chr.4: 108047548-108047548 on Build GRCh38
rs545302118 Chr.4: 108047741-108047741 on Build GRCh38
rs1015250115 Chr.4: 108047404-108047404 on Build GRCh38
rs549936803 Chr.4: 108047300-108047300 on Build GRCh38
rs934688077 Chr.4: 108047605-108047605 on Build GRCh38
rs924602504 Chr.4: 108047602-108047602 on Build GRCh38
rs907062743 Chr.4: 108047664-108047664 on Build GRCh38
rs76952910 Chr.4: 108047712-108047712 on Build GRCh38
rs573606188 Chr.4: 108047684-108047684 on Build GRCh38
rs1004342662 Chr.4: 108047396-108047396 on Build GRCh38
rs187773509 Chr.4: 108047525-108047525 on Build GRCh38
rs986322546 Chr.4: 108047586-108047586 on Build GRCh38
rs149801845 Chr.4: 108047710-108047710 on Build GRCh38
rs1027873264 Chr.4: 108047807-108047807 on Build GRCh38
rs796185394 Chr.4: 108047343-108047344 on Build GRCh38
rs558818076 Chr.4: 108047337-108047344 on Build GRCh38
rs948523545 Chr.4: 108047645-108047645 on Build GRCh38
rs896800892 Chr.4: 108047779-108047779 on Build GRCh38
rs11941749 Chr.4: 108047345-108047345 on Build GRCh38
rs557538461 Chr.4: 108047635-108047635 on Build GRCh38
rs1044511885 Chr.4: 108047287-108047287 on Build GRCh38
rs558066780 Chr.4: 108047539-108047539 on Build GRCh38
rs537268952 Chr.4: 108047299-108047299 on Build GRCh38
rs780279077 Chr.4: 108047369-108047369 on Build GRCh38
rs184077366 Chr.4: 108047333-108047333 on Build GRCh38
rs1048384745 Chr.4: 108047777-108047777 on Build GRCh38
rs948699245 Chr.4: 108047763-108047763 on Build GRCh38
rs552296042 Chr.4: 108047685-108047685 on Build GRCh38
rs35125664 Chr.4: 108047400-108047401 on Build GRCh38
rs966153552 Chr.4: 108047595-108047595 on Build GRCh38
rs976607904 Chr.4: 108047596-108047596 on Build GRCh38
rs116637005 Chr.4: 108047431-108047431 on Build GRCh38
rs1026998216 Chr.4: 108047293-108047293 on Build GRCh38
rs576943283 Chr.4: 108047346-108047350 on Build GRCh38
rs995184147 Chr.4: 108047806-108047806 on Build GRCh38
rs989166306 Chr.4: 108047574-108047574 on Build GRCh38
rs768642450 Chr.4: 108047654-108047654 on Build GRCh38
rs570010858 Chr.4: 108047326-108047326 on Build GRCh38
rs543628648 Chr.4: 108047628-108047628 on Build GRCh38
rs993731875 Chr.4: 108047444-108047444 on Build GRCh38

View Full Product Details