Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89919951-89920547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MC1R
Entrez Gene ID
4157
Gene Name
melanocortin 1 receptor
Gene Aliases
CMM5, MSH-R, SHEP2
UniGene
Hs.513829
Gene Chromosome Location
Chr.16: 89917879-89920977 on Build GRCh38
Transcripts
NM_002386.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6709269 c.894C>T p.Y298Y Chr.16: 89920152-89920152 on Build GRCh38
6583864 c.860T>A p.I287N Chr.16: 89920118-89920118 on Build GRCh38
975013 c.853G>A p.A285T Chr.16: 89920111-89920111 on Build GRCh38
4579392 c.949T>C p.W317R Chr.16: 89920207-89920207 on Build GRCh38
6458077 c.832A>G p.K278E Chr.16: 89920090-89920090 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536644599 Chr.16: 89920466-89920466 on Build GRCh38
rs1054207997 Chr.16: 89920025-89920025 on Build GRCh38
rs772969877 Chr.16: 89920150-89920150 on Build GRCh38
rs552512218 Chr.16: 89920063-89920063 on Build GRCh38
rs767276470 Chr.16: 89920013-89920013 on Build GRCh38
rs141177570 Chr.16: 89920100-89920100 on Build GRCh38
rs747847234 Chr.16: 89920174-89920174 on Build GRCh38
rs925012763 Chr.16: 89920114-89920114 on Build GRCh38
rs370073863 Chr.16: 89920045-89920045 on Build GRCh38
rs373703770 Chr.16: 89920127-89920127 on Build GRCh38
rs749154025 Chr.16: 89920233-89920233 on Build GRCh38
rs949262951 Chr.16: 89920132-89920132 on Build GRCh38
rs924429375 Chr.16: 89920019-89920019 on Build GRCh38
rs3212369 Chr.16: 89920352-89920352 on Build GRCh38
rs376508354 Chr.16: 89920028-89920028 on Build GRCh38
rs972412422 Chr.16: 89920493-89920493 on Build GRCh38
rs970473748 Chr.16: 89920345-89920345 on Build GRCh38
rs1805009 Chr.16: 89920138-89920138 on Build GRCh38
rs763065230 Chr.16: 89920037-89920037 on Build GRCh38
rs753883273 Chr.16: 89919973-89919973 on Build GRCh38
rs774110096 Chr.16: 89920187-89920187 on Build GRCh38
rs34490506 Chr.16: 89919975-89919975 on Build GRCh38
rs747968193 Chr.16: 89920143-89920143 on Build GRCh38
rs1024445964 Chr.16: 89920450-89920450 on Build GRCh38
rs753139067 Chr.16: 89920247-89920248 on Build GRCh38
rs575176457 Chr.16: 89920296-89920296 on Build GRCh38
rs377248188 Chr.16: 89920202-89920202 on Build GRCh38
rs776867314 Chr.16: 89919989-89920000 on Build GRCh38
rs926154015 Chr.16: 89920186-89920186 on Build GRCh38
rs528202567 Chr.16: 89920521-89920521 on Build GRCh38
rs973514189 Chr.16: 89920518-89920518 on Build GRCh38
rs373467427 Chr.16: 89920147-89920147 on Build GRCh38
rs780265657 Chr.16: 89920039-89920039 on Build GRCh38
rs1037626233 Chr.16: 89920363-89920363 on Build GRCh38
rs371583010 Chr.16: 89920064-89920064 on Build GRCh38
rs1006964619 Chr.16: 89920362-89920362 on Build GRCh38
rs772323341 Chr.16: 89920199-89920199 on Build GRCh38
rs1017629048 Chr.16: 89920277-89920277 on Build GRCh38
rs746134737 Chr.16: 89919978-89919978 on Build GRCh38
rs771648556 Chr.16: 89920093-89920093 on Build GRCh38
rs899007216 Chr.16: 89920235-89920235 on Build GRCh38
rs766530750 Chr.16: 89920117-89920117 on Build GRCh38
rs368507952 Chr.16: 89920175-89920175 on Build GRCh38
rs770382929 Chr.16: 89920281-89920281 on Build GRCh38
rs780347303 Chr.16: 89919984-89919984 on Build GRCh38
rs911435490 Chr.16: 89920524-89920524 on Build GRCh38
rs755331408 Chr.16: 89920046-89920046 on Build GRCh38
rs748521098 Chr.16: 89920078-89920078 on Build GRCh38
rs551043523 Chr.16: 89920421-89920421 on Build GRCh38
rs34628168 Chr.16: 89920516-89920517 on Build GRCh38
rs376425491 Chr.16: 89920133-89920133 on Build GRCh38
rs780594587 Chr.16: 89920231-89920234 on Build GRCh38
rs762846553 Chr.16: 89920113-89920113 on Build GRCh38
rs564507704 Chr.16: 89920259-89920259 on Build GRCh38
rs910479534 Chr.16: 89919998-89919998 on Build GRCh38
rs780773419 Chr.16: 89920140-89920140 on Build GRCh38
rs375127718 Chr.16: 89920181-89920181 on Build GRCh38
rs536045780 Chr.16: 89920125-89920125 on Build GRCh38
rs587783376 Chr.16: 89920051-89920051 on Build GRCh38
rs748972076 Chr.16: 89920182-89920182 on Build GRCh38
rs3212368 Chr.16: 89920224-89920224 on Build GRCh38
rs988972413 Chr.16: 89920333-89920333 on Build GRCh38
rs760209791 Chr.16: 89920062-89920062 on Build GRCh38
rs761538502 Chr.16: 89920103-89920103 on Build GRCh38
rs886052507 Chr.16: 89920325-89920326 on Build GRCh38
rs181269865 Chr.16: 89920050-89920050 on Build GRCh38
rs370472871 Chr.16: 89920153-89920153 on Build GRCh38
rs182215467 Chr.16: 89920391-89920391 on Build GRCh38
rs757162834 Chr.16: 89920026-89920026 on Build GRCh38
rs781382183 Chr.16: 89920047-89920047 on Build GRCh38
rs375067758 Chr.16: 89920242-89920242 on Build GRCh38
rs750200309 Chr.16: 89920072-89920073 on Build GRCh38
rs761804253 Chr.16: 89920071-89920071 on Build GRCh38
rs550480383 Chr.16: 89920110-89920110 on Build GRCh38
rs778370171 Chr.16: 89920089-89920089 on Build GRCh38
rs1011893326 Chr.16: 89920434-89920434 on Build GRCh38
rs914341120 Chr.16: 89920111-89920111 on Build GRCh38
rs545888975 Chr.16: 89920288-89920288 on Build GRCh38
rs3212367 Chr.16: 89920158-89920158 on Build GRCh38
rs754488118 Chr.16: 89920227-89920245 on Build GRCh38
rs974171310 Chr.16: 89919974-89919974 on Build GRCh38
rs751102007 Chr.16: 89920168-89920168 on Build GRCh38
rs376692024 Chr.16: 89920094-89920094 on Build GRCh38
rs766386985 Chr.16: 89920131-89920133 on Build GRCh38
rs367652300 Chr.16: 89920364-89920364 on Build GRCh38
rs918228951 Chr.16: 89920511-89920511 on Build GRCh38
rs527697723 Chr.16: 89920061-89920061 on Build GRCh38
rs369402699 Chr.16: 89919987-89919987 on Build GRCh38
rs926242817 Chr.16: 89920492-89920492 on Build GRCh38
rs778930648 Chr.16: 89919976-89919976 on Build GRCh38
rs373872609 Chr.16: 89920157-89920157 on Build GRCh38
rs750496563 Chr.16: 89920032-89920032 on Build GRCh38
rs879682994 Chr.16: 89920396-89920396 on Build GRCh38
rs369542041 Chr.16: 89920123-89920123 on Build GRCh38
rs765813858 Chr.16: 89920209-89920209 on Build GRCh38
rs573848209 Chr.16: 89920475-89920475 on Build GRCh38
rs760466103 Chr.16: 89920022-89920022 on Build GRCh38
rs774718264 Chr.16: 89920438-89920438 on Build GRCh38
rs752932359 Chr.16: 89920154-89920154 on Build GRCh38
rs373109025 Chr.16: 89920225-89920225 on Build GRCh38
rs763966451 Chr.16: 89920023-89920023 on Build GRCh38
rs560742802 Chr.16: 89920482-89920482 on Build GRCh38
rs745421220 Chr.16: 89920146-89920146 on Build GRCh38
rs138058610 Chr.16: 89920519-89920519 on Build GRCh38
rs929117769 Chr.16: 89920365-89920365 on Build GRCh38
rs981893689 Chr.16: 89920483-89920483 on Build GRCh38
rs748780226 Chr.16: 89920001-89920001 on Build GRCh38
rs1000459034 Chr.16: 89920287-89920287 on Build GRCh38
rs768777016 Chr.16: 89919985-89919986 on Build GRCh38
rs938331667 Chr.16: 89920271-89920271 on Build GRCh38
rs779680061 Chr.16: 89920137-89920137 on Build GRCh38
rs569349623 Chr.16: 89920437-89920437 on Build GRCh38
rs773094322 Chr.16: 89920112-89920112 on Build GRCh38
rs756738524 Chr.16: 89920122-89920122 on Build GRCh38
rs776589029 Chr.16: 89920497-89920497 on Build GRCh38
rs375867067 Chr.16: 89920161-89920161 on Build GRCh38
rs762901370 Chr.16: 89920115-89920117 on Build GRCh38
rs905089222 Chr.16: 89920216-89920216 on Build GRCh38
rs956319227 Chr.16: 89920428-89920428 on Build GRCh38
rs200051702 Chr.16: 89919983-89919983 on Build GRCh38
rs1039410521 Chr.16: 89920378-89920378 on Build GRCh38
rs990355156 Chr.16: 89920105-89920105 on Build GRCh38
rs200215218 Chr.16: 89920024-89920024 on Build GRCh38
rs754442823 Chr.16: 89920131-89920131 on Build GRCh38
rs373957223 Chr.16: 89920119-89920119 on Build GRCh38
rs765016133 Chr.16: 89920071-89920082 on Build GRCh38
rs1008891042 Chr.16: 89920274-89920274 on Build GRCh38
rs375813196 Chr.16: 89920077-89920077 on Build GRCh38
rs150450096 Chr.16: 89920389-89920389 on Build GRCh38
rs151318945 Chr.16: 89920206-89920206 on Build GRCh38
rs112759721 Chr.16: 89920386-89920386 on Build GRCh38
rs901231418 Chr.16: 89920368-89920368 on Build GRCh38
rs368281517 Chr.16: 89920076-89920076 on Build GRCh38
rs897318380 Chr.16: 89920054-89920054 on Build GRCh38
rs762063106 Chr.16: 89920207-89920207 on Build GRCh38
rs564084675 Chr.16: 89920316-89920316 on Build GRCh38
rs188982766 Chr.16: 89920214-89920214 on Build GRCh38
rs561571645 Chr.16: 89920011-89920011 on Build GRCh38
rs979894076 Chr.16: 89920439-89920439 on Build GRCh38
rs753749977 Chr.16: 89920118-89920118 on Build GRCh38
rs965299910 Chr.16: 89920280-89920280 on Build GRCh38
rs756696134 Chr.16: 89920085-89920085 on Build GRCh38
rs776413951 Chr.16: 89920101-89920101 on Build GRCh38
rs903642958 Chr.16: 89920014-89920014 on Build GRCh38
rs572754025 Chr.16: 89920344-89920344 on Build GRCh38
rs1037285800 Chr.16: 89920141-89920141 on Build GRCh38
rs956202422 Chr.16: 89920324-89920325 on Build GRCh38
rs745994833 Chr.16: 89920193-89920193 on Build GRCh38
rs778725176 Chr.16: 89920262-89920262 on Build GRCh38
rs754405679 Chr.16: 89920169-89920169 on Build GRCh38
rs546300801 Chr.16: 89920228-89920228 on Build GRCh38
rs867971391 Chr.16: 89920483-89920483 on Build GRCh38
rs575607312 Chr.16: 89920166-89920166 on Build GRCh38
rs777147258 Chr.16: 89919989-89919989 on Build GRCh38
rs140568765 Chr.16: 89920317-89920317 on Build GRCh38
rs375200283 Chr.16: 89920223-89920223 on Build GRCh38
rs374760264 Chr.16: 89920074-89920074 on Build GRCh38
rs750924468 Chr.16: 89920211-89920211 on Build GRCh38
rs926324042 Chr.16: 89920354-89920354 on Build GRCh38
rs761973172 Chr.16: 89920011-89920011 on Build GRCh38
rs202197434 Chr.16: 89920095-89920095 on Build GRCh38
rs769056179 Chr.16: 89919988-89919988 on Build GRCh38
rs928322951 Chr.16: 89920268-89920268 on Build GRCh38
rs367998996 Chr.16: 89920053-89920053 on Build GRCh38
rs765090747 Chr.16: 89920072-89920072 on Build GRCh38
rs3212370 Chr.16: 89920369-89920369 on Build GRCh38
rs751594275 Chr.16: 89920152-89920153 on Build GRCh38
rs913639956 Chr.16: 89920339-89920339 on Build GRCh38
rs760953265 Chr.16: 89920155-89920155 on Build GRCh38
rs758623879 Chr.16: 89920033-89920033 on Build GRCh38
rs34020587 Chr.16: 89920226-89920226 on Build GRCh38
rs371214731 Chr.16: 89920021-89920021 on Build GRCh38
rs761041641 Chr.16: 89920201-89920201 on Build GRCh38
rs376659314 Chr.16: 89920002-89920002 on Build GRCh38
rs2228478 Chr.16: 89920200-89920200 on Build GRCh38
rs780650451 Chr.16: 89920172-89920172 on Build GRCh38
rs886052508 Chr.16: 89920522-89920522 on Build GRCh38
rs143395134 Chr.16: 89920152-89920152 on Build GRCh38
rs555253543 Chr.16: 89920468-89920468 on Build GRCh38
rs201171524 Chr.16: 89920090-89920090 on Build GRCh38
rs201028944 Chr.16: 89920012-89920012 on Build GRCh38
rs1044424142 Chr.16: 89920048-89920048 on Build GRCh38

View Full Product Details