Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 56072916-56073474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS20
Entrez Gene ID
6224
Gene Name
ribosomal protein S20
Gene Aliases
S20
UniGene
Hs.8102
Gene Chromosome Location
Chr.8: 56068180-56074581 on Build GRCh38
Transcripts
NM_001023.3 NM_001146227.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
384691 c.260G>T p.R87L Chr.8: 56073190-56073190 on Build GRCh38
6329574 c.252T>C p.I84I Chr.8: 56073198-56073198 on Build GRCh38
6336789 c.317T>C p.I106T Chr.8: 56073133-56073133 on Build GRCh38
1151130 c.280C>G p.P94A Chr.8: 56073170-56073170 on Build GRCh38
1597746 c.197G>T p.R66I Chr.8: 56073253-56073253 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762595301 Chr.8: 56073094-56073094 on Build GRCh38
rs755023684 Chr.8: 56073301-56073301 on Build GRCh38
rs139428379 Chr.8: 56073386-56073386 on Build GRCh38
rs544105403 Chr.8: 56072989-56072989 on Build GRCh38
rs994831715 Chr.8: 56073002-56073002 on Build GRCh38
rs867989887 Chr.8: 56073428-56073428 on Build GRCh38
rs1804368 Chr.8: 56073181-56073181 on Build GRCh38
rs560024590 Chr.8: 56073064-56073064 on Build GRCh38
rs1050511816 Chr.8: 56073439-56073439 on Build GRCh38
rs747254002 Chr.8: 56073178-56073178 on Build GRCh38
rs930724850 Chr.8: 56073066-56073066 on Build GRCh38
rs531289733 Chr.8: 56073234-56073234 on Build GRCh38
rs867773964 Chr.8: 56073252-56073252 on Build GRCh38
rs777824927 Chr.8: 56073141-56073141 on Build GRCh38
rs11537565 Chr.8: 56073176-56073176 on Build GRCh38
rs113128051 Chr.8: 56073060-56073060 on Build GRCh38
rs540124601 Chr.8: 56073222-56073222 on Build GRCh38
rs752279003 Chr.8: 56073276-56073276 on Build GRCh38
rs965052096 Chr.8: 56072940-56072940 on Build GRCh38
rs767366285 Chr.8: 56072967-56072967 on Build GRCh38
rs904 Chr.8: 56073214-56073214 on Build GRCh38
rs532258451 Chr.8: 56073082-56073082 on Build GRCh38
rs1049066812 Chr.8: 56073366-56073366 on Build GRCh38
rs368670078 Chr.8: 56073078-56073078 on Build GRCh38
rs930649684 Chr.8: 56073433-56073433 on Build GRCh38
rs763728957 Chr.8: 56073286-56073286 on Build GRCh38
rs766012489 Chr.8: 56073240-56073240 on Build GRCh38
rs142278807 Chr.8: 56073259-56073259 on Build GRCh38
rs879777423 Chr.8: 56073397-56073397 on Build GRCh38
rs898077873 Chr.8: 56073334-56073334 on Build GRCh38
rs754631108 Chr.8: 56073102-56073102 on Build GRCh38
rs772032278 Chr.8: 56073049-56073049 on Build GRCh38
rs947836738 Chr.8: 56073284-56073284 on Build GRCh38
rs773942943 Chr.8: 56073347-56073347 on Build GRCh38
rs751272740 Chr.8: 56073099-56073099 on Build GRCh38
rs762255340 Chr.8: 56073069-56073069 on Build GRCh38
rs765925127 Chr.8: 56073096-56073096 on Build GRCh38
rs956405729 Chr.8: 56073101-56073101 on Build GRCh38
rs993081450 Chr.8: 56073351-56073351 on Build GRCh38
rs79584175 Chr.8: 56072999-56072999 on Build GRCh38
rs988490352 Chr.8: 56072947-56072947 on Build GRCh38
rs968406813 Chr.8: 56073248-56073248 on Build GRCh38
rs142522386 Chr.8: 56073004-56073004 on Build GRCh38
rs961916137 Chr.8: 56073383-56073383 on Build GRCh38
rs749176723 Chr.8: 56073144-56073144 on Build GRCh38
rs1003810451 Chr.8: 56072960-56072960 on Build GRCh38
rs1019960217 Chr.8: 56073246-56073246 on Build GRCh38
rs568734901 Chr.8: 56073267-56073267 on Build GRCh38
rs768823942 Chr.8: 56073179-56073179 on Build GRCh38
rs781530727 Chr.8: 56073304-56073304 on Build GRCh38
rs77156325 Chr.8: 56073453-56073453 on Build GRCh38
rs199637313 Chr.8: 56073180-56073180 on Build GRCh38
rs757489007 Chr.8: 56073302-56073302 on Build GRCh38
rs554329892 Chr.8: 56073349-56073349 on Build GRCh38
rs889252729 Chr.8: 56073133-56073133 on Build GRCh38
rs201535420 Chr.8: 56073321-56073321 on Build GRCh38
rs372054935 Chr.8: 56073189-56073189 on Build GRCh38
rs906660228 Chr.8: 56072952-56072952 on Build GRCh38
rs755256481 Chr.8: 56073345-56073345 on Build GRCh38
rs371589912 Chr.8: 56073150-56073150 on Build GRCh38
rs901043587 Chr.8: 56073411-56073411 on Build GRCh38
rs917171934 Chr.8: 56073008-56073008 on Build GRCh38
rs940664823 Chr.8: 56073014-56073014 on Build GRCh38
rs748590989 Chr.8: 56073190-56073190 on Build GRCh38
rs878869472 Chr.8: 56073275-56073275 on Build GRCh38
rs749298650 Chr.8: 56073314-56073314 on Build GRCh38
rs756005160 Chr.8: 56073278-56073278 on Build GRCh38
rs1019991396 Chr.8: 56072979-56072979 on Build GRCh38
rs978547374 Chr.8: 56072990-56072990 on Build GRCh38
rs997965601 Chr.8: 56073412-56073412 on Build GRCh38
rs989322222 Chr.8: 56073444-56073444 on Build GRCh38
rs1019430847 Chr.8: 56073401-56073401 on Build GRCh38
rs773566296 Chr.8: 56073084-56073084 on Build GRCh38
rs749847601 Chr.8: 56073320-56073320 on Build GRCh38
rs11537563 Chr.8: 56073245-56073245 on Build GRCh38
rs752579707 Chr.8: 56073117-56073117 on Build GRCh38
rs755967009 Chr.8: 56073053-56073053 on Build GRCh38
rs769020380 Chr.8: 56073061-56073061 on Build GRCh38
rs767329543 Chr.8: 56073274-56073274 on Build GRCh38
rs958542903 Chr.8: 56073325-56073325 on Build GRCh38
rs928171604 Chr.8: 56073296-56073296 on Build GRCh38
rs770211614 Chr.8: 56073312-56073312 on Build GRCh38
rs927140096 Chr.8: 56072997-56072997 on Build GRCh38
rs1050403 Chr.8: 56073255-56073255 on Build GRCh38
rs777901837 Chr.8: 56073015-56073015 on Build GRCh38
rs1030915064 Chr.8: 56073425-56073425 on Build GRCh38
rs750399928 Chr.8: 56073039-56073039 on Build GRCh38
rs780136373 Chr.8: 56073294-56073294 on Build GRCh38
rs767452255 Chr.8: 56073106-56073106 on Build GRCh38
rs570923381 Chr.8: 56073372-56073372 on Build GRCh38
rs575607291 Chr.8: 56073430-56073430 on Build GRCh38
rs1045235671 Chr.8: 56072944-56072944 on Build GRCh38
rs773777295 Chr.8: 56073231-56073231 on Build GRCh38
rs751676310 Chr.8: 56073295-56073295 on Build GRCh38
rs768430426 Chr.8: 56073326-56073326 on Build GRCh38
rs753827762 Chr.8: 56073290-56073290 on Build GRCh38
rs145164279 Chr.8: 56073069-56073069 on Build GRCh38
rs889208761 Chr.8: 56073037-56073037 on Build GRCh38
rs755842545 Chr.8: 56073138-56073138 on Build GRCh38
rs748254952 Chr.8: 56073306-56073306 on Build GRCh38
rs1009848079 Chr.8: 56073223-56073223 on Build GRCh38
rs540399554 Chr.8: 56073050-56073050 on Build GRCh38

View Full Product Details