Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 19306797-19307324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR7A2
Entrez Gene ID
8574
Gene Name
aldo-keto reductase family 7 member A2
Gene Aliases
AFAR, AFAR1, AFB1-AR1, AKR7
UniGene
Hs.571886
Gene Chromosome Location
Chr.1: 19302708-19312146 on Build GRCh38
Transcripts
NM_001320979.1 NM_003689.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7090513 c.730G>T p.G244W Chr.1: 19307060-19307060 on Build GRCh38
6597992 c.746G>A p.G249D Chr.1: 19307044-19307044 on Build GRCh38
6597991 c.692delG p.G231fs*3 Chr.1: 19307098-19307098 on Build GRCh38
6123217 c.752T>C p.F251S Chr.1: 19307038-19307038 on Build GRCh38
7101590 c.772G>A p.E258K Chr.1: 19307018-19307018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527832954 Chr.1: 19307091-19307091 on Build GRCh38
rs749350832 Chr.1: 19307103-19307104 on Build GRCh38
rs759237815 Chr.1: 19307302-19307302 on Build GRCh38
rs759856519 Chr.1: 19306954-19306954 on Build GRCh38
rs202217638 Chr.1: 19307106-19307106 on Build GRCh38
rs150297156 Chr.1: 19307028-19307028 on Build GRCh38
rs373229523 Chr.1: 19307141-19307141 on Build GRCh38
rs770988259 Chr.1: 19307131-19307131 on Build GRCh38
rs538343102 Chr.1: 19307174-19307174 on Build GRCh38
rs931219049 Chr.1: 19306854-19306855 on Build GRCh38
rs775810957 Chr.1: 19307143-19307143 on Build GRCh38
rs372724113 Chr.1: 19306908-19306908 on Build GRCh38
rs908449908 Chr.1: 19307262-19307262 on Build GRCh38
rs753148581 Chr.1: 19306956-19306956 on Build GRCh38
rs994516635 Chr.1: 19307293-19307293 on Build GRCh38
rs145694835 Chr.1: 19307002-19307002 on Build GRCh38
rs140964634 Chr.1: 19307256-19307256 on Build GRCh38
rs529708119 Chr.1: 19306930-19306930 on Build GRCh38
rs774654989 Chr.1: 19307125-19307126 on Build GRCh38
rs565194687 Chr.1: 19306826-19306826 on Build GRCh38
rs377294347 Chr.1: 19306995-19306995 on Build GRCh38
rs777615652 Chr.1: 19307052-19307053 on Build GRCh38
rs1003664028 Chr.1: 19307048-19307048 on Build GRCh38
rs200730767 Chr.1: 19307055-19307055 on Build GRCh38
rs756060304 Chr.1: 19306989-19306990 on Build GRCh38
rs376469810 Chr.1: 19307270-19307270 on Build GRCh38
rs778623206 Chr.1: 19307097-19307097 on Build GRCh38
rs377621683 Chr.1: 19307087-19307087 on Build GRCh38
rs750131969 Chr.1: 19306992-19306992 on Build GRCh38
rs771092024 Chr.1: 19307031-19307031 on Build GRCh38
rs1025194987 Chr.1: 19306903-19306903 on Build GRCh38
rs899056845 Chr.1: 19307288-19307288 on Build GRCh38
rs764649602 Chr.1: 19306963-19306963 on Build GRCh38
rs2231203 Chr.1: 19307026-19307026 on Build GRCh38
rs1002154399 Chr.1: 19307195-19307195 on Build GRCh38
rs1009369280 Chr.1: 19306836-19306836 on Build GRCh38
rs374214238 Chr.1: 19307086-19307086 on Build GRCh38
rs777722858 Chr.1: 19307123-19307123 on Build GRCh38
rs567174054 Chr.1: 19307061-19307061 on Build GRCh38
rs774182583 Chr.1: 19307299-19307299 on Build GRCh38
rs776044049 Chr.1: 19307043-19307043 on Build GRCh38
rs973910609 Chr.1: 19307127-19307127 on Build GRCh38
rs370539329 Chr.1: 19307128-19307128 on Build GRCh38
rs906453607 Chr.1: 19306916-19306916 on Build GRCh38
rs2231201 Chr.1: 19307153-19307153 on Build GRCh38
rs748089759 Chr.1: 19307011-19307011 on Build GRCh38
rs759911672 Chr.1: 19307041-19307041 on Build GRCh38
rs1025949183 Chr.1: 19307304-19307304 on Build GRCh38
rs556666007 Chr.1: 19307199-19307199 on Build GRCh38
rs762841259 Chr.1: 19307167-19307167 on Build GRCh38
rs769076615 Chr.1: 19307267-19307267 on Build GRCh38
rs547866923 Chr.1: 19307016-19307016 on Build GRCh38
rs771154452 Chr.1: 19307123-19307125 on Build GRCh38
rs1026086129 Chr.1: 19307010-19307010 on Build GRCh38
rs2231202 Chr.1: 19307037-19307037 on Build GRCh38
rs185921220 Chr.1: 19306952-19306952 on Build GRCh38
rs765959035 Chr.1: 19307088-19307088 on Build GRCh38
rs970873962 Chr.1: 19306820-19306821 on Build GRCh38
rs757967137 Chr.1: 19306994-19306994 on Build GRCh38
rs756027825 Chr.1: 19306827-19306827 on Build GRCh38
rs746089623 Chr.1: 19307269-19307270 on Build GRCh38
rs141023615 Chr.1: 19307003-19307003 on Build GRCh38
rs574904978 Chr.1: 19307158-19307158 on Build GRCh38
rs762427018 Chr.1: 19307271-19307271 on Build GRCh38
rs113986007 Chr.1: 19307049-19307049 on Build GRCh38
rs976563268 Chr.1: 19307275-19307275 on Build GRCh38
rs1047441736 Chr.1: 19306883-19306883 on Build GRCh38
rs548864318 Chr.1: 19307060-19307060 on Build GRCh38
rs769953755 Chr.1: 19307025-19307025 on Build GRCh38
rs765982492 Chr.1: 19307280-19307280 on Build GRCh38
rs769788229 Chr.1: 19306825-19306825 on Build GRCh38
rs973413731 Chr.1: 19306829-19306829 on Build GRCh38
rs906350999 Chr.1: 19307194-19307194 on Build GRCh38
rs761375151 Chr.1: 19306957-19306957 on Build GRCh38
rs752727831 Chr.1: 19307105-19307105 on Build GRCh38
rs369846651 Chr.1: 19307042-19307042 on Build GRCh38
rs779168148 Chr.1: 19307138-19307138 on Build GRCh38

View Full Product Details