Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27224569-27225167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAD
Entrez Gene ID
790
Gene Name
carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase
Gene Aliases
CDG1Z
UniGene
Hs.377010
Gene Chromosome Location
Chr.2: 27217257-27243792 on Build GRCh38
Transcripts
NM_001306079.1 NM_004341.4 XM_005264555.3 XM_006712101.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4838437 c.1357C>G p.L453V Chr.2: 27224847-27224847 on Build GRCh38
3972192 c.1335G>A p.G445G Chr.2: 27224825-27224825 on Build GRCh38
4401764 c.1523C>T p.T508I Chr.2: 27225146-27225146 on Build GRCh38
7136448 c.1342G>T p.D448Y Chr.2: 27224832-27224832 on Build GRCh38
6467144 c.1434C>T p.G478G Chr.2: 27225057-27225057 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531016443 Chr.2: 27224604-27224604 on Build GRCh38
rs761858738 Chr.2: 27224873-27224873 on Build GRCh38
rs769331637 Chr.2: 27224832-27224832 on Build GRCh38
rs754635926 Chr.2: 27224789-27224789 on Build GRCh38
rs532828205 Chr.2: 27224718-27224718 on Build GRCh38
rs200806293 Chr.2: 27224771-27224771 on Build GRCh38
rs769007549 Chr.2: 27224860-27224860 on Build GRCh38
rs777131756 Chr.2: 27224917-27224917 on Build GRCh38
rs762949328 Chr.2: 27224984-27224984 on Build GRCh38
rs747632467 Chr.2: 27224697-27224697 on Build GRCh38
rs780879805 Chr.2: 27224909-27224909 on Build GRCh38
rs752047141 Chr.2: 27224987-27224987 on Build GRCh38
rs889031080 Chr.2: 27224633-27224633 on Build GRCh38
rs770057935 Chr.2: 27225071-27225071 on Build GRCh38
rs766809763 Chr.2: 27224895-27224895 on Build GRCh38
rs919994787 Chr.2: 27225098-27225098 on Build GRCh38
rs760066835 Chr.2: 27225099-27225099 on Build GRCh38
rs1001678249 Chr.2: 27224950-27224950 on Build GRCh38
rs759347908 Chr.2: 27224859-27224859 on Build GRCh38
rs577259994 Chr.2: 27225097-27225097 on Build GRCh38
rs201924140 Chr.2: 27224831-27224831 on Build GRCh38
rs749219227 Chr.2: 27224838-27224838 on Build GRCh38
rs369735111 Chr.2: 27224614-27224614 on Build GRCh38
rs139440330 Chr.2: 27224780-27224780 on Build GRCh38
rs1019200218 Chr.2: 27224819-27224819 on Build GRCh38
rs771984144 Chr.2: 27224962-27224962 on Build GRCh38
rs552916887 Chr.2: 27225115-27225115 on Build GRCh38
rs759940468 Chr.2: 27224989-27224989 on Build GRCh38
rs202046521 Chr.2: 27224807-27224807 on Build GRCh38
rs765340947 Chr.2: 27224874-27224874 on Build GRCh38
rs372581198 Chr.2: 27224927-27224927 on Build GRCh38
rs769622608 Chr.2: 27224706-27224706 on Build GRCh38
rs147758336 Chr.2: 27225037-27225037 on Build GRCh38
rs746719792 Chr.2: 27225063-27225063 on Build GRCh38
rs777619723 Chr.2: 27224837-27224837 on Build GRCh38
rs753980659 Chr.2: 27224765-27224765 on Build GRCh38
rs369864220 Chr.2: 27224741-27224741 on Build GRCh38
rs769614222 Chr.2: 27224653-27224653 on Build GRCh38
rs1033969711 Chr.2: 27224974-27224974 on Build GRCh38
rs758419246 Chr.2: 27225122-27225122 on Build GRCh38
rs748934301 Chr.2: 27225031-27225031 on Build GRCh38
rs745654977 Chr.2: 27224601-27224601 on Build GRCh38
rs763596085 Chr.2: 27225016-27225016 on Build GRCh38
rs755330351 Chr.2: 27224898-27224898 on Build GRCh38
rs746886288 Chr.2: 27225126-27225126 on Build GRCh38
rs756898493 Chr.2: 27225044-27225050 on Build GRCh38
rs773298563 Chr.2: 27225090-27225090 on Build GRCh38
rs568984892 Chr.2: 27224937-27224937 on Build GRCh38
rs535714254 Chr.2: 27224852-27224852 on Build GRCh38
rs564017743 Chr.2: 27224713-27224713 on Build GRCh38
rs369732907 Chr.2: 27224694-27224694 on Build GRCh38
rs895880344 Chr.2: 27224996-27224996 on Build GRCh38
rs751868593 Chr.2: 27224788-27224788 on Build GRCh38
rs539264086 Chr.2: 27224960-27224960 on Build GRCh38
rs762712513 Chr.2: 27224716-27224716 on Build GRCh38
rs751921953 Chr.2: 27224896-27224896 on Build GRCh38
rs190930020 Chr.2: 27224787-27224787 on Build GRCh38
rs778928183 Chr.2: 27224959-27224959 on Build GRCh38
rs755618660 Chr.2: 27224923-27224923 on Build GRCh38
rs756878122 Chr.2: 27225114-27225114 on Build GRCh38
rs755814746 Chr.2: 27225010-27225010 on Build GRCh38
rs199506509 Chr.2: 27225127-27225127 on Build GRCh38
rs771006887 Chr.2: 27224928-27224928 on Build GRCh38
rs552210993 Chr.2: 27224623-27224623 on Build GRCh38
rs200964128 Chr.2: 27225109-27225109 on Build GRCh38
rs745782990 Chr.2: 27225030-27225030 on Build GRCh38
rs557970896 Chr.2: 27225093-27225093 on Build GRCh38
rs773352253 Chr.2: 27224982-27224982 on Build GRCh38
rs375757875 Chr.2: 27225020-27225020 on Build GRCh38
rs764362669 Chr.2: 27224749-27224749 on Build GRCh38
rs767836058 Chr.2: 27225000-27225000 on Build GRCh38
rs942589760 Chr.2: 27224680-27224680 on Build GRCh38
rs776993320 Chr.2: 27224862-27224862 on Build GRCh38
rs753530814 Chr.2: 27225110-27225110 on Build GRCh38
rs752371232 Chr.2: 27224919-27224919 on Build GRCh38
rs754714468 Chr.2: 27225129-27225129 on Build GRCh38
rs759224415 Chr.2: 27224628-27224628 on Build GRCh38
rs776136132 Chr.2: 27224695-27224695 on Build GRCh38
rs918097614 Chr.2: 27225026-27225026 on Build GRCh38
rs190822986 Chr.2: 27224717-27224717 on Build GRCh38
rs757298956 Chr.2: 27224769-27224769 on Build GRCh38
rs770974703 Chr.2: 27225096-27225096 on Build GRCh38
rs769856396 Chr.2: 27224978-27224978 on Build GRCh38
rs779056625 Chr.2: 27225025-27225025 on Build GRCh38
rs753393523 Chr.2: 27225017-27225017 on Build GRCh38
rs1034519493 Chr.2: 27224973-27224973 on Build GRCh38
rs780755058 Chr.2: 27224824-27224824 on Build GRCh38
rs978733286 Chr.2: 27224615-27224615 on Build GRCh38
rs766938241 Chr.2: 27224986-27224986 on Build GRCh38
rs1014363759 Chr.2: 27225006-27225006 on Build GRCh38
rs770953907 Chr.2: 27224842-27224842 on Build GRCh38
rs762066850 Chr.2: 27224970-27224970 on Build GRCh38
rs780424025 Chr.2: 27224783-27224783 on Build GRCh38
rs748979465 Chr.2: 27224987-27224988 on Build GRCh38
rs779784937 Chr.2: 27225124-27225124 on Build GRCh38
rs765734732 Chr.2: 27224770-27224770 on Build GRCh38
rs141764429 Chr.2: 27225021-27225021 on Build GRCh38
rs779917471 Chr.2: 27225034-27225034 on Build GRCh38
rs763535399 Chr.2: 27224891-27224891 on Build GRCh38
rs931301550 Chr.2: 27224595-27224595 on Build GRCh38
rs778103898 Chr.2: 27225136-27225136 on Build GRCh38
rs142632262 Chr.2: 27225106-27225106 on Build GRCh38
rs779450544 Chr.2: 27225060-27225061 on Build GRCh38
rs758741978 Chr.2: 27224781-27224781 on Build GRCh38
rs182598111 Chr.2: 27224881-27224881 on Build GRCh38

View Full Product Details