Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90441552-90442148 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQGAP1
Entrez Gene ID
8826
Gene Name
IQ motif containing GTPase activating protein 1
Gene Aliases
HUMORFA01, SAR1, p195
UniGene
Hs.430551
Gene Chromosome Location
Chr.15: 90388241-90502243 on Build GRCh38
Transcripts
NM_003870.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3505401 c.727C>T p.P243S Chr.15: 90441583-90441583 on Build GRCh38
6078043 c.809T>C p.M270T Chr.15: 90441665-90441665 on Build GRCh38
3505400 c.698C>T p.P233L Chr.15: 90441554-90441554 on Build GRCh38
6464166 c.766T>G p.S256A Chr.15: 90441622-90441622 on Build GRCh38
5614301 c.757C>G p.P253A Chr.15: 90441613-90441613 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008443944 Chr.15: 90441780-90441780 on Build GRCh38
rs542898729 Chr.15: 90442052-90442052 on Build GRCh38
rs924468238 Chr.15: 90441807-90441807 on Build GRCh38
rs935869216 Chr.15: 90441834-90441834 on Build GRCh38
rs1001328636 Chr.15: 90441831-90441831 on Build GRCh38
rs57169094 Chr.15: 90441702-90441702 on Build GRCh38
rs1032457810 Chr.15: 90441844-90441844 on Build GRCh38
rs769512178 Chr.15: 90441635-90441635 on Build GRCh38
rs776760717 Chr.15: 90441705-90441705 on Build GRCh38
rs575610220 Chr.15: 90441957-90441957 on Build GRCh38
rs147782101 Chr.15: 90441584-90441584 on Build GRCh38
rs149182787 Chr.15: 90441671-90441671 on Build GRCh38
rs538812894 Chr.15: 90441891-90441891 on Build GRCh38
rs769243678 Chr.15: 90441659-90441659 on Build GRCh38
rs578053151 Chr.15: 90441820-90441820 on Build GRCh38
rs773503651 Chr.15: 90441697-90441697 on Build GRCh38
rs748800809 Chr.15: 90441643-90441643 on Build GRCh38
rs948034929 Chr.15: 90441885-90441885 on Build GRCh38
rs764142290 Chr.15: 90441720-90441720 on Build GRCh38
rs553402609 Chr.15: 90441730-90441730 on Build GRCh38
rs891412876 Chr.15: 90441754-90441756 on Build GRCh38
rs374214826 Chr.15: 90441613-90441613 on Build GRCh38
rs897980297 Chr.15: 90442012-90442012 on Build GRCh38
rs114889611 Chr.15: 90441630-90441630 on Build GRCh38
rs1009443392 Chr.15: 90441880-90441880 on Build GRCh38
rs186012149 Chr.15: 90442056-90442056 on Build GRCh38
rs767826518 Chr.15: 90441573-90441573 on Build GRCh38
rs1053787965 Chr.15: 90441852-90441852 on Build GRCh38
rs139541147 Chr.15: 90441598-90441598 on Build GRCh38
rs1003796104 Chr.15: 90441943-90441943 on Build GRCh38
rs758389834 Chr.15: 90441596-90441596 on Build GRCh38
rs977616400 Chr.15: 90441801-90441801 on Build GRCh38
rs117727888 Chr.15: 90441909-90441909 on Build GRCh38
rs911781631 Chr.15: 90441759-90441759 on Build GRCh38
rs751829471 Chr.15: 90441703-90441706 on Build GRCh38
rs750227948 Chr.15: 90441735-90441735 on Build GRCh38
rs753859958 Chr.15: 90441592-90441592 on Build GRCh38
rs201948534 Chr.15: 90441669-90441669 on Build GRCh38
rs12324924 Chr.15: 90441622-90441622 on Build GRCh38
rs377277102 Chr.15: 90442083-90442084 on Build GRCh38
rs865854946 Chr.15: 90441994-90441994 on Build GRCh38
rs893917065 Chr.15: 90441872-90441872 on Build GRCh38
rs1007196482 Chr.15: 90442086-90442086 on Build GRCh38
rs766453620 Chr.15: 90441585-90441585 on Build GRCh38
rs143921317 Chr.15: 90441673-90441673 on Build GRCh38
rs374042010 Chr.15: 90441729-90441729 on Build GRCh38
rs963086774 Chr.15: 90442065-90442065 on Build GRCh38
rs147237687 Chr.15: 90442118-90442118 on Build GRCh38
rs1045059882 Chr.15: 90441887-90441887 on Build GRCh38
rs1037055110 Chr.15: 90441972-90441972 on Build GRCh38
rs780430509 Chr.15: 90441682-90441683 on Build GRCh38
rs1033321857 Chr.15: 90441959-90441959 on Build GRCh38
rs865969402 Chr.15: 90441572-90441572 on Build GRCh38
rs760903110 Chr.15: 90441700-90441700 on Build GRCh38
rs973546738 Chr.15: 90442066-90442066 on Build GRCh38
rs956728651 Chr.15: 90441876-90441876 on Build GRCh38
rs944646691 Chr.15: 90441800-90441800 on Build GRCh38
rs752483726 Chr.15: 90441848-90441848 on Build GRCh38
rs1027381771 Chr.15: 90442059-90442059 on Build GRCh38
rs759506274 Chr.15: 90441709-90441709 on Build GRCh38
rs953119792 Chr.15: 90442068-90442068 on Build GRCh38
rs138352975 Chr.15: 90441637-90441637 on Build GRCh38
rs756357188 Chr.15: 90441983-90441984 on Build GRCh38
rs34759220 Chr.15: 90441634-90441635 on Build GRCh38

View Full Product Details