Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 93708914-93709501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM133A
Entrez Gene ID
286499
Gene Name
family with sequence similarity 133 member A
Gene Aliases
CT115
UniGene
Hs.110069
Gene Chromosome Location
Chr.X: 93673730-93712274 on Build GRCh38
Transcripts
NM_001171109.1 NM_001171110.1 NM_001171111.1 NM_173698.2 XM_006724641.3 XM_011530925.2 XM_011530926.2 XM_011530927.2 XM_011530928.1 XM_011530929.1 XM_011530931.2 XM_017029454.1 XM_017029455.1 XM_017029456.1

SNP Detail

SNP ID Chromosome Location
rs750047908 Chr.X: 93709018-93709018 on Build GRCh38
rs978494798 Chr.X: 93709393-93709393 on Build GRCh38
rs937621560 Chr.X: 93709037-93709037 on Build GRCh38
rs1020091640 Chr.X: 93709328-93709328 on Build GRCh38
rs893357684 Chr.X: 93709216-93709216 on Build GRCh38
rs150149449 Chr.X: 93709225-93709225 on Build GRCh38
rs755773424 Chr.X: 93709379-93709379 on Build GRCh38
rs748080321 Chr.X: 93709360-93709360 on Build GRCh38
rs376895079 Chr.X: 93709305-93709305 on Build GRCh38
rs4893046 Chr.X: 93709151-93709151 on Build GRCh38
rs748787393 Chr.X: 93709397-93709397 on Build GRCh38
rs778761063 Chr.X: 93709429-93709429 on Build GRCh38
rs768599301 Chr.X: 93709408-93709408 on Build GRCh38
rs995815276 Chr.X: 93709343-93709343 on Build GRCh38
rs771919183 Chr.X: 93709465-93709465 on Build GRCh38
rs779473973 Chr.X: 93709394-93709394 on Build GRCh38
rs1041921317 Chr.X: 93709438-93709438 on Build GRCh38
rs1001526727 Chr.X: 93709110-93709110 on Build GRCh38
rs185797210 Chr.X: 93709293-93709293 on Build GRCh38
rs778820429 Chr.X: 93709359-93709359 on Build GRCh38
rs940764095 Chr.X: 93708986-93708986 on Build GRCh38
rs1028911179 Chr.X: 93709430-93709430 on Build GRCh38
rs767966052 Chr.X: 93709224-93709224 on Build GRCh38
rs774692697 Chr.X: 93709003-93709003 on Build GRCh38
rs1045746400 Chr.X: 93708996-93708996 on Build GRCh38
rs1053564 Chr.X: 93709422-93709422 on Build GRCh38
rs902294179 Chr.X: 93709261-93709261 on Build GRCh38
rs959948363 Chr.X: 93709229-93709229 on Build GRCh38
rs925731998 Chr.X: 93709478-93709478 on Build GRCh38
rs747939385 Chr.X: 93709441-93709441 on Build GRCh38
rs888436887 Chr.X: 93709106-93709106 on Build GRCh38
rs1027058565 Chr.X: 93708951-93708951 on Build GRCh38
rs965690789 Chr.X: 93709341-93709341 on Build GRCh38
rs928619090 Chr.X: 93709025-93709025 on Build GRCh38
rs755822847 Chr.X: 93709074-93709074 on Build GRCh38

View Full Product Details