Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179765232-179765770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAML1
Entrez Gene ID
9794
Gene Name
mastermind like transcriptional coactivator 1
Gene Aliases
Mam-1, Mam1
UniGene
Hs.631951
Gene Chromosome Location
Chr.5: 179732850-179777286 on Build GRCh38
Transcripts
NM_014757.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5351183 c.497G>T p.G166V Chr.5: 179765396-179765396 on Build GRCh38
1066687 c.372C>T p.G124G Chr.5: 179765382-179765382 on Build GRCh38
3730159 c.562A>G p.K188E Chr.5: 179765572-179765572 on Build GRCh38
7376106 c.680G>A p.R227H Chr.5: 179765579-179765579 on Build GRCh38
5014554 c.717C>G p.N239K Chr.5: 179765727-179765727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760897990 Chr.5: 179765557-179765557 on Build GRCh38
rs111702598 Chr.5: 179765264-179765264 on Build GRCh38
rs759419227 Chr.5: 179765702-179765702 on Build GRCh38
rs765585677 Chr.5: 179765280-179765280 on Build GRCh38
rs780729908 Chr.5: 179765582-179765582 on Build GRCh38
rs776761529 Chr.5: 179765561-179765561 on Build GRCh38
rs548354642 Chr.5: 179765493-179765493 on Build GRCh38
rs137983624 Chr.5: 179765520-179765520 on Build GRCh38
rs578168072 Chr.5: 179765635-179765635 on Build GRCh38
rs766359390 Chr.5: 179765578-179765578 on Build GRCh38
rs913439165 Chr.5: 179765568-179765568 on Build GRCh38
rs781325199 Chr.5: 179765315-179765315 on Build GRCh38
rs761292293 Chr.5: 179765562-179765562 on Build GRCh38
rs148455396 Chr.5: 179765718-179765718 on Build GRCh38
rs557255881 Chr.5: 179765751-179765751 on Build GRCh38
rs763315128 Chr.5: 179765288-179765288 on Build GRCh38
rs759600308 Chr.5: 179765401-179765401 on Build GRCh38
rs374498133 Chr.5: 179765362-179765362 on Build GRCh38
rs764356260 Chr.5: 179765488-179765488 on Build GRCh38
rs113636707 Chr.5: 179765579-179765579 on Build GRCh38
rs745817360 Chr.5: 179765434-179765434 on Build GRCh38
rs368699923 Chr.5: 179765475-179765475 on Build GRCh38
rs996292105 Chr.5: 179765744-179765744 on Build GRCh38
rs754468218 Chr.5: 179765505-179765505 on Build GRCh38
rs750380237 Chr.5: 179765490-179765490 on Build GRCh38
rs113715124 Chr.5: 179765400-179765400 on Build GRCh38
rs753511638 Chr.5: 179765424-179765424 on Build GRCh38
rs768909571 Chr.5: 179765642-179765642 on Build GRCh38
rs758342415 Chr.5: 179765569-179765569 on Build GRCh38
rs770161505 Chr.5: 179765380-179765380 on Build GRCh38
rs779486597 Chr.5: 179765437-179765437 on Build GRCh38
rs767728069 Chr.5: 179765472-179765472 on Build GRCh38
rs61748798 Chr.5: 179765457-179765457 on Build GRCh38
rs150744171 Chr.5: 179765492-179765492 on Build GRCh38
rs752741971 Chr.5: 179765293-179765294 on Build GRCh38
rs141968004 Chr.5: 179765489-179765489 on Build GRCh38
rs751546910 Chr.5: 179765700-179765700 on Build GRCh38
rs772554689 Chr.5: 179765607-179765607 on Build GRCh38
rs766966774 Chr.5: 179765704-179765704 on Build GRCh38
rs985779841 Chr.5: 179765431-179765431 on Build GRCh38
rs752185395 Chr.5: 179765706-179765706 on Build GRCh38
rs139314513 Chr.5: 179765435-179765435 on Build GRCh38
rs149446543 Chr.5: 179765521-179765521 on Build GRCh38
rs780775920 Chr.5: 179765611-179765611 on Build GRCh38
rs182954629 Chr.5: 179765304-179765304 on Build GRCh38
rs777743610 Chr.5: 179765423-179765423 on Build GRCh38
rs367807730 Chr.5: 179765365-179765365 on Build GRCh38
rs758706595 Chr.5: 179765309-179765309 on Build GRCh38
rs10045384 Chr.5: 179765484-179765484 on Build GRCh38
rs764268425 Chr.5: 179765560-179765560 on Build GRCh38
rs778777699 Chr.5: 179765432-179765432 on Build GRCh38
rs774727873 Chr.5: 179765524-179765524 on Build GRCh38
rs747778003 Chr.5: 179765583-179765583 on Build GRCh38
rs746692411 Chr.5: 179765440-179765440 on Build GRCh38
rs894513597 Chr.5: 179765361-179765361 on Build GRCh38
rs750162656 Chr.5: 179765567-179765567 on Build GRCh38
rs982358554 Chr.5: 179765506-179765506 on Build GRCh38
rs747545832 Chr.5: 179765734-179765734 on Build GRCh38
rs773290253 Chr.5: 179765668-179765668 on Build GRCh38
rs950534988 Chr.5: 179765481-179765481 on Build GRCh38
rs373596613 Chr.5: 179765592-179765592 on Build GRCh38
rs974777786 Chr.5: 179765346-179765346 on Build GRCh38
rs370463621 Chr.5: 179765494-179765494 on Build GRCh38
rs891980148 Chr.5: 179765408-179765408 on Build GRCh38
rs768228572 Chr.5: 179765444-179765444 on Build GRCh38
rs766734835 Chr.5: 179765384-179765384 on Build GRCh38
rs764220320 Chr.5: 179765374-179765388 on Build GRCh38
rs558589991 Chr.5: 179765600-179765600 on Build GRCh38
rs763516997 Chr.5: 179765382-179765382 on Build GRCh38
rs773522134 Chr.5: 179765381-179765381 on Build GRCh38
rs879206269 Chr.5: 179765730-179765730 on Build GRCh38
rs199837253 Chr.5: 179765523-179765523 on Build GRCh38
rs370391455 Chr.5: 179765289-179765289 on Build GRCh38
rs142619738 Chr.5: 179765726-179765726 on Build GRCh38
rs769996954 Chr.5: 179765658-179765658 on Build GRCh38
rs377200438 Chr.5: 179765278-179765278 on Build GRCh38
rs758663524 Chr.5: 179765724-179765724 on Build GRCh38
rs151328555 Chr.5: 179765369-179765369 on Build GRCh38
rs753016394 Chr.5: 179765410-179765410 on Build GRCh38
rs146593514 Chr.5: 179765340-179765340 on Build GRCh38
rs551874527 Chr.5: 179765465-179765465 on Build GRCh38
rs146285187 Chr.5: 179765673-179765673 on Build GRCh38
rs998990754 Chr.5: 179765396-179765396 on Build GRCh38
rs756012391 Chr.5: 179765329-179765329 on Build GRCh38
rs199828835 Chr.5: 179765293-179765293 on Build GRCh38
rs200741540 Chr.5: 179765462-179765462 on Build GRCh38
rs767672825 Chr.5: 179765404-179765404 on Build GRCh38
rs771321305 Chr.5: 179765461-179765461 on Build GRCh38
rs1025289275 Chr.5: 179765468-179765468 on Build GRCh38
rs992467819 Chr.5: 179765274-179765274 on Build GRCh38
rs750974261 Chr.5: 179765281-179765281 on Build GRCh38
rs778196575 Chr.5: 179765735-179765735 on Build GRCh38
rs146382198 Chr.5: 179765417-179765417 on Build GRCh38
rs371142896 Chr.5: 179765345-179765345 on Build GRCh38
rs191317254 Chr.5: 179765526-179765526 on Build GRCh38
rs766129941 Chr.5: 179765674-179765674 on Build GRCh38
rs777540931 Chr.5: 179765717-179765717 on Build GRCh38
rs1033756028 Chr.5: 179765256-179765256 on Build GRCh38
rs904680782 Chr.5: 179765602-179765602 on Build GRCh38
rs772642568 Chr.5: 179765555-179765555 on Build GRCh38
rs780772338 Chr.5: 179765510-179765510 on Build GRCh38
rs1013518330 Chr.5: 179765466-179765466 on Build GRCh38
rs139386509 Chr.5: 179765427-179765427 on Build GRCh38
rs376219588 Chr.5: 179765339-179765339 on Build GRCh38
rs776727858 Chr.5: 179765643-179765643 on Build GRCh38
rs764968262 Chr.5: 179765566-179765566 on Build GRCh38
rs542745888 Chr.5: 179765372-179765372 on Build GRCh38
rs143467944 Chr.5: 179765623-179765623 on Build GRCh38
rs139828482 Chr.5: 179765621-179765621 on Build GRCh38
rs761942273 Chr.5: 179765653-179765653 on Build GRCh38
rs777690278 Chr.5: 179765337-179765337 on Build GRCh38
rs755737129 Chr.5: 179765707-179765707 on Build GRCh38
rs779150459 Chr.5: 179765596-179765596 on Build GRCh38
rs752963138 Chr.5: 179765316-179765316 on Build GRCh38
rs147120877 Chr.5: 179765458-179765458 on Build GRCh38
rs373516653 Chr.5: 179765528-179765528 on Build GRCh38
rs907300008 Chr.5: 179765326-179765326 on Build GRCh38
rs775726916 Chr.5: 179765556-179765556 on Build GRCh38

View Full Product Details