Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 136225898-136226451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPC7
Entrez Gene ID
57113
Gene Name
transient receptor potential cation channel subfamily C member 7
Gene Aliases
TRP7
UniGene
Hs.591263
Gene Chromosome Location
Chr.5: 136213311-136365476 on Build GRCh38
Transcripts
NM_001167576.1 NM_001167577.1 NM_020389.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
448623 c.2188A>C p.I730L Chr.5: 136226108-136226108 on Build GRCh38
5867324 c.2209G>A p.A737T Chr.5: 136226087-136226087 on Build GRCh38
4478036 c.2204C>T p.S735F Chr.5: 136226092-136226092 on Build GRCh38
1061082 c.2171G>T p.R724I Chr.5: 136226125-136226125 on Build GRCh38
1433265 c.2204C>A p.S735Y Chr.5: 136226092-136226092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775250939 Chr.5: 136225988-136225988 on Build GRCh38
rs773941734 Chr.5: 136225999-136225999 on Build GRCh38
rs576602589 Chr.5: 136226054-136226054 on Build GRCh38
rs777830189 Chr.5: 136226097-136226097 on Build GRCh38
rs755434913 Chr.5: 136225964-136225964 on Build GRCh38
rs1036134438 Chr.5: 136226425-136226425 on Build GRCh38
rs529754841 Chr.5: 136226304-136226304 on Build GRCh38
rs968598364 Chr.5: 136226286-136226286 on Build GRCh38
rs765788703 Chr.5: 136225942-136225942 on Build GRCh38
rs377352489 Chr.5: 136226253-136226253 on Build GRCh38
rs911568560 Chr.5: 136226374-136226374 on Build GRCh38
rs940398227 Chr.5: 136226378-136226378 on Build GRCh38
rs200515814 Chr.5: 136226127-136226127 on Build GRCh38
rs756658492 Chr.5: 136226271-136226273 on Build GRCh38
rs549143570 Chr.5: 136226341-136226341 on Build GRCh38
rs555647049 Chr.5: 136225935-136225957 on Build GRCh38
rs748434740 Chr.5: 136226297-136226297 on Build GRCh38
rs759911194 Chr.5: 136226284-136226284 on Build GRCh38
rs866424613 Chr.5: 136226191-136226191 on Build GRCh38
rs957116934 Chr.5: 136226219-136226219 on Build GRCh38
rs781750138 Chr.5: 136226176-136226176 on Build GRCh38
rs921876832 Chr.5: 136226312-136226312 on Build GRCh38
rs559361658 Chr.5: 136226278-136226278 on Build GRCh38
rs908408357 Chr.5: 136225981-136225981 on Build GRCh38
rs754289003 Chr.5: 136226041-136226041 on Build GRCh38
rs575279127 Chr.5: 136225940-136225940 on Build GRCh38
rs771451450 Chr.5: 136226273-136226273 on Build GRCh38
rs975093481 Chr.5: 136225920-136225920 on Build GRCh38
rs975627549 Chr.5: 136226269-136226269 on Build GRCh38
rs766978394 Chr.5: 136226030-136226030 on Build GRCh38
rs915707923 Chr.5: 136226400-136226400 on Build GRCh38
rs765646335 Chr.5: 136226296-136226296 on Build GRCh38
rs1051442246 Chr.5: 136225935-136225935 on Build GRCh38
rs191198339 Chr.5: 136226116-136226116 on Build GRCh38
rs1046803902 Chr.5: 136225977-136225977 on Build GRCh38
rs747719435 Chr.5: 136226264-136226264 on Build GRCh38
rs781503013 Chr.5: 136225987-136225987 on Build GRCh38
rs748962753 Chr.5: 136226244-136226244 on Build GRCh38
rs757312499 Chr.5: 136226109-136226109 on Build GRCh38
rs1056985557 Chr.5: 136226398-136226398 on Build GRCh38
rs948648670 Chr.5: 136226410-136226410 on Build GRCh38
rs770231934 Chr.5: 136226399-136226399 on Build GRCh38
rs541955811 Chr.5: 136226416-136226416 on Build GRCh38
rs913015809 Chr.5: 136226275-136226275 on Build GRCh38
rs779668597 Chr.5: 136226235-136226235 on Build GRCh38
rs982960415 Chr.5: 136226319-136226319 on Build GRCh38
rs374184967 Chr.5: 136226307-136226307 on Build GRCh38
rs762658003 Chr.5: 136225990-136225990 on Build GRCh38
rs543301333 Chr.5: 136225941-136225941 on Build GRCh38
rs774145396 Chr.5: 136226263-136226263 on Build GRCh38
rs1045049830 Chr.5: 136226108-136226108 on Build GRCh38
rs1057427399 Chr.5: 136225995-136225995 on Build GRCh38
rs60568559 Chr.5: 136225996-136225996 on Build GRCh38
rs746422378 Chr.5: 136226016-136226020 on Build GRCh38
rs370291152 Chr.5: 136226347-136226347 on Build GRCh38
rs1008384190 Chr.5: 136226224-136226224 on Build GRCh38
rs938400944 Chr.5: 136226348-136226348 on Build GRCh38
rs765476576 Chr.5: 136226064-136226064 on Build GRCh38
rs369028086 Chr.5: 136225938-136225960 on Build GRCh38
rs758555309 Chr.5: 136226073-136226073 on Build GRCh38
rs752986415 Chr.5: 136226070-136226070 on Build GRCh38
rs761268607 Chr.5: 136226004-136226004 on Build GRCh38
rs375971976 Chr.5: 136226229-136226229 on Build GRCh38
rs751702897 Chr.5: 136226102-136226102 on Build GRCh38

View Full Product Details