Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 132344677-132345118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACPP
Entrez Gene ID
55
Gene Name
acid phosphatase, prostate
Gene Aliases
5'-NT, ACP-3, ACP3
UniGene
Hs.433060
Gene Chromosome Location
Chr.3: 132317367-132368765 on Build GRCh38
Transcripts
NM_001099.4 NM_001134194.1 NM_001292037.1 XM_011512946.1 XM_011512947.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3587560 c.663C>T p.F221F Chr.3: 132344941-132344941 on Build GRCh38
7078844 c.738C>T p.L246L Chr.3: 132345016-132345016 on Build GRCh38
3587562 c.679G>A p.A227T Chr.3: 132344957-132344957 on Build GRCh38
5626381 c.769A>G p.R257G Chr.3: 132345047-132345047 on Build GRCh38
6894353 c.685G>C p.E229Q Chr.3: 132344963-132344963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762872737 Chr.3: 132344899-132344899 on Build GRCh38
rs769927105 Chr.3: 132345069-132345069 on Build GRCh38
rs761510622 Chr.3: 132344911-132344911 on Build GRCh38
rs774280963 Chr.3: 132345007-132345007 on Build GRCh38
rs986526814 Chr.3: 132344885-132344885 on Build GRCh38
rs552010348 Chr.3: 132344759-132344759 on Build GRCh38
rs764907124 Chr.3: 132344766-132344766 on Build GRCh38
rs748722251 Chr.3: 132344980-132344980 on Build GRCh38
rs748000374 Chr.3: 132345094-132345094 on Build GRCh38
rs972775 Chr.3: 132344799-132344799 on Build GRCh38
rs777042059 Chr.3: 132344863-132344864 on Build GRCh38
rs768680586 Chr.3: 132345075-132345075 on Build GRCh38
rs755940786 Chr.3: 132345040-132345040 on Build GRCh38
rs368755340 Chr.3: 132345079-132345079 on Build GRCh38
rs749919801 Chr.3: 132344927-132344927 on Build GRCh38
rs912307708 Chr.3: 132344887-132344887 on Build GRCh38
rs1034242748 Chr.3: 132344778-132344778 on Build GRCh38
rs753216319 Chr.3: 132345045-132345045 on Build GRCh38
rs180682200 Chr.3: 132345026-132345026 on Build GRCh38
rs936284340 Chr.3: 132345078-132345078 on Build GRCh38
rs1025794873 Chr.3: 132344703-132344703 on Build GRCh38
rs200171664 Chr.3: 132345060-132345060 on Build GRCh38
rs769721271 Chr.3: 132344987-132344987 on Build GRCh38
rs981189384 Chr.3: 132344776-132344776 on Build GRCh38
rs758893967 Chr.3: 132345050-132345050 on Build GRCh38
rs778932702 Chr.3: 132344887-132344888 on Build GRCh38
rs755490029 Chr.3: 132344930-132344930 on Build GRCh38
rs1015094324 Chr.3: 132344871-132344871 on Build GRCh38
rs778178548 Chr.3: 132345055-132345055 on Build GRCh38
rs753166910 Chr.3: 132344944-132344944 on Build GRCh38
rs747290939 Chr.3: 132344968-132344968 on Build GRCh38
rs771182185 Chr.3: 132344970-132344970 on Build GRCh38
rs1014175238 Chr.3: 132345028-132345028 on Build GRCh38
rs945776077 Chr.3: 132344876-132344876 on Build GRCh38
rs200598576 Chr.3: 132344985-132344985 on Build GRCh38
rs766999544 Chr.3: 132344912-132344912 on Build GRCh38
rs573870675 Chr.3: 132345043-132345043 on Build GRCh38
rs768627199 Chr.3: 132345004-132345004 on Build GRCh38
rs781175659 Chr.3: 132344983-132344983 on Build GRCh38
rs17856253 Chr.3: 132344954-132344954 on Build GRCh38
rs762922022 Chr.3: 132344994-132344994 on Build GRCh38
rs752603617 Chr.3: 132344847-132344847 on Build GRCh38
rs781506317 Chr.3: 132345064-132345064 on Build GRCh38
rs753633230 Chr.3: 132344722-132344722 on Build GRCh38
rs972774 Chr.3: 132345070-132345070 on Build GRCh38
rs892391852 Chr.3: 132344734-132344734 on Build GRCh38
rs749998275 Chr.3: 132345013-132345013 on Build GRCh38
rs751266647 Chr.3: 132344909-132344909 on Build GRCh38
rs775573290 Chr.3: 132344988-132344988 on Build GRCh38
rs778054006 Chr.3: 132344966-132344966 on Build GRCh38
rs767178800 Chr.3: 132345009-132345009 on Build GRCh38
rs969288048 Chr.3: 132344788-132344788 on Build GRCh38
rs989969117 Chr.3: 132344832-132344832 on Build GRCh38
rs141708902 Chr.3: 132344948-132344948 on Build GRCh38
rs931597549 Chr.3: 132345001-132345001 on Build GRCh38
rs779423026 Chr.3: 132344937-132344937 on Build GRCh38
rs1052330374 Chr.3: 132344733-132344733 on Build GRCh38
rs1034409313 Chr.3: 132344840-132344840 on Build GRCh38
rs750014567 Chr.3: 132344775-132344775 on Build GRCh38
rs528037752 Chr.3: 132344854-132344854 on Build GRCh38
rs751924131 Chr.3: 132345057-132345057 on Build GRCh38
rs746010362 Chr.3: 132345066-132345066 on Build GRCh38
rs760267787 Chr.3: 132345019-132345019 on Build GRCh38
rs775518172 Chr.3: 132344886-132344886 on Build GRCh38

View Full Product Details