Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 128027714-128028280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC4
Entrez Gene ID
64101
Gene Name
leucine rich repeat containing 4
Gene Aliases
NAG14, NGL-2
UniGene
Hs.655003
Gene Chromosome Location
Chr.7: 128027071-128032273 on Build GRCh38
Transcripts
NM_022143.4 XM_011516461.2 XM_017012505.1 NM_014390.2

Gene Symbol
SND1
Entrez Gene ID
27044
Gene Name
staphylococcal nuclease and tudor domain containing 1
Gene Aliases
TDRD11, p100
UniGene
Hs.122523
Gene Chromosome Location
Chr.7: 127652119-128092607 on Build GRCh38
Transcripts
NM_022143.4 XM_011516461.2 XM_017012505.1 NM_014390.2

SNP Detail

SNP ID Chromosome Location
rs1029197290 Chr.7: 128028017-128028017 on Build GRCh38
rs544455948 Chr.7: 128028151-128028151 on Build GRCh38
rs771869118 Chr.7: 128028073-128028073 on Build GRCh38
rs775305528 Chr.7: 128027995-128027995 on Build GRCh38
rs551448221 Chr.7: 128027783-128027783 on Build GRCh38
rs538805490 Chr.7: 128027949-128027949 on Build GRCh38
rs770621771 Chr.7: 128027824-128027824 on Build GRCh38
rs914629741 Chr.7: 128028191-128028192 on Build GRCh38
rs562386132 Chr.7: 128028154-128028154 on Build GRCh38
rs912703487 Chr.7: 128027909-128027909 on Build GRCh38
rs1016606895 Chr.7: 128028070-128028070 on Build GRCh38
rs10713009 Chr.7: 128027808-128027808 on Build GRCh38
rs775660173 Chr.7: 128027813-128027813 on Build GRCh38
rs149027335 Chr.7: 128028001-128028002 on Build GRCh38
rs555883061 Chr.7: 128027924-128027924 on Build GRCh38
rs781110176 Chr.7: 128027925-128027925 on Build GRCh38
rs987636351 Chr.7: 128027893-128027893 on Build GRCh38
rs901834724 Chr.7: 128028206-128028206 on Build GRCh38
rs1051078862 Chr.7: 128028214-128028214 on Build GRCh38
rs572738000 Chr.7: 128027952-128027952 on Build GRCh38
rs893763831 Chr.7: 128028098-128028098 on Build GRCh38
rs1041750548 Chr.7: 128028196-128028196 on Build GRCh38
rs887231079 Chr.7: 128027811-128027811 on Build GRCh38
rs921511195 Chr.7: 128028243-128028243 on Build GRCh38
rs957673798 Chr.7: 128027852-128027852 on Build GRCh38
rs1011244329 Chr.7: 128028099-128028099 on Build GRCh38
rs946096655 Chr.7: 128028192-128028192 on Build GRCh38
rs966793178 Chr.7: 128028115-128028115 on Build GRCh38
rs913638546 Chr.7: 128027870-128027870 on Build GRCh38
rs1006599782 Chr.7: 128028031-128028031 on Build GRCh38
rs553829060 Chr.7: 128027950-128027950 on Build GRCh38
rs989195952 Chr.7: 128027865-128027865 on Build GRCh38
rs909593204 Chr.7: 128027843-128027843 on Build GRCh38
rs966617302 Chr.7: 128027781-128027781 on Build GRCh38
rs972634264 Chr.7: 128027872-128027872 on Build GRCh38
rs3808058 Chr.7: 128028018-128028018 on Build GRCh38
rs987216132 Chr.7: 128028118-128028118 on Build GRCh38
rs567096978 Chr.7: 128027810-128027810 on Build GRCh38
rs893144182 Chr.7: 128027975-128027975 on Build GRCh38
rs35973393 Chr.7: 128028048-128028048 on Build GRCh38
rs567981545 Chr.7: 128027827-128027827 on Build GRCh38
rs1024769073 Chr.7: 128027819-128027819 on Build GRCh38
rs949939551 Chr.7: 128027899-128027899 on Build GRCh38
rs995546441 Chr.7: 128027774-128027774 on Build GRCh38
rs965506798 Chr.7: 128028166-128028166 on Build GRCh38
rs774662670 Chr.7: 128028230-128028230 on Build GRCh38
rs906341845 Chr.7: 128028021-128028021 on Build GRCh38
rs191496565 Chr.7: 128027812-128027812 on Build GRCh38
rs975584150 Chr.7: 128028167-128028167 on Build GRCh38
rs918483267 Chr.7: 128027880-128027880 on Build GRCh38
rs937118004 Chr.7: 128028180-128028180 on Build GRCh38
rs940085653 Chr.7: 128028010-128028010 on Build GRCh38
rs981755190 Chr.7: 128028174-128028174 on Build GRCh38
rs1048428267 Chr.7: 128027982-128027982 on Build GRCh38
rs555114737 Chr.7: 128028040-128028040 on Build GRCh38
rs868717122 Chr.7: 128028085-128028085 on Build GRCh38
rs1029558910 Chr.7: 128027816-128027816 on Build GRCh38
rs1002060772 Chr.7: 128028063-128028063 on Build GRCh38
rs933628405 Chr.7: 128028208-128028208 on Build GRCh38
rs538482041 Chr.7: 128027892-128027892 on Build GRCh38
rs35189586 Chr.7: 128027788-128027789 on Build GRCh38
rs927185982 Chr.7: 128028179-128028180 on Build GRCh38
rs931541031 Chr.7: 128028245-128028245 on Build GRCh38
rs1039948652 Chr.7: 128027748-128027748 on Build GRCh38
rs962368529 Chr.7: 128028102-128028102 on Build GRCh38
rs571605391 Chr.7: 128027923-128027923 on Build GRCh38
rs866262161 Chr.7: 128027931-128027931 on Build GRCh38
rs879141553 Chr.7: 128027738-128027738 on Build GRCh38
rs563693011 Chr.7: 128028136-128028138 on Build GRCh38
rs541663547 Chr.7: 128028127-128028127 on Build GRCh38
rs745487067 Chr.7: 128027939-128027939 on Build GRCh38
rs1020585535 Chr.7: 128028109-128028109 on Build GRCh38
rs773976020 Chr.7: 128027744-128027744 on Build GRCh38
rs549564149 Chr.7: 128027822-128027822 on Build GRCh38
rs990240146 Chr.7: 128028180-128028184 on Build GRCh38
rs1048942398 Chr.7: 128027805-128027805 on Build GRCh38
rs998164276 Chr.7: 128028014-128028016 on Build GRCh38
rs887109432 Chr.7: 128027999-128027999 on Build GRCh38
rs899822169 Chr.7: 128027765-128027765 on Build GRCh38
rs1033614531 Chr.7: 128027838-128027838 on Build GRCh38
rs540621923 Chr.7: 128028131-128028131 on Build GRCh38
rs966447911 Chr.7: 128027883-128027883 on Build GRCh38
rs556290745 Chr.7: 128027912-128027912 on Build GRCh38

View Full Product Details