Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75942452-75942960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOX1
Entrez Gene ID
51
Gene Name
acyl-CoA oxidase 1
Gene Aliases
ACOX, PALMCOX, SCOX
UniGene
Hs.464137
Gene Chromosome Location
Chr.17: 75941507-75979434 on Build GRCh38
Transcripts
NM_001185039.1 NM_004035.6 NM_007292.5 XM_011524868.2 XM_011524869.2

SNP Detail

SNP ID Chromosome Location
rs964649647 Chr.17: 75942742-75942742 on Build GRCh38
rs150647165 Chr.17: 75942489-75942489 on Build GRCh38
rs567320717 Chr.17: 75942595-75942595 on Build GRCh38
rs1013260199 Chr.17: 75942627-75942633 on Build GRCh38
rs907465838 Chr.17: 75942661-75942661 on Build GRCh38
rs941776798 Chr.17: 75942558-75942558 on Build GRCh38
rs998998120 Chr.17: 75942677-75942677 on Build GRCh38
rs372407045 Chr.17: 75942630-75942630 on Build GRCh38
rs992985966 Chr.17: 75942776-75942776 on Build GRCh38
rs910947165 Chr.17: 75942882-75942882 on Build GRCh38
rs897051989 Chr.17: 75942637-75942637 on Build GRCh38
rs536938567 Chr.17: 75942727-75942727 on Build GRCh38
rs894903652 Chr.17: 75942879-75942879 on Build GRCh38
rs559485748 Chr.17: 75942833-75942833 on Build GRCh38
rs577207820 Chr.17: 75942709-75942709 on Build GRCh38
rs1031818724 Chr.17: 75942678-75942678 on Build GRCh38
rs1024526692 Chr.17: 75942655-75942655 on Build GRCh38
rs563313208 Chr.17: 75942542-75942542 on Build GRCh38
rs889601629 Chr.17: 75942532-75942532 on Build GRCh38
rs1041290804 Chr.17: 75942624-75942624 on Build GRCh38
rs1020872917 Chr.17: 75942804-75942804 on Build GRCh38
rs1034467110 Chr.17: 75942747-75942747 on Build GRCh38
rs979015012 Chr.17: 75942829-75942829 on Build GRCh38
rs777677912 Chr.17: 75942904-75942904 on Build GRCh38
rs112541815 Chr.17: 75942498-75942498 on Build GRCh38
rs1047573165 Chr.17: 75942509-75942509 on Build GRCh38
rs148585794 Chr.17: 75942936-75942936 on Build GRCh38
rs867359154 Chr.17: 75942602-75942602 on Build GRCh38
rs16968333 Chr.17: 75942805-75942805 on Build GRCh38
rs558901332 Chr.17: 75942816-75942816 on Build GRCh38
rs957725645 Chr.17: 75942870-75942870 on Build GRCh38
rs796084410 Chr.17: 75942918-75942918 on Build GRCh38
rs994089593 Chr.17: 75942700-75942700 on Build GRCh38
rs368770598 Chr.17: 75942503-75942503 on Build GRCh38
rs144701048 Chr.17: 75942839-75942839 on Build GRCh38
rs887630690 Chr.17: 75942548-75942548 on Build GRCh38
rs546147725 Chr.17: 75942739-75942739 on Build GRCh38
rs909038775 Chr.17: 75942813-75942813 on Build GRCh38
rs556930840 Chr.17: 75942738-75942738 on Build GRCh38
rs149774605 Chr.17: 75942760-75942760 on Build GRCh38
rs2608880 Chr.17: 75942876-75942876 on Build GRCh38
rs943805838 Chr.17: 75942890-75942890 on Build GRCh38
rs1055316575 Chr.17: 75942585-75942585 on Build GRCh38
rs764816387 Chr.17: 75942798-75942798 on Build GRCh38
rs759138629 Chr.17: 75942780-75942780 on Build GRCh38

View Full Product Details