Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 55007086-55007634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSND
Entrez Gene ID
7809
Gene Name
barttin CLCNK type accessory beta subunit
Gene Aliases
BART, DFNB73
UniGene
Hs.151291
Gene Chromosome Location
Chr.1: 54998944-55008792 on Build GRCh38
Transcripts
NM_057176.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4009012 c.484G>A p.V162M Chr.1: 55007208-55007208 on Build GRCh38
1639921 c.422T>C p.L141P Chr.1: 55007146-55007146 on Build GRCh38
5992272 c.478G>A p.A160T Chr.1: 55007202-55007202 on Build GRCh38
4009011 c.456C>T p.G152G Chr.1: 55007180-55007180 on Build GRCh38
1627125 c.548+6T>C p.? Chr.1: 55007278-55007278 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs979847754 Chr.1: 55007298-55007298 on Build GRCh38
rs968386063 Chr.1: 55007612-55007612 on Build GRCh38
rs575448589 Chr.1: 55007492-55007492 on Build GRCh38
rs889865679 Chr.1: 55007443-55007443 on Build GRCh38
rs202128855 Chr.1: 55007181-55007181 on Build GRCh38
rs774559680 Chr.1: 55007238-55007238 on Build GRCh38
rs777582230 Chr.1: 55007203-55007203 on Build GRCh38
rs992673810 Chr.1: 55007367-55007367 on Build GRCh38
rs762620750 Chr.1: 55007156-55007156 on Build GRCh38
rs769608234 Chr.1: 55007149-55007149 on Build GRCh38
rs1049867679 Chr.1: 55007438-55007438 on Build GRCh38
rs77339832 Chr.1: 55007574-55007574 on Build GRCh38
rs746540356 Chr.1: 55007139-55007139 on Build GRCh38
rs143711308 Chr.1: 55007251-55007251 on Build GRCh38
rs1056638175 Chr.1: 55007546-55007546 on Build GRCh38
rs894066625 Chr.1: 55007344-55007344 on Build GRCh38
rs375605563 Chr.1: 55007454-55007454 on Build GRCh38
rs769806934 Chr.1: 55007224-55007224 on Build GRCh38
rs749600956 Chr.1: 55007133-55007133 on Build GRCh38
rs758111009 Chr.1: 55007135-55007135 on Build GRCh38
rs749127391 Chr.1: 55007231-55007231 on Build GRCh38
rs775406885 Chr.1: 55007153-55007153 on Build GRCh38
rs750027126 Chr.1: 55007271-55007271 on Build GRCh38
rs779959793 Chr.1: 55007354-55007354 on Build GRCh38
rs1037038332 Chr.1: 55007179-55007179 on Build GRCh38
rs780910610 Chr.1: 55007208-55007208 on Build GRCh38
rs6687801 Chr.1: 55007370-55007370 on Build GRCh38
rs529780817 Chr.1: 55007184-55007184 on Build GRCh38
rs1010508291 Chr.1: 55007561-55007561 on Build GRCh38
rs891923096 Chr.1: 55007506-55007506 on Build GRCh38
rs369882063 Chr.1: 55007282-55007282 on Build GRCh38
rs774504119 Chr.1: 55007180-55007180 on Build GRCh38
rs138974602 Chr.1: 55007183-55007183 on Build GRCh38
rs750527043 Chr.1: 55007232-55007233 on Build GRCh38
rs755527408 Chr.1: 55007287-55007287 on Build GRCh38
rs761777830 Chr.1: 55007112-55007113 on Build GRCh38
rs984031117 Chr.1: 55007525-55007525 on Build GRCh38
rs540317305 Chr.1: 55007189-55007189 on Build GRCh38
rs927034309 Chr.1: 55007299-55007299 on Build GRCh38
rs939110347 Chr.1: 55007445-55007445 on Build GRCh38
rs749110043 Chr.1: 55007315-55007315 on Build GRCh38
rs768951289 Chr.1: 55007170-55007170 on Build GRCh38
rs375551000 Chr.1: 55007270-55007270 on Build GRCh38
rs532738229 Chr.1: 55007240-55007240 on Build GRCh38
rs547462190 Chr.1: 55007382-55007382 on Build GRCh38
rs745871123 Chr.1: 55007148-55007148 on Build GRCh38
rs780533402 Chr.1: 55007129-55007129 on Build GRCh38
rs760632724 Chr.1: 55007258-55007258 on Build GRCh38
rs201991745 Chr.1: 55007250-55007250 on Build GRCh38
rs770508418 Chr.1: 55007140-55007140 on Build GRCh38
rs920560793 Chr.1: 55007501-55007501 on Build GRCh38
rs201370435 Chr.1: 55007304-55007304 on Build GRCh38
rs1033303952 Chr.1: 55007294-55007294 on Build GRCh38
rs908436037 Chr.1: 55007526-55007526 on Build GRCh38
rs367549117 Chr.1: 55007110-55007110 on Build GRCh38
rs907662035 Chr.1: 55007369-55007369 on Build GRCh38
rs765766484 Chr.1: 55007273-55007273 on Build GRCh38
rs765135576 Chr.1: 55007174-55007174 on Build GRCh38
rs757283527 Chr.1: 55007109-55007109 on Build GRCh38
rs758169410 Chr.1: 55007346-55007346 on Build GRCh38
rs754262236 Chr.1: 55007190-55007190 on Build GRCh38
rs201387711 Chr.1: 55007117-55007117 on Build GRCh38
rs558585133 Chr.1: 55007481-55007481 on Build GRCh38
rs560696544 Chr.1: 55007226-55007226 on Build GRCh38
rs761300365 Chr.1: 55007185-55007185 on Build GRCh38
rs763459351 Chr.1: 55007302-55007302 on Build GRCh38
rs745352422 Chr.1: 55007219-55007219 on Build GRCh38
rs757022807 Chr.1: 55007276-55007276 on Build GRCh38
rs566596233 Chr.1: 55007406-55007406 on Build GRCh38
rs777252686 Chr.1: 55007136-55007136 on Build GRCh38
rs776717215 Chr.1: 55007141-55007141 on Build GRCh38
rs755249026 Chr.1: 55007192-55007192 on Build GRCh38
rs146647907 Chr.1: 55007126-55007126 on Build GRCh38
rs548152750 Chr.1: 55007335-55007335 on Build GRCh38
rs538816952 Chr.1: 55007433-55007433 on Build GRCh38
rs568229262 Chr.1: 55007347-55007347 on Build GRCh38
rs1037994469 Chr.1: 55007394-55007394 on Build GRCh38
rs768275152 Chr.1: 55007232-55007232 on Build GRCh38
rs1041217563 Chr.1: 55007483-55007483 on Build GRCh38
rs756969977 Chr.1: 55007207-55007207 on Build GRCh38
rs369618892 Chr.1: 55007206-55007206 on Build GRCh38
rs749976995 Chr.1: 55007286-55007286 on Build GRCh38
rs867203115 Chr.1: 55007113-55007113 on Build GRCh38

View Full Product Details