Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17954537-17955076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNN1
Entrez Gene ID
3780
Gene Name
potassium calcium-activated channel subfamily N member 1
Gene Aliases
KCa2.1, SK1, SKCA1, hSK1
UniGene
Hs.158173
Gene Chromosome Location
Chr.19: 17951302-17999121 on Build GRCh38
Transcripts
NM_002248.4 XM_011528005.2

SNP Detail

SNP ID Chromosome Location
rs868773304 Chr.19: 17955037-17955037 on Build GRCh38
rs745996842 Chr.19: 17954572-17954572 on Build GRCh38
rs886165629 Chr.19: 17954795-17954795 on Build GRCh38
rs575267700 Chr.19: 17955023-17955023 on Build GRCh38
rs939997153 Chr.19: 17954692-17954692 on Build GRCh38
rs1049324415 Chr.19: 17955002-17955002 on Build GRCh38
rs866158803 Chr.19: 17954916-17954916 on Build GRCh38
rs1021107080 Chr.19: 17954644-17954644 on Build GRCh38
rs113416446 Chr.19: 17954893-17954893 on Build GRCh38
rs557634078 Chr.19: 17954858-17954858 on Build GRCh38
rs965897278 Chr.19: 17954817-17954817 on Build GRCh38
rs559542527 Chr.19: 17954899-17954899 on Build GRCh38
rs1048722580 Chr.19: 17954785-17954785 on Build GRCh38
rs980680883 Chr.19: 17954969-17954969 on Build GRCh38
rs1048884830 Chr.19: 17954755-17954755 on Build GRCh38
rs1002091136 Chr.19: 17954775-17954775 on Build GRCh38
rs987021703 Chr.19: 17954675-17954675 on Build GRCh38
rs1044643542 Chr.19: 17954739-17954739 on Build GRCh38
rs190172272 Chr.19: 17954659-17954659 on Build GRCh38
rs950291219 Chr.19: 17954732-17954732 on Build GRCh38
rs375489168 Chr.19: 17954992-17954992 on Build GRCh38
rs974675986 Chr.19: 17954651-17954651 on Build GRCh38
rs555256651 Chr.19: 17954702-17954702 on Build GRCh38
rs967963181 Chr.19: 17954866-17954866 on Build GRCh38
rs1009636205 Chr.19: 17954854-17954854 on Build GRCh38
rs1019137706 Chr.19: 17954814-17954814 on Build GRCh38
rs917357198 Chr.19: 17954927-17954927 on Build GRCh38
rs927516600 Chr.19: 17954991-17954991 on Build GRCh38
rs926478518 Chr.19: 17954993-17954993 on Build GRCh38
rs34300645 Chr.19: 17955037-17955038 on Build GRCh38
rs896178545 Chr.19: 17954851-17954851 on Build GRCh38
rs12104302 Chr.19: 17954995-17954995 on Build GRCh38
rs919899948 Chr.19: 17954963-17954963 on Build GRCh38
rs781107544 Chr.19: 17954563-17954563 on Build GRCh38
rs577503875 Chr.19: 17954890-17954890 on Build GRCh38
rs370871622 Chr.19: 17954894-17954894 on Build GRCh38
rs1037147877 Chr.19: 17954694-17954694 on Build GRCh38
rs755995351 Chr.19: 17954598-17954598 on Build GRCh38
rs1028873839 Chr.19: 17954853-17954853 on Build GRCh38
rs1004617669 Chr.19: 17954806-17954806 on Build GRCh38
rs960623827 Chr.19: 17954561-17954561 on Build GRCh38
rs999092590 Chr.19: 17955033-17955033 on Build GRCh38
rs779981997 Chr.19: 17954613-17954613 on Build GRCh38
rs968202812 Chr.19: 17954868-17954868 on Build GRCh38
rs544106964 Chr.19: 17954716-17954716 on Build GRCh38
rs765822562 Chr.19: 17954792-17954792 on Build GRCh38
rs184861031 Chr.19: 17954962-17954962 on Build GRCh38
rs749020652 Chr.19: 17954950-17954950 on Build GRCh38
rs1000581477 Chr.19: 17954873-17954873 on Build GRCh38
rs919997362 Chr.19: 17954706-17954706 on Build GRCh38
rs116528247 Chr.19: 17954677-17954677 on Build GRCh38
rs912793548 Chr.19: 17954918-17954918 on Build GRCh38
rs540859726 Chr.19: 17954904-17954904 on Build GRCh38
rs561745952 Chr.19: 17954632-17954632 on Build GRCh38
rs968045936 Chr.19: 17954657-17954657 on Build GRCh38
rs796319823 Chr.19: 17954575-17954575 on Build GRCh38
rs192780985 Chr.19: 17954660-17954660 on Build GRCh38
rs575063272 Chr.19: 17954713-17954713 on Build GRCh38
rs905001636 Chr.19: 17954762-17954762 on Build GRCh38
rs930517362 Chr.19: 17954746-17954746 on Build GRCh38
rs996450017 Chr.19: 17954850-17954850 on Build GRCh38
rs538395964 Chr.19: 17954647-17954647 on Build GRCh38
rs994422378 Chr.19: 17954761-17954761 on Build GRCh38
rs1056288299 Chr.19: 17954776-17954776 on Build GRCh38
rs961436155 Chr.19: 17954941-17954941 on Build GRCh38
rs925995036 Chr.19: 17954670-17954670 on Build GRCh38
rs886093440 Chr.19: 17955011-17955011 on Build GRCh38
rs931406293 Chr.19: 17954975-17954975 on Build GRCh38

View Full Product Details