Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151161691-151162103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYSMD1
Entrez Gene ID
388695
Gene Name
LysM domain containing 1
Gene Aliases
SB145
UniGene
Hs.591482
Gene Chromosome Location
Chr.1: 151148496-151165948 on Build GRCh38
Transcripts
NM_001136543.1 NM_212551.4 NM_001204848.1

Gene Symbol
TNFAIP8L2-SCNM1
Entrez Gene ID
100534012
Gene Name
TNFAIP8L2-SCNM1 readthrough
Gene Aliases
-
UniGene
Hs.732060
Gene Chromosome Location
Chr.1: 151156629-151170297 on Build GRCh38
Transcripts
NM_001136543.1 NM_212551.4 NM_001204848.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
895718 c.260T>C p.L87P Chr.1: 151162021-151162021 on Build GRCh38
4022141 c.534A>G p.K178K Chr.1: 151161747-151161747 on Build GRCh38
895717 c.388G>A p.A130T Chr.1: 151161893-151161893 on Build GRCh38
6705349 c.61C>T p.R21C Chr.1: 151162076-151162076 on Build GRCh38
6579618 c.322G>A p.D108N Chr.1: 151161815-151161815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777240571 Chr.1: 151162075-151162075 on Build GRCh38
rs879033634 Chr.1: 151161858-151161858 on Build GRCh38
rs754782146 Chr.1: 151161841-151161841 on Build GRCh38
rs200735103 Chr.1: 151162062-151162062 on Build GRCh38
rs752612746 Chr.1: 151161865-151161865 on Build GRCh38
rs74888559 Chr.1: 151162059-151162059 on Build GRCh38
rs1035101785 Chr.1: 151161820-151161820 on Build GRCh38
rs745340535 Chr.1: 151161746-151161746 on Build GRCh38
rs77292984 Chr.1: 151161886-151161886 on Build GRCh38
rs759933455 Chr.1: 151162016-151162017 on Build GRCh38
rs141690686 Chr.1: 151162028-151162028 on Build GRCh38
rs757987576 Chr.1: 151162055-151162055 on Build GRCh38
rs761171483 Chr.1: 151161939-151161939 on Build GRCh38
rs111870485 Chr.1: 151161971-151161971 on Build GRCh38
rs777820791 Chr.1: 151161877-151161877 on Build GRCh38
rs370583370 Chr.1: 151161983-151161983 on Build GRCh38
rs768117541 Chr.1: 151162076-151162076 on Build GRCh38
rs149420524 Chr.1: 151161844-151161844 on Build GRCh38
rs772366935 Chr.1: 151161928-151161928 on Build GRCh38
rs755999714 Chr.1: 151161867-151161867 on Build GRCh38
rs138522958 Chr.1: 151161845-151161845 on Build GRCh38
rs750716325 Chr.1: 151161838-151161838 on Build GRCh38
rs903135613 Chr.1: 151161881-151161881 on Build GRCh38
rs773508537 Chr.1: 151161969-151161969 on Build GRCh38
rs192433758 Chr.1: 151161817-151161817 on Build GRCh38
rs368807734 Chr.1: 151162035-151162035 on Build GRCh38
rs753802519 Chr.1: 151162026-151162026 on Build GRCh38
rs752420143 Chr.1: 151161984-151161984 on Build GRCh38
rs942357118 Chr.1: 151162034-151162034 on Build GRCh38
rs774036336 Chr.1: 151161714-151161714 on Build GRCh38
rs755982668 Chr.1: 151162003-151162003 on Build GRCh38
rs774467416 Chr.1: 151162004-151162005 on Build GRCh38
rs587699753 Chr.1: 151161825-151161825 on Build GRCh38
rs748376465 Chr.1: 151161901-151161907 on Build GRCh38
rs764172759 Chr.1: 151161816-151161816 on Build GRCh38
rs769031385 Chr.1: 151161944-151161944 on Build GRCh38
rs761102404 Chr.1: 151161802-151161802 on Build GRCh38
rs935531417 Chr.1: 151162067-151162067 on Build GRCh38
rs772454142 Chr.1: 151161786-151161786 on Build GRCh38
rs761955171 Chr.1: 151161967-151161967 on Build GRCh38
rs748769586 Chr.1: 151161880-151161880 on Build GRCh38
rs75993764 Chr.1: 151161985-151161985 on Build GRCh38
rs759860606 Chr.1: 151161762-151161762 on Build GRCh38
rs587674416 Chr.1: 151161911-151161911 on Build GRCh38
rs763903092 Chr.1: 151162011-151162011 on Build GRCh38
rs777245106 Chr.1: 151161963-151161963 on Build GRCh38
rs770989273 Chr.1: 151161907-151161909 on Build GRCh38
rs748958647 Chr.1: 151161827-151161827 on Build GRCh38
rs778411362 Chr.1: 151162029-151162029 on Build GRCh38
rs1013300211 Chr.1: 151162037-151162037 on Build GRCh38
rs778493466 Chr.1: 151161887-151161887 on Build GRCh38
rs1043262450 Chr.1: 151161872-151161872 on Build GRCh38
rs756705505 Chr.1: 151162027-151162027 on Build GRCh38
rs776019822 Chr.1: 151161788-151161788 on Build GRCh38
rs775726153 Chr.1: 151161934-151161934 on Build GRCh38

View Full Product Details