Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 30230492-30230988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGEB3
Entrez Gene ID
4114
Gene Name
MAGE family member B3
Gene Aliases
CT3.5
UniGene
Hs.113290
Gene Chromosome Location
Chr.X: 30230436-30237493 on Build GRCh38
Transcripts
NM_002365.4 XM_011545514.2

SNP Detail

SNP ID Chromosome Location
rs1030082834 Chr.X: 30230645-30230645 on Build GRCh38
rs748895014 Chr.X: 30230649-30230649 on Build GRCh38
rs746202912 Chr.X: 30230635-30230635 on Build GRCh38
rs937883559 Chr.X: 30230752-30230752 on Build GRCh38
rs747925826 Chr.X: 30230753-30230753 on Build GRCh38
rs946571026 Chr.X: 30230842-30230842 on Build GRCh38
rs764288461 Chr.X: 30230890-30230890 on Build GRCh38
rs148337997 Chr.X: 30230517-30230518 on Build GRCh38
rs981828305 Chr.X: 30230741-30230741 on Build GRCh38
rs868311587 Chr.X: 30230518-30230518 on Build GRCh38
rs939673546 Chr.X: 30230668-30230668 on Build GRCh38
rs190106943 Chr.X: 30230844-30230844 on Build GRCh38
rs777326641 Chr.X: 30230580-30230580 on Build GRCh38
rs927851013 Chr.X: 30230750-30230750 on Build GRCh38
rs755412838 Chr.X: 30230514-30230515 on Build GRCh38
rs778907385 Chr.X: 30230665-30230666 on Build GRCh38
rs181678853 Chr.X: 30230869-30230869 on Build GRCh38
rs890347178 Chr.X: 30230652-30230652 on Build GRCh38
rs903298237 Chr.X: 30230856-30230856 on Build GRCh38
rs962107373 Chr.X: 30230658-30230658 on Build GRCh38
rs565255927 Chr.X: 30230823-30230823 on Build GRCh38
rs762052815 Chr.X: 30230967-30230967 on Build GRCh38
rs1007497620 Chr.X: 30230737-30230737 on Build GRCh38
rs769702725 Chr.X: 30230864-30230864 on Build GRCh38
rs185979703 Chr.X: 30230886-30230886 on Build GRCh38
rs1017430879 Chr.X: 30230792-30230792 on Build GRCh38
rs1055055991 Chr.X: 30230765-30230765 on Build GRCh38
rs754670080 Chr.X: 30230651-30230651 on Build GRCh38
rs901181862 Chr.X: 30230948-30230948 on Build GRCh38
rs55752735 Chr.X: 30230957-30230957 on Build GRCh38
rs908326418 Chr.X: 30230637-30230637 on Build GRCh38
rs772983013 Chr.X: 30230843-30230843 on Build GRCh38
rs998156680 Chr.X: 30230578-30230578 on Build GRCh38
rs897141226 Chr.X: 30230529-30230529 on Build GRCh38

View Full Product Details