Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50931357-50931897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL7A
Entrez Gene ID
25840
Gene Name
methyltransferase like 7A
Gene Aliases
AAM-B
UniGene
Hs.711113
Gene Chromosome Location
Chr.12: 50923811-50932517 on Build GRCh38
Transcripts
NM_014033.3 XM_006719332.1

SNP Detail

SNP ID Chromosome Location
rs3180989 Chr.12: 50931636-50931636 on Build GRCh38
rs918603041 Chr.12: 50931435-50931435 on Build GRCh38
rs35013187 Chr.12: 50931546-50931547 on Build GRCh38
rs879288414 Chr.12: 50931436-50931436 on Build GRCh38
rs3180343 Chr.12: 50931454-50931454 on Build GRCh38
rs762022573 Chr.12: 50931701-50931711 on Build GRCh38
rs939784486 Chr.12: 50931735-50931735 on Build GRCh38
rs3180345 Chr.12: 50931504-50931504 on Build GRCh38
rs1041861133 Chr.12: 50931736-50931736 on Build GRCh38
rs961831386 Chr.12: 50931580-50931580 on Build GRCh38
rs1034106442 Chr.12: 50931546-50931546 on Build GRCh38
rs1046108516 Chr.12: 50931743-50931743 on Build GRCh38
rs3180985 Chr.12: 50931455-50931455 on Build GRCh38
rs1003129550 Chr.12: 50931461-50931461 on Build GRCh38
rs565085447 Chr.12: 50931645-50931645 on Build GRCh38
rs377548291 Chr.12: 50931411-50931411 on Build GRCh38
rs319940 Chr.12: 50931515-50931515 on Build GRCh38
rs3209374 Chr.12: 50931806-50931806 on Build GRCh38
rs967878957 Chr.12: 50931627-50931627 on Build GRCh38
rs965464126 Chr.12: 50931424-50931425 on Build GRCh38
rs78329969 Chr.12: 50931715-50931715 on Build GRCh38
rs3180987 Chr.12: 50931607-50931607 on Build GRCh38
rs995170185 Chr.12: 50931420-50931420 on Build GRCh38
rs3209373 Chr.12: 50931803-50931803 on Build GRCh38
rs561292157 Chr.12: 50931448-50931448 on Build GRCh38
rs927568106 Chr.12: 50931445-50931445 on Build GRCh38
rs377657191 Chr.12: 50931671-50931671 on Build GRCh38
rs370491373 Chr.12: 50931684-50931684 on Build GRCh38
rs925763948 Chr.12: 50931739-50931740 on Build GRCh38
rs3209369 Chr.12: 50931794-50931794 on Build GRCh38
rs767344349 Chr.12: 50931660-50931660 on Build GRCh38
rs1022061478 Chr.12: 50931858-50931858 on Build GRCh38
rs977507792 Chr.12: 50931642-50931642 on Build GRCh38
rs539284466 Chr.12: 50931799-50931799 on Build GRCh38
rs1015520661 Chr.12: 50931447-50931447 on Build GRCh38
rs899514137 Chr.12: 50931408-50931408 on Build GRCh38
rs71089713 Chr.12: 50931547-50931548 on Build GRCh38
rs897673521 Chr.12: 50931848-50931848 on Build GRCh38
rs960660347 Chr.12: 50931576-50931576 on Build GRCh38
rs117919526 Chr.12: 50931807-50931807 on Build GRCh38
rs546985900 Chr.12: 50931707-50931707 on Build GRCh38
rs765524209 Chr.12: 50931456-50931456 on Build GRCh38
rs319941 Chr.12: 50931646-50931646 on Build GRCh38
rs972826066 Chr.12: 50931425-50931425 on Build GRCh38
rs3180347 Chr.12: 50931754-50931754 on Build GRCh38
rs1027805481 Chr.12: 50931496-50931496 on Build GRCh38
rs187621980 Chr.12: 50931659-50931659 on Build GRCh38
rs958941720 Chr.12: 50931492-50931492 on Build GRCh38
rs907377790 Chr.12: 50931759-50931759 on Build GRCh38
rs917434157 Chr.12: 50931672-50931672 on Build GRCh38
rs3209370 Chr.12: 50931795-50931795 on Build GRCh38
rs3180986 Chr.12: 50931596-50931596 on Build GRCh38
rs886883454 Chr.12: 50931442-50931442 on Build GRCh38
rs945802913 Chr.12: 50931474-50931474 on Build GRCh38
rs886750253 Chr.12: 50931730-50931730 on Build GRCh38
rs958965881 Chr.12: 50931666-50931666 on Build GRCh38
rs3180991 Chr.12: 50931657-50931657 on Build GRCh38
rs3209375 Chr.12: 50931808-50931808 on Build GRCh38
rs185500414 Chr.12: 50931643-50931643 on Build GRCh38
rs550991099 Chr.12: 50931648-50931648 on Build GRCh38
rs3209367 Chr.12: 50931765-50931765 on Build GRCh38
rs1048059498 Chr.12: 50931421-50931421 on Build GRCh38
rs536612179 Chr.12: 50931706-50931706 on Build GRCh38
rs3180346 Chr.12: 50931523-50931523 on Build GRCh38
rs543664528 Chr.12: 50931553-50931553 on Build GRCh38
rs1040241220 Chr.12: 50931814-50931814 on Build GRCh38
rs914564920 Chr.12: 50931571-50931571 on Build GRCh38
rs992158529 Chr.12: 50931579-50931579 on Build GRCh38
rs1034566123 Chr.12: 50931462-50931462 on Build GRCh38
rs1018758836 Chr.12: 50931567-50931567 on Build GRCh38
rs994674967 Chr.12: 50931634-50931634 on Build GRCh38
rs754366448 Chr.12: 50931711-50931712 on Build GRCh38
rs1003954716 Chr.12: 50931443-50931443 on Build GRCh38
rs993656892 Chr.12: 50931484-50931484 on Build GRCh38
rs3209368 Chr.12: 50931766-50931766 on Build GRCh38
rs567121530 Chr.12: 50931785-50931785 on Build GRCh38
rs569006330 Chr.12: 50931828-50931828 on Build GRCh38
rs952716446 Chr.12: 50931440-50931440 on Build GRCh38
rs779962175 Chr.12: 50931778-50931778 on Build GRCh38
rs3180992 Chr.12: 50931661-50931661 on Build GRCh38
rs3180344 Chr.12: 50931497-50931497 on Build GRCh38
rs967364294 Chr.12: 50931864-50931864 on Build GRCh38
rs398019537 Chr.12: 50931561-50931562 on Build GRCh38
rs3180984 Chr.12: 50931419-50931419 on Build GRCh38
rs980850850 Chr.12: 50931542-50931542 on Build GRCh38
rs3209372 Chr.12: 50931802-50931802 on Build GRCh38
rs946201203 Chr.12: 50931732-50931732 on Build GRCh38
rs3209371 Chr.12: 50931800-50931800 on Build GRCh38
rs537522287 Chr.12: 50931865-50931865 on Build GRCh38

View Full Product Details