Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 173909806-173910322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SP3
Entrez Gene ID
6670
Gene Name
Sp3 transcription factor
Gene Aliases
SPR2
UniGene
Hs.531587
Gene Chromosome Location
Chr.2: 173906459-173965702 on Build GRCh38
Transcripts
NM_001017371.4 NM_001172712.1 NM_003111.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5539571 c.2283C>T p.I761I Chr.2: 173910004-173910004 on Build GRCh38
7209219 c.2174A>G p.D725G Chr.2: 173910113-173910113 on Build GRCh38
1752110 c.2166G>A p.A722A Chr.2: 173910121-173910121 on Build GRCh38
6407588 c.2119A>G p.K707E Chr.2: 173910168-173910168 on Build GRCh38
1728563 c.2096A>G p.H699R Chr.2: 173910191-173910191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556988115 Chr.2: 173910047-173910047 on Build GRCh38
rs556890630 Chr.2: 173909996-173909996 on Build GRCh38
rs762550108 Chr.2: 173910278-173910278 on Build GRCh38
rs201170496 Chr.2: 173910220-173910220 on Build GRCh38
rs761430393 Chr.2: 173910122-173910122 on Build GRCh38
rs772816470 Chr.2: 173910135-173910135 on Build GRCh38
rs570621079 Chr.2: 173909917-173909917 on Build GRCh38
rs753961846 Chr.2: 173910154-173910154 on Build GRCh38
rs780355136 Chr.2: 173910187-173910187 on Build GRCh38
rs764798004 Chr.2: 173910133-173910133 on Build GRCh38
rs777854788 Chr.2: 173910051-173910051 on Build GRCh38
rs758298021 Chr.2: 173910024-173910024 on Build GRCh38
rs536762496 Chr.2: 173910042-173910042 on Build GRCh38
rs1027732476 Chr.2: 173910110-173910110 on Build GRCh38
rs759911765 Chr.2: 173909960-173909960 on Build GRCh38
rs776373136 Chr.2: 173909863-173909863 on Build GRCh38
rs149409403 Chr.2: 173910025-173910025 on Build GRCh38
rs13392217 Chr.2: 173909983-173909983 on Build GRCh38
rs775979032 Chr.2: 173910099-173910099 on Build GRCh38
rs202184835 Chr.2: 173910062-173910062 on Build GRCh38
rs751290873 Chr.2: 173910044-173910044 on Build GRCh38
rs776089745 Chr.2: 173910121-173910121 on Build GRCh38
rs779421195 Chr.2: 173910085-173910085 on Build GRCh38
rs747842049 Chr.2: 173910260-173910260 on Build GRCh38
rs778985458 Chr.2: 173909919-173909919 on Build GRCh38
rs760421387 Chr.2: 173909993-173909993 on Build GRCh38
rs757360982 Chr.2: 173910155-173910155 on Build GRCh38
rs758967155 Chr.2: 173910166-173910166 on Build GRCh38
rs775039079 Chr.2: 173909971-173909971 on Build GRCh38
rs568181781 Chr.2: 173909904-173909904 on Build GRCh38
rs1047477997 Chr.2: 173910256-173910256 on Build GRCh38
rs7561583 Chr.2: 173910145-173910145 on Build GRCh38
rs886581775 Chr.2: 173909836-173909837 on Build GRCh38
rs774776403 Chr.2: 173909958-173909958 on Build GRCh38
rs750583077 Chr.2: 173910265-173910266 on Build GRCh38
rs374953528 Chr.2: 173909964-173909964 on Build GRCh38
rs754663414 Chr.2: 173910148-173910148 on Build GRCh38
rs764393808 Chr.2: 173910153-173910153 on Build GRCh38
rs1012004654 Chr.2: 173910268-173910268 on Build GRCh38
rs747326757 Chr.2: 173910100-173910100 on Build GRCh38
rs750246347 Chr.2: 173909922-173909922 on Build GRCh38
rs771496015 Chr.2: 173909950-173909950 on Build GRCh38
rs763696767 Chr.2: 173909893-173909893 on Build GRCh38
rs746337075 Chr.2: 173910086-173910086 on Build GRCh38
rs368858604 Chr.2: 173909999-173909999 on Build GRCh38
rs769549495 Chr.2: 173910265-173910265 on Build GRCh38
rs970711099 Chr.2: 173909857-173909858 on Build GRCh38
rs781585294 Chr.2: 173909928-173909928 on Build GRCh38
rs769876944 Chr.2: 173909938-173909938 on Build GRCh38
rs61748248 Chr.2: 173910119-173910119 on Build GRCh38
rs1006893594 Chr.2: 173909842-173909842 on Build GRCh38
rs371446873 Chr.2: 173910018-173910018 on Build GRCh38
rs763034180 Chr.2: 173910138-173910138 on Build GRCh38
rs772543416 Chr.2: 173910097-173910097 on Build GRCh38
rs748360180 Chr.2: 173909936-173909936 on Build GRCh38
rs1035643032 Chr.2: 173909924-173909924 on Build GRCh38
rs779008205 Chr.2: 173910162-173910162 on Build GRCh38
rs758379337 Chr.2: 173910285-173910286 on Build GRCh38
rs185888392 Chr.2: 173909949-173909949 on Build GRCh38
rs554621330 Chr.2: 173910069-173910069 on Build GRCh38
rs753262221 Chr.2: 173909913-173909913 on Build GRCh38
rs137998852 Chr.2: 173910287-173910287 on Build GRCh38
rs765365105 Chr.2: 173909935-173909936 on Build GRCh38
rs575224872 Chr.2: 173909998-173909998 on Build GRCh38
rs745932652 Chr.2: 173910164-173910164 on Build GRCh38
rs1017579900 Chr.2: 173910281-173910281 on Build GRCh38
rs768853671 Chr.2: 173910222-173910222 on Build GRCh38
rs757487613 Chr.2: 173909894-173909896 on Build GRCh38
rs765278428 Chr.2: 173910017-173910017 on Build GRCh38
rs777960563 Chr.2: 173909946-173909946 on Build GRCh38
rs1016479935 Chr.2: 173909845-173909845 on Build GRCh38
rs776375159 Chr.2: 173910237-173910237 on Build GRCh38
rs532647295 Chr.2: 173910273-173910273 on Build GRCh38

View Full Product Details