Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 24275311-24275816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1217
Entrez Gene ID
56243
Gene Name
KIAA1217
Gene Aliases
SKT
UniGene
Hs.445885
Gene Chromosome Location
Chr.10: 23694746-24547848 on Build GRCh38
Transcripts
NM_001098500.2 NM_001282767.1 NM_001282768.1 NM_019590.4 XM_011519552.2 XM_011519555.2 XM_011519558.2 XM_011519559.2 XM_011519562.2 XM_011519564.2 XM_011519565.2 XM_011519566.2 XM_017016416.1 XM_017016417.1 XM_017016418.1 XM_017016419.1 XM_017016420.1 XM_017016421.1 XM_017016422.1 XM_017016425.1 XM_017016426.1 XM_017016427.1 XM_017016429.1 NR_030334.1

Gene Symbol
MIR603
Entrez Gene ID
693188
Gene Name
microRNA 603
Gene Aliases
MIRN603, hsa-mir-603
UniGene
Gene Chromosome Location
Chr.10: 24275685-24275781 on Build GRCh38
Transcripts
NM_001098500.2 NM_001282767.1 NM_001282768.1 NM_019590.4 XM_011519552.2 XM_011519555.2 XM_011519558.2 XM_011519559.2 XM_011519562.2 XM_011519564.2 XM_011519565.2 XM_011519566.2 XM_017016416.1 XM_017016417.1 XM_017016418.1 XM_017016419.1 XM_017016420.1 XM_017016421.1 XM_017016422.1 XM_017016425.1 XM_017016426.1 XM_017016427.1 XM_017016429.1 NR_030334.1

SNP Detail

SNP ID Chromosome Location
rs758263826 Chr.10: 24275756-24275756 on Build GRCh38
rs949583421 Chr.10: 24275447-24275447 on Build GRCh38
rs376669016 Chr.10: 24275456-24275456 on Build GRCh38
rs146060498 Chr.10: 24275663-24275663 on Build GRCh38
rs142760601 Chr.10: 24275441-24275441 on Build GRCh38
rs369057409 Chr.10: 24275646-24275646 on Build GRCh38
rs974227979 Chr.10: 24275733-24275733 on Build GRCh38
rs757089689 Chr.10: 24275637-24275637 on Build GRCh38
rs773716103 Chr.10: 24275717-24275717 on Build GRCh38
rs748400551 Chr.10: 24275789-24275789 on Build GRCh38
rs1009411406 Chr.10: 24275524-24275524 on Build GRCh38
rs532988442 Chr.10: 24275745-24275745 on Build GRCh38
rs542109865 Chr.10: 24275473-24275473 on Build GRCh38
rs747179363 Chr.10: 24275678-24275678 on Build GRCh38
rs372348663 Chr.10: 24275465-24275465 on Build GRCh38
rs948724194 Chr.10: 24275463-24275463 on Build GRCh38
rs777040577 Chr.10: 24275691-24275691 on Build GRCh38
rs377666570 Chr.10: 24275669-24275669 on Build GRCh38
rs758454803 Chr.10: 24275657-24275657 on Build GRCh38
rs755147142 Chr.10: 24275771-24275771 on Build GRCh38
rs1028437245 Chr.10: 24275625-24275625 on Build GRCh38
rs527458545 Chr.10: 24275501-24275501 on Build GRCh38
rs781521931 Chr.10: 24275776-24275776 on Build GRCh38
rs907270891 Chr.10: 24275469-24275469 on Build GRCh38
rs934427756 Chr.10: 24275434-24275434 on Build GRCh38
rs965230696 Chr.10: 24275531-24275531 on Build GRCh38
rs888491215 Chr.10: 24275486-24275486 on Build GRCh38
rs577997259 Chr.10: 24275435-24275435 on Build GRCh38
rs11014002 Chr.10: 24275724-24275724 on Build GRCh38
rs1004319701 Chr.10: 24275499-24275499 on Build GRCh38
rs1022181583 Chr.10: 24275577-24275577 on Build GRCh38
rs770310311 Chr.10: 24275703-24275703 on Build GRCh38
rs967673079 Chr.10: 24275359-24275359 on Build GRCh38
rs1046614086 Chr.10: 24275455-24275455 on Build GRCh38
rs932300097 Chr.10: 24275333-24275333 on Build GRCh38
rs980107180 Chr.10: 24275770-24275770 on Build GRCh38
rs878967501 Chr.10: 24275727-24275727 on Build GRCh38
rs760239002 Chr.10: 24275739-24275739 on Build GRCh38
rs978455658 Chr.10: 24275398-24275398 on Build GRCh38
rs531645932 Chr.10: 24275462-24275462 on Build GRCh38
rs747373840 Chr.10: 24275672-24275673 on Build GRCh38
rs768985215 Chr.10: 24275688-24275688 on Build GRCh38
rs778185770 Chr.10: 24275645-24275646 on Build GRCh38
rs768494913 Chr.10: 24275588-24275588 on Build GRCh38
rs761557273 Chr.10: 24275746-24275746 on Build GRCh38
rs769971847 Chr.10: 24275475-24275475 on Build GRCh38
rs776423486 Chr.10: 24275658-24275658 on Build GRCh38
rs1028050617 Chr.10: 24275372-24275372 on Build GRCh38
rs561885901 Chr.10: 24275500-24275500 on Build GRCh38
rs925658915 Chr.10: 24275624-24275624 on Build GRCh38
rs368831545 Chr.10: 24275736-24275736 on Build GRCh38
rs771429679 Chr.10: 24275735-24275739 on Build GRCh38
rs768455504 Chr.10: 24275631-24275631 on Build GRCh38
rs549516435 Chr.10: 24275790-24275790 on Build GRCh38
rs763717489 Chr.10: 24275744-24275744 on Build GRCh38
rs372743442 Chr.10: 24275738-24275738 on Build GRCh38
rs780096355 Chr.10: 24275768-24275768 on Build GRCh38
rs778789530 Chr.10: 24275645-24275645 on Build GRCh38
rs750318040 Chr.10: 24275751-24275751 on Build GRCh38
rs978382212 Chr.10: 24275605-24275605 on Build GRCh38
rs955313770 Chr.10: 24275402-24275402 on Build GRCh38
rs996498126 Chr.10: 24275598-24275598 on Build GRCh38
rs188364596 Chr.10: 24275365-24275365 on Build GRCh38
rs746953286 Chr.10: 24275483-24275483 on Build GRCh38
rs373927490 Chr.10: 24275632-24275632 on Build GRCh38
rs908740374 Chr.10: 24275429-24275429 on Build GRCh38
rs563888729 Chr.10: 24275788-24275788 on Build GRCh38
rs986975954 Chr.10: 24275412-24275412 on Build GRCh38
rs748490402 Chr.10: 24275697-24275697 on Build GRCh38
rs374852885 Chr.10: 24275686-24275686 on Build GRCh38

View Full Product Details