Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 116536245-116536741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00173
Entrez Gene ID
100287569
Gene Name
long intergenic non-protein coding RNA 173
Gene Aliases
NCRNA00173
UniGene
Hs.441601
Gene Chromosome Location
Chr.12: 116533422-116536513 on Build GRCh38
Transcripts
NR_027346.1

SNP Detail

SNP ID Chromosome Location
rs569949575 Chr.12: 116536490-116536490 on Build GRCh38
rs891044651 Chr.12: 116536383-116536383 on Build GRCh38
rs533703588 Chr.12: 116536616-116536616 on Build GRCh38
rs10850662 Chr.12: 116536323-116536323 on Build GRCh38
rs143535036 Chr.12: 116536331-116536331 on Build GRCh38
rs909296832 Chr.12: 116536324-116536324 on Build GRCh38
rs777960420 Chr.12: 116536377-116536377 on Build GRCh38
rs910487514 Chr.12: 116536670-116536670 on Build GRCh38
rs909315999 Chr.12: 116536283-116536283 on Build GRCh38
rs186308575 Chr.12: 116536336-116536336 on Build GRCh38
rs932495336 Chr.12: 116536352-116536352 on Build GRCh38
rs1022739474 Chr.12: 116536458-116536458 on Build GRCh38
rs1050666479 Chr.12: 116536339-116536339 on Build GRCh38
rs902943844 Chr.12: 116536407-116536407 on Build GRCh38
rs1016244972 Chr.12: 116536542-116536542 on Build GRCh38
rs963410886 Chr.12: 116536563-116536563 on Build GRCh38
rs569047529 Chr.12: 116536591-116536591 on Build GRCh38
rs1035401805 Chr.12: 116536419-116536419 on Build GRCh38
rs1042763362 Chr.12: 116536406-116536406 on Build GRCh38
rs987040683 Chr.12: 116536648-116536648 on Build GRCh38
rs559181799 Chr.12: 116536568-116536568 on Build GRCh38
rs75839519 Chr.12: 116536662-116536662 on Build GRCh38
rs191653479 Chr.12: 116536446-116536446 on Build GRCh38
rs934303832 Chr.12: 116536432-116536432 on Build GRCh38
rs553789367 Chr.12: 116536649-116536649 on Build GRCh38
rs1011356284 Chr.12: 116536449-116536449 on Build GRCh38
rs947182204 Chr.12: 116536673-116536673 on Build GRCh38
rs906004867 Chr.12: 116536459-116536459 on Build GRCh38
rs11068062 Chr.12: 116536611-116536611 on Build GRCh38
rs1011377221 Chr.12: 116536362-116536362 on Build GRCh38
rs1036368712 Chr.12: 116536280-116536280 on Build GRCh38
rs962157251 Chr.12: 116536594-116536594 on Build GRCh38
rs969524150 Chr.12: 116536539-116536539 on Build GRCh38
rs766266878 Chr.12: 116536578-116536578 on Build GRCh38
rs1042878504 Chr.12: 116536693-116536693 on Build GRCh38
rs909264954 Chr.12: 116536573-116536573 on Build GRCh38
rs74678942 Chr.12: 116536604-116536604 on Build GRCh38
rs951077120 Chr.12: 116536535-116536535 on Build GRCh38
rs888975872 Chr.12: 116536341-116536341 on Build GRCh38
rs998892198 Chr.12: 116536414-116536414 on Build GRCh38
rs892878871 Chr.12: 116536447-116536449 on Build GRCh38
rs902932634 Chr.12: 116536695-116536695 on Build GRCh38
rs111942413 Chr.12: 116536326-116536326 on Build GRCh38
rs538790980 Chr.12: 116536502-116536502 on Build GRCh38
rs1023906959 Chr.12: 116536536-116536536 on Build GRCh38
rs530049724 Chr.12: 116536443-116536443 on Build GRCh38

View Full Product Details