Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 32825181-32825713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00486
Entrez Gene ID
285045
Gene Name
long intergenic non-protein coding RNA 486
Gene Aliases
-
UniGene
Hs.434660
Gene Chromosome Location
Chr.2: 32825443-32937203 on Build GRCh38
Transcripts
NR_027098.1

SNP Detail

SNP ID Chromosome Location
rs1047233485 Chr.2: 32825239-32825239 on Build GRCh38
rs1002243959 Chr.2: 32825364-32825364 on Build GRCh38
rs908681311 Chr.2: 32825277-32825277 on Build GRCh38
rs34577739 Chr.2: 32825236-32825236 on Build GRCh38
rs776610831 Chr.2: 32825303-32825303 on Build GRCh38
rs752084075 Chr.2: 32825565-32825565 on Build GRCh38
rs192574309 Chr.2: 32825334-32825334 on Build GRCh38
rs529229941 Chr.2: 32825216-32825216 on Build GRCh38
rs943984593 Chr.2: 32825676-32825676 on Build GRCh38
rs371199661 Chr.2: 32825455-32825455 on Build GRCh38
rs1008056896 Chr.2: 32825454-32825454 on Build GRCh38
rs567985905 Chr.2: 32825682-32825682 on Build GRCh38
rs887037463 Chr.2: 32825635-32825635 on Build GRCh38
rs748091502 Chr.2: 32825408-32825408 on Build GRCh38
rs760267206 Chr.2: 32825388-32825388 on Build GRCh38
rs967574636 Chr.2: 32825434-32825434 on Build GRCh38
rs554489550 Chr.2: 32825630-32825630 on Build GRCh38
rs746238067 Chr.2: 32825615-32825615 on Build GRCh38
rs764045740 Chr.2: 32825386-32825386 on Build GRCh38
rs370158497 Chr.2: 32825684-32825684 on Build GRCh38
rs753503404 Chr.2: 32825409-32825409 on Build GRCh38
rs1019990531 Chr.2: 32825459-32825459 on Build GRCh38
rs754501187 Chr.2: 32825473-32825473 on Build GRCh38
rs780306983 Chr.2: 32825646-32825646 on Build GRCh38
rs7602181 Chr.2: 32825569-32825569 on Build GRCh38
rs879835852 Chr.2: 32825381-32825381 on Build GRCh38
rs150182698 Chr.2: 32825563-32825563 on Build GRCh38
rs919702078 Chr.2: 32825209-32825209 on Build GRCh38
rs893922684 Chr.2: 32825404-32825404 on Build GRCh38
rs551904214 Chr.2: 32825509-32825509 on Build GRCh38
rs952483706 Chr.2: 32825561-32825561 on Build GRCh38
rs1045826350 Chr.2: 32825258-32825258 on Build GRCh38
rs957781795 Chr.2: 32825493-32825493 on Build GRCh38
rs531722105 Chr.2: 32825466-32825466 on Build GRCh38
rs749520866 Chr.2: 32825683-32825683 on Build GRCh38
rs905436321 Chr.2: 32825355-32825355 on Build GRCh38
rs975079207 Chr.2: 32825485-32825485 on Build GRCh38
rs1055614447 Chr.2: 32825302-32825302 on Build GRCh38
rs990363047 Chr.2: 32825508-32825508 on Build GRCh38
rs898361290 Chr.2: 32825372-32825372 on Build GRCh38
rs982299448 Chr.2: 32825589-32825589 on Build GRCh38
rs1007864563 Chr.2: 32825442-32825442 on Build GRCh38
rs778677509 Chr.2: 32825585-32825587 on Build GRCh38
rs1046940632 Chr.2: 32825616-32825616 on Build GRCh38
rs949665124 Chr.2: 32825290-32825290 on Build GRCh38
rs549134731 Chr.2: 32825220-32825220 on Build GRCh38
rs1019291507 Chr.2: 32825449-32825450 on Build GRCh38
rs772713169 Chr.2: 32825629-32825629 on Build GRCh38
rs937439279 Chr.2: 32825566-32825566 on Build GRCh38
rs1053767447 Chr.2: 32825394-32825394 on Build GRCh38
rs1020568468 Chr.2: 32825539-32825539 on Build GRCh38
rs757870775 Chr.2: 32825568-32825568 on Build GRCh38
rs938230067 Chr.2: 32825641-32825641 on Build GRCh38
rs963912740 Chr.2: 32825477-32825477 on Build GRCh38
rs967343607 Chr.2: 32825543-32825543 on Build GRCh38
rs79950238 Chr.2: 32825210-32825210 on Build GRCh38
rs919565699 Chr.2: 32825519-32825519 on Build GRCh38

View Full Product Details