Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 32927144-32927682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00486
Entrez Gene ID
285045
Gene Name
long intergenic non-protein coding RNA 486
Gene Aliases
-
UniGene
Hs.434660
Gene Chromosome Location
Chr.2: 32825443-32937203 on Build GRCh38
Transcripts
NR_027098.1 NR_027099.1 NR_027097.1

Gene Symbol
LOC100271832
Entrez Gene ID
100271832
Gene Name
uncharacterized LOC100271832
Gene Aliases
-
UniGene
Hs.743603
Gene Chromosome Location
Chr.2: 32927127-32946136 on Build GRCh38
Transcripts
NR_027098.1 NR_027099.1 NR_027097.1

SNP Detail

SNP ID Chromosome Location
rs113816503 Chr.2: 32927331-32927331 on Build GRCh38
rs945707059 Chr.2: 32927232-32927232 on Build GRCh38
rs749689105 Chr.2: 32927407-32927407 on Build GRCh38
rs772127460 Chr.2: 32927522-32927522 on Build GRCh38
rs557861207 Chr.2: 32927646-32927646 on Build GRCh38
rs917552429 Chr.2: 32927184-32927184 on Build GRCh38
rs1055244130 Chr.2: 32927278-32927278 on Build GRCh38
rs951377918 Chr.2: 32927548-32927548 on Build GRCh38
rs981105005 Chr.2: 32927517-32927517 on Build GRCh38
rs981192969 Chr.2: 32927246-32927246 on Build GRCh38
rs537845285 Chr.2: 32927636-32927636 on Build GRCh38
rs918400182 Chr.2: 32927540-32927540 on Build GRCh38
rs925020257 Chr.2: 32927643-32927643 on Build GRCh38
rs555831468 Chr.2: 32927578-32927578 on Build GRCh38
rs530852202 Chr.2: 32927306-32927306 on Build GRCh38
rs562111442 Chr.2: 32927267-32927267 on Build GRCh38
rs930808702 Chr.2: 32927591-32927591 on Build GRCh38
rs916714020 Chr.2: 32927587-32927587 on Build GRCh38
rs962567958 Chr.2: 32927492-32927492 on Build GRCh38
rs879505206 Chr.2: 32927564-32927564 on Build GRCh38
rs1020640818 Chr.2: 32927510-32927510 on Build GRCh38
rs1040418945 Chr.2: 32927645-32927645 on Build GRCh38
rs1005310520 Chr.2: 32927357-32927357 on Build GRCh38
rs372854952 Chr.2: 32927419-32927419 on Build GRCh38
rs188121227 Chr.2: 32927451-32927451 on Build GRCh38
rs958488256 Chr.2: 32927526-32927526 on Build GRCh38
rs967043723 Chr.2: 32927514-32927517 on Build GRCh38
rs111277234 Chr.2: 32927465-32927465 on Build GRCh38
rs947805849 Chr.2: 32927187-32927187 on Build GRCh38
rs936393330 Chr.2: 32927644-32927644 on Build GRCh38
rs140128546 Chr.2: 32927397-32927397 on Build GRCh38
rs577831043 Chr.2: 32927652-32927652 on Build GRCh38
rs904068146 Chr.2: 32927279-32927279 on Build GRCh38
rs1040453125 Chr.2: 32927392-32927392 on Build GRCh38
rs35054628 Chr.2: 32927166-32927167 on Build GRCh38
rs570613461 Chr.2: 32927339-32927339 on Build GRCh38
rs372971015 Chr.2: 32927349-32927349 on Build GRCh38
rs4444590 Chr.2: 32927178-32927178 on Build GRCh38
rs759751276 Chr.2: 32927186-32927186 on Build GRCh38
rs1011454516 Chr.2: 32927468-32927468 on Build GRCh38
rs529173555 Chr.2: 32927212-32927212 on Build GRCh38
rs953370000 Chr.2: 32927430-32927430 on Build GRCh38
rs143140969 Chr.2: 32927264-32927264 on Build GRCh38
rs973918104 Chr.2: 32927527-32927527 on Build GRCh38
rs947743648 Chr.2: 32927607-32927607 on Build GRCh38
rs908133290 Chr.2: 32927628-32927628 on Build GRCh38
rs760576235 Chr.2: 32927626-32927626 on Build GRCh38
rs557810653 Chr.2: 32927592-32927592 on Build GRCh38

View Full Product Details