Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73735616-73736110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD11
Entrez Gene ID
83451
Gene Name
abhydrolase domain containing 11
Gene Aliases
PP1226, WBSCR21
UniGene
Hs.647045
Gene Chromosome Location
Chr.7: 73736094-73738867 on Build GRCh38
Transcripts
NM_001145364.2 NM_001301058.1 NM_001321382.1 NM_001321383.1 NM_148912.3 NM_148913.3 NR_026912.2 NR_135627.1 XM_006716144.2 XM_011516590.2 XM_011516591.1 NR_026690.1

Gene Symbol
ABHD11-AS1
Entrez Gene ID
171022
Gene Name
ABHD11 antisense RNA 1 (tail to tail)
Gene Aliases
LINC00035, NCRNA00035, WBSCR26
UniGene
Hs.650513
Gene Chromosome Location
Chr.7: 73735069-73736000 on Build GRCh38
Transcripts
NM_001145364.2 NM_001301058.1 NM_001321382.1 NM_001321383.1 NM_148912.3 NM_148913.3 NR_026912.2 NR_135627.1 XM_006716144.2 XM_011516590.2 XM_011516591.1 NR_026690.1

SNP Detail

SNP ID Chromosome Location
rs782676002 Chr.7: 73736030-73736030 on Build GRCh38
rs117303091 Chr.7: 73735829-73735829 on Build GRCh38
rs782506956 Chr.7: 73735937-73735937 on Build GRCh38
rs143145722 Chr.7: 73735764-73735764 on Build GRCh38
rs1005219563 Chr.7: 73735736-73735736 on Build GRCh38
rs1043283300 Chr.7: 73735809-73735809 on Build GRCh38
rs916160744 Chr.7: 73736055-73736055 on Build GRCh38
rs903309551 Chr.7: 73735651-73735651 on Build GRCh38
rs999256838 Chr.7: 73735664-73735664 on Build GRCh38
rs952555486 Chr.7: 73735717-73735717 on Build GRCh38
rs782547525 Chr.7: 73736067-73736067 on Build GRCh38
rs1041421581 Chr.7: 73735835-73735835 on Build GRCh38
rs200403289 Chr.7: 73735777-73735777 on Build GRCh38
rs184322075 Chr.7: 73735693-73735693 on Build GRCh38
rs782605222 Chr.7: 73735828-73735828 on Build GRCh38
rs782715554 Chr.7: 73735737-73735737 on Build GRCh38
rs961398964 Chr.7: 73735974-73735974 on Build GRCh38
rs996684938 Chr.7: 73735904-73735904 on Build GRCh38
rs1035067905 Chr.7: 73735997-73735997 on Build GRCh38
rs1005332474 Chr.7: 73735965-73735965 on Build GRCh38
rs35181612 Chr.7: 73735791-73735791 on Build GRCh38
rs149788470 Chr.7: 73735709-73735710 on Build GRCh38
rs924824250 Chr.7: 73736086-73736086 on Build GRCh38
rs557103599 Chr.7: 73735712-73735712 on Build GRCh38
rs901069172 Chr.7: 73735696-73735696 on Build GRCh38
rs978908722 Chr.7: 73736068-73736068 on Build GRCh38
rs370893374 Chr.7: 73736057-73736057 on Build GRCh38
rs540181318 Chr.7: 73735776-73735776 on Build GRCh38
rs888402645 Chr.7: 73735946-73735946 on Build GRCh38
rs34891294 Chr.7: 73735705-73735705 on Build GRCh38
rs754915211 Chr.7: 73735935-73735935 on Build GRCh38
rs961187223 Chr.7: 73735745-73735745 on Build GRCh38
rs374107642 Chr.7: 73735752-73735752 on Build GRCh38
rs371076218 Chr.7: 73736008-73736008 on Build GRCh38
rs562705747 Chr.7: 73736033-73736033 on Build GRCh38
rs558019839 Chr.7: 73735760-73735760 on Build GRCh38
rs535685463 Chr.7: 73735707-73735707 on Build GRCh38
rs569088042 Chr.7: 73735748-73735748 on Build GRCh38
rs546779539 Chr.7: 73735672-73735672 on Build GRCh38
rs947536417 Chr.7: 73735795-73735795 on Build GRCh38
rs549126223 Chr.7: 73735851-73735851 on Build GRCh38
rs1028084731 Chr.7: 73735713-73735713 on Build GRCh38
rs544099611 Chr.7: 73735989-73735989 on Build GRCh38
rs1015456073 Chr.7: 73735973-73735973 on Build GRCh38
rs189308070 Chr.7: 73735878-73735878 on Build GRCh38

View Full Product Details