Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 72233826-72234374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC392232
Entrez Gene ID
392232
Gene Name
transient receptor potential cation channel subfamily A member 1 pseudogene
Gene Aliases
-
UniGene
Hs.454043
Gene Chromosome Location
Chr.8: 72202752-72251634 on Build GRCh38
Transcripts
NR_033867.2

SNP Detail

SNP ID Chromosome Location
rs72534880 Chr.8: 72233940-72233941 on Build GRCh38
rs116140299 Chr.8: 72233900-72233900 on Build GRCh38
rs917741270 Chr.8: 72234344-72234344 on Build GRCh38
rs887964317 Chr.8: 72233936-72233936 on Build GRCh38
rs189444918 Chr.8: 72234060-72234060 on Build GRCh38
rs937431213 Chr.8: 72233919-72233919 on Build GRCh38
rs988486385 Chr.8: 72233878-72233878 on Build GRCh38
rs1005118344 Chr.8: 72233942-72233942 on Build GRCh38
rs546098050 Chr.8: 72234192-72234192 on Build GRCh38
rs913222018 Chr.8: 72233881-72233881 on Build GRCh38
rs532474413 Chr.8: 72234150-72234150 on Build GRCh38
rs1030457829 Chr.8: 72234213-72234213 on Build GRCh38
rs926500188 Chr.8: 72233957-72233957 on Build GRCh38
rs917834197 Chr.8: 72234013-72234013 on Build GRCh38
rs935239100 Chr.8: 72233866-72233866 on Build GRCh38
rs1015293632 Chr.8: 72234263-72234263 on Build GRCh38
rs1033865797 Chr.8: 72234303-72234303 on Build GRCh38
rs559244489 Chr.8: 72234211-72234211 on Build GRCh38
rs868155815 Chr.8: 72233940-72233940 on Build GRCh38
rs900553793 Chr.8: 72234196-72234196 on Build GRCh38
rs376820245 Chr.8: 72234207-72234207 on Build GRCh38
rs67199705 Chr.8: 72233935-72233936 on Build GRCh38
rs563556600 Chr.8: 72234071-72234071 on Build GRCh38
rs997143787 Chr.8: 72234005-72234005 on Build GRCh38
rs115275635 Chr.8: 72233923-72233923 on Build GRCh38
rs1000928014 Chr.8: 72234152-72234152 on Build GRCh38
rs572616976 Chr.8: 72233882-72233882 on Build GRCh38
rs962484722 Chr.8: 72233939-72233939 on Build GRCh38
rs745538322 Chr.8: 72234260-72234260 on Build GRCh38
rs1015215076 Chr.8: 72233943-72233943 on Build GRCh38
rs3736140 Chr.8: 72234233-72234233 on Build GRCh38
rs992410158 Chr.8: 72234319-72234319 on Build GRCh38
rs1036532591 Chr.8: 72234337-72234337 on Build GRCh38
rs956774721 Chr.8: 72234165-72234165 on Build GRCh38
rs116835916 Chr.8: 72234229-72234229 on Build GRCh38
rs574881852 Chr.8: 72233988-72233988 on Build GRCh38
rs974214402 Chr.8: 72233947-72233947 on Build GRCh38
rs558858433 Chr.8: 72233879-72233879 on Build GRCh38
rs1028669625 Chr.8: 72234006-72234006 on Build GRCh38
rs567858358 Chr.8: 72234307-72234307 on Build GRCh38
rs1056271052 Chr.8: 72233996-72233996 on Build GRCh38
rs375770708 Chr.8: 72233941-72233942 on Build GRCh38
rs903859940 Chr.8: 72234091-72234091 on Build GRCh38
rs962359545 Chr.8: 72234270-72234270 on Build GRCh38
rs937859829 Chr.8: 72233972-72233972 on Build GRCh38
rs530409933 Chr.8: 72234308-72234308 on Build GRCh38
rs182074992 Chr.8: 72234302-72234302 on Build GRCh38
rs538826488 Chr.8: 72233851-72233851 on Build GRCh38
rs575024230 Chr.8: 72234203-72234203 on Build GRCh38
rs749744186 Chr.8: 72234179-72234179 on Build GRCh38
rs982455515 Chr.8: 72233902-72233902 on Build GRCh38

View Full Product Details