Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 69208265-69208679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPH
Entrez Gene ID
64946
Gene Name
centromere protein H
Gene Aliases
-
UniGene
Hs.631967
Gene Chromosome Location
Chr.5: 69189548-69210357 on Build GRCh38
Transcripts
NM_022909.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3617633 c.591C>T p.I197I Chr.5: 69208299-69208299 on Build GRCh38
7179206 c.568G>A p.E190K Chr.5: 69208276-69208276 on Build GRCh38
4139248 c.604C>T p.L202L Chr.5: 69208312-69208312 on Build GRCh38
1200752 c.570A>C p.E190D Chr.5: 69208278-69208278 on Build GRCh38
6229146 c.595C>T p.R199* Chr.5: 69208303-69208303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936376522 Chr.5: 69208371-69208371 on Build GRCh38
rs762218556 Chr.5: 69208382-69208382 on Build GRCh38
rs774833788 Chr.5: 69208380-69208380 on Build GRCh38
rs1046270990 Chr.5: 69208613-69208613 on Build GRCh38
rs937882447 Chr.5: 69208619-69208619 on Build GRCh38
rs745430240 Chr.5: 69208365-69208365 on Build GRCh38
rs777628754 Chr.5: 69208347-69208347 on Build GRCh38
rs111738491 Chr.5: 69208457-69208457 on Build GRCh38
rs779708820 Chr.5: 69208636-69208636 on Build GRCh38
rs758092279 Chr.5: 69208345-69208345 on Build GRCh38
rs959024292 Chr.5: 69208503-69208503 on Build GRCh38
rs548297135 Chr.5: 69208445-69208445 on Build GRCh38
rs952281663 Chr.5: 69208556-69208556 on Build GRCh38
rs751395109 Chr.5: 69208349-69208349 on Build GRCh38
rs757341786 Chr.5: 69208365-69208368 on Build GRCh38
rs150256446 Chr.5: 69208304-69208304 on Build GRCh38
rs933923933 Chr.5: 69208435-69208435 on Build GRCh38
rs906378618 Chr.5: 69208614-69208614 on Build GRCh38
rs116486664 Chr.5: 69208601-69208601 on Build GRCh38
rs371778599 Chr.5: 69208520-69208520 on Build GRCh38
rs564923468 Chr.5: 69208620-69208620 on Build GRCh38
rs780677432 Chr.5: 69208362-69208362 on Build GRCh38
rs896715783 Chr.5: 69208548-69208548 on Build GRCh38
rs1009437821 Chr.5: 69208431-69208431 on Build GRCh38
rs368701429 Chr.5: 69208372-69208372 on Build GRCh38
rs112298462 Chr.5: 69208522-69208522 on Build GRCh38
rs145927480 Chr.5: 69208446-69208446 on Build GRCh38
rs762392898 Chr.5: 69208290-69208290 on Build GRCh38
rs534721342 Chr.5: 69208496-69208496 on Build GRCh38
rs556875146 Chr.5: 69208548-69208548 on Build GRCh38
rs569890413 Chr.5: 69208303-69208303 on Build GRCh38
rs769314658 Chr.5: 69208368-69208368 on Build GRCh38
rs754138643 Chr.5: 69208338-69208338 on Build GRCh38
rs894592280 Chr.5: 69208651-69208651 on Build GRCh38
rs921053799 Chr.5: 69208561-69208561 on Build GRCh38
rs768841268 Chr.5: 69208379-69208379 on Build GRCh38
rs138892809 Chr.5: 69208333-69208333 on Build GRCh38
rs920940389 Chr.5: 69208336-69208336 on Build GRCh38
rs552579972 Chr.5: 69208469-69208469 on Build GRCh38
rs1014972103 Chr.5: 69208578-69208578 on Build GRCh38
rs757278506 Chr.5: 69208354-69208354 on Build GRCh38
rs200671961 Chr.5: 69208546-69208546 on Build GRCh38
rs759888765 Chr.5: 69208451-69208451 on Build GRCh38
rs774650504 Chr.5: 69208289-69208289 on Build GRCh38
rs1053426486 Chr.5: 69208574-69208574 on Build GRCh38
rs539949847 Chr.5: 69208610-69208610 on Build GRCh38
rs1026978243 Chr.5: 69208514-69208514 on Build GRCh38
rs767689138 Chr.5: 69208302-69208302 on Build GRCh38
rs75093994 Chr.5: 69208602-69208602 on Build GRCh38
rs1046219117 Chr.5: 69208436-69208436 on Build GRCh38
rs2972360 Chr.5: 69208423-69208423 on Build GRCh38
rs139119884 Chr.5: 69208459-69208459 on Build GRCh38
rs2637128 Chr.5: 69208504-69208504 on Build GRCh38
rs548003077 Chr.5: 69208547-69208547 on Build GRCh38
rs557683544 Chr.5: 69208376-69208377 on Build GRCh38
rs530789579 Chr.5: 69208422-69208422 on Build GRCh38
rs748975057 Chr.5: 69208375-69208375 on Build GRCh38
rs757245407 Chr.5: 69208566-69208566 on Build GRCh38
rs936411666 Chr.5: 69208567-69208567 on Build GRCh38
rs570752058 Chr.5: 69208494-69208494 on Build GRCh38
rs2637129 Chr.5: 69208549-69208549 on Build GRCh38
rs763458354 Chr.5: 69208334-69208334 on Build GRCh38
rs571317004 Chr.5: 69208533-69208533 on Build GRCh38
rs754015650 Chr.5: 69208360-69208361 on Build GRCh38

View Full Product Details