Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 77508374-77508509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRX
Entrez Gene ID
546
Gene Name
ATRX, chromatin remodeler
Gene Aliases
ATR2, JMS, MRX52, MRXHF1, RAD54, RAD54L, SFM1, SHS, XH2, XNP, ZNF-HX
UniGene
Hs.533526
Gene Chromosome Location
Chr.X: 77504878-77786269 on Build GRCh38
Transcripts
NM_000489.4 NM_138270.3 XM_005262153.4 XM_005262154.4 XM_005262155.4 XM_005262156.4 XM_005262157.4 XM_006724666.3 XM_006724667.3 XM_017029601.1 XM_017029602.1 XM_017029603.1 XM_017029604.1 XM_017029605.1 XM_017029606.1 XM_017029607.1 XM_017029608.1 XM_017029609.1 XM_017029610.1 XM_017029611.1

SNP Detail

SNP ID Chromosome Location
rs782330964 Chr.X: 77508435-77508435 on Build GRCh38
rs398123428 Chr.X: 77508436-77508436 on Build GRCh38
rs782169215 Chr.X: 77508451-77508451 on Build GRCh38
rs797044723 Chr.X: 77508463-77508464 on Build GRCh38
rs782771681 Chr.X: 77508481-77508481 on Build GRCh38
rs782540364 Chr.X: 77508434-77508434 on Build GRCh38
rs782416494 Chr.X: 77508429-77508429 on Build GRCh38
rs782445691 Chr.X: 77508424-77508424 on Build GRCh38
rs782020537 Chr.X: 77508437-77508437 on Build GRCh38
rs1044007508 Chr.X: 77508405-77508405 on Build GRCh38
rs781975871 Chr.X: 77508431-77508431 on Build GRCh38
rs781835425 Chr.X: 77508409-77508409 on Build GRCh38
rs782631992 Chr.X: 77508430-77508430 on Build GRCh38
rs782050050 Chr.X: 77508432-77508432 on Build GRCh38
rs782570568 Chr.X: 77508428-77508428 on Build GRCh38

View Full Product Details