Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7910332-7910573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP2K7
Entrez Gene ID
5609
Gene Name
mitogen-activated protein kinase kinase 7
Gene Aliases
JNKK2, MAPKK7, MEK, MEK 7, MKK7, PRKMK7, SAPKK-4, SAPKK4
UniGene
Hs.531754
Gene Chromosome Location
Chr.19: 7903780-7914483 on Build GRCh38
Transcripts
NM_001297555.1 NM_001297556.1 NM_145185.3 XM_006722800.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5564878 c.506G>A p.R169H Chr.19: 7910463-7910463 on Build GRCh38
4547375 c.420G>A p.R140R Chr.19: 7910346-7910346 on Build GRCh38
4288889 c.587C>A p.S196Y Chr.19: 7910466-7910466 on Build GRCh38
4082909 c.584G>A p.R195H Chr.19: 7910463-7910463 on Build GRCh38
4082906 c.573+2T>C p.? Chr.19: 7910375-7910375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982421514 Chr.19: 7910397-7910397 on Build GRCh38
rs781180446 Chr.19: 7910393-7910393 on Build GRCh38
rs200200323 Chr.19: 7910362-7910362 on Build GRCh38
rs28395772 Chr.19: 7910400-7910400 on Build GRCh38
rs1049003237 Chr.19: 7910359-7910359 on Build GRCh38
rs768319722 Chr.19: 7910434-7910434 on Build GRCh38
rs373472944 Chr.19: 7910364-7910364 on Build GRCh38
rs765198147 Chr.19: 7910376-7910376 on Build GRCh38
rs775088413 Chr.19: 7910432-7910432 on Build GRCh38
rs779365415 Chr.19: 7910534-7910534 on Build GRCh38
rs766336838 Chr.19: 7910504-7910504 on Build GRCh38
rs749643554 Chr.19: 7910401-7910401 on Build GRCh38
rs371522920 Chr.19: 7910418-7910418 on Build GRCh38
rs759108043 Chr.19: 7910352-7910352 on Build GRCh38
rs756198072 Chr.19: 7910510-7910510 on Build GRCh38
rs528497143 Chr.19: 7910495-7910495 on Build GRCh38
rs975881933 Chr.19: 7910538-7910538 on Build GRCh38
rs200460916 Chr.19: 7910367-7910367 on Build GRCh38
rs377698263 Chr.19: 7910417-7910417 on Build GRCh38
rs551841220 Chr.19: 7910539-7910539 on Build GRCh38
rs367833467 Chr.19: 7910475-7910475 on Build GRCh38
rs761655517 Chr.19: 7910459-7910459 on Build GRCh38
rs752744062 Chr.19: 7910509-7910509 on Build GRCh38
rs746313373 Chr.19: 7910419-7910419 on Build GRCh38
rs772804453 Chr.19: 7910395-7910395 on Build GRCh38
rs969606750 Chr.19: 7910391-7910391 on Build GRCh38
rs896455254 Chr.19: 7910383-7910383 on Build GRCh38
rs199768951 Chr.19: 7910394-7910394 on Build GRCh38
rs1045580606 Chr.19: 7910460-7910460 on Build GRCh38
rs746502176 Chr.19: 7910543-7910543 on Build GRCh38
rs201775683 Chr.19: 7910533-7910533 on Build GRCh38
rs371056501 Chr.19: 7910386-7910386 on Build GRCh38
rs760290803 Chr.19: 7910433-7910433 on Build GRCh38
rs778044336 Chr.19: 7910527-7910527 on Build GRCh38
rs55772368 Chr.19: 7910542-7910542 on Build GRCh38
rs956946281 Chr.19: 7910409-7910409 on Build GRCh38
rs771779482 Chr.19: 7910353-7910353 on Build GRCh38
rs962404278 Chr.19: 7910408-7910408 on Build GRCh38
rs373551377 Chr.19: 7910467-7910467 on Build GRCh38
rs775147511 Chr.19: 7910358-7910358 on Build GRCh38
rs760421768 Chr.19: 7910545-7910545 on Build GRCh38
rs750417403 Chr.19: 7910385-7910385 on Build GRCh38
rs776192507 Chr.19: 7910436-7910436 on Build GRCh38
rs746254265 Chr.19: 7910535-7910535 on Build GRCh38
rs370478429 Chr.19: 7910530-7910530 on Build GRCh38
rs754078664 Chr.19: 7910528-7910528 on Build GRCh38
rs761549769 Chr.19: 7910368-7910368 on Build GRCh38
rs772722551 Chr.19: 7910428-7910428 on Build GRCh38

View Full Product Details