Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97167472-97167737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD36
Entrez Gene ID
375248
Gene Name
ankyrin repeat domain 36
Gene Aliases
UNQ2430
UniGene
Hs.541894
Gene Chromosome Location
Chr.2: 97113186-97264521 on Build GRCh38
Transcripts
NM_001164315.1 XM_006712514.3 XM_011511130.1 XM_011511131.1 XM_017004009.1 XM_017004010.1 XM_017004011.1 XM_017004012.1 XM_017004013.1 XM_017004014.1 XM_017004015.1 XM_017004016.1 XM_017004017.1 XM_017004018.1 XM_017004019.1 XM_017004020.1 XM_017004021.1 XM_017004022.1 XM_017004023.1 XM_017004024.1 XM_017004025.1 XM_017004026.1 XM_017004027.1 XM_017004028.1 XM_017004029.1 XM_017004030.1 XM_017004031.1 XM_017004032.1 XM_017004033.1 XM_017004034.1 XM_017004035.1 XM_017004036.1 XM_017004037.1 XM_017004038.1 XM_017004039.1 XM_017004040.1 XM_017004041.1 XM_017004042.1 XM_017004043.1 XM_017004044.1 XM_017004045.1 XM_017004046.1 XM_017004047.1 XM_017004048.1 XM_017004049.1 XM_017004050.1 XM_017004051.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4134186 c.1560G>A p.T520T Chr.2: 97167605-97167605 on Build GRCh38
4134185 c.1560G>A p.T520T Chr.2: 97167605-97167605 on Build GRCh38
4594331 c.1597G>A p.A533T Chr.2: 97167731-97167731 on Build GRCh38
4590981 c.1595C>T p.A532V Chr.2: 97167729-97167729 on Build GRCh38
4316992 c.1599C>T p.A533A Chr.2: 97167733-97167733 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916314110 Chr.2: 97167666-97167666 on Build GRCh38
rs371262503 Chr.2: 97167660-97167661 on Build GRCh38
rs530702144 Chr.2: 97167656-97167656 on Build GRCh38
rs758605502 Chr.2: 97167677-97167677 on Build GRCh38
rs5832832 Chr.2: 97167664-97167665 on Build GRCh38
rs1004692306 Chr.2: 97167611-97167611 on Build GRCh38
rs777914199 Chr.2: 97167717-97167717 on Build GRCh38
rs1010966605 Chr.2: 97167517-97167517 on Build GRCh38
rs1636143 Chr.2: 97167657-97167657 on Build GRCh38
rs201686430 Chr.2: 97167603-97167603 on Build GRCh38
rs540366604 Chr.2: 97167536-97167536 on Build GRCh38
rs776363536 Chr.2: 97167533-97167533 on Build GRCh38
rs766800559 Chr.2: 97167623-97167623 on Build GRCh38
rs1358633 Chr.2: 97167624-97167624 on Build GRCh38
rs762690660 Chr.2: 97167588-97167588 on Build GRCh38
rs973153588 Chr.2: 97167647-97167647 on Build GRCh38
rs550493362 Chr.2: 97167674-97167674 on Build GRCh38
rs377717858 Chr.2: 97167664-97167664 on Build GRCh38
rs963170558 Chr.2: 97167616-97167616 on Build GRCh38
rs764633193 Chr.2: 97167552-97167552 on Build GRCh38
rs560646949 Chr.2: 97167574-97167574 on Build GRCh38
rs5008284 Chr.2: 97167605-97167605 on Build GRCh38
rs770723630 Chr.2: 97167528-97167528 on Build GRCh38
rs747535601 Chr.2: 97167672-97167673 on Build GRCh38
rs745410546 Chr.2: 97167663-97167664 on Build GRCh38
rs544826880 Chr.2: 97167604-97167604 on Build GRCh38
rs564802885 Chr.2: 97167615-97167615 on Build GRCh38
rs869080724 Chr.2: 97167665-97167666 on Build GRCh38
rs2924009 Chr.2: 97167500-97167500 on Build GRCh38
rs201829271 Chr.2: 97167662-97167662 on Build GRCh38

View Full Product Details