Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 142784673-142784936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPC1
Entrez Gene ID
7220
Gene Name
transient receptor potential cation channel subfamily C member 1
Gene Aliases
HTRP-1, TRP1
UniGene
Hs.250687
Gene Chromosome Location
Chr.3: 142724032-142807888 on Build GRCh38
Transcripts
NM_001251845.1 NM_003304.4 XM_005247738.3 XM_005247739.2 XM_017007121.1 XM_017007122.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4319672 c.1012C>T p.Q338* Chr.3: 142784755-142784755 on Build GRCh38
7315610 c.1171G>T p.G391* Chr.3: 142784914-142784914 on Build GRCh38
1419655 c.1055A>C p.K352T Chr.3: 142784900-142784900 on Build GRCh38
728953 c.926G>T p.R309L Chr.3: 142784771-142784771 on Build GRCh38
5615212 c.908G>T p.G303V Chr.3: 142784753-142784753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781497762 Chr.3: 142784735-142784735 on Build GRCh38
rs761607753 Chr.3: 142784808-142784808 on Build GRCh38
rs377020498 Chr.3: 142784853-142784853 on Build GRCh38
rs760914870 Chr.3: 142784744-142784749 on Build GRCh38
rs767321616 Chr.3: 142784758-142784758 on Build GRCh38
rs756151939 Chr.3: 142784756-142784756 on Build GRCh38
rs776318165 Chr.3: 142784773-142784773 on Build GRCh38
rs866163394 Chr.3: 142784742-142784745 on Build GRCh38
rs751261909 Chr.3: 142784893-142784893 on Build GRCh38
rs779189422 Chr.3: 142784717-142784717 on Build GRCh38
rs569515062 Chr.3: 142784759-142784759 on Build GRCh38
rs762915978 Chr.3: 142784868-142784868 on Build GRCh38
rs148727851 Chr.3: 142784762-142784762 on Build GRCh38
rs746269004 Chr.3: 142784766-142784766 on Build GRCh38
rs775548212 Chr.3: 142784717-142784720 on Build GRCh38
rs748182947 Chr.3: 142784741-142784741 on Build GRCh38
rs7621642 Chr.3: 142784763-142784763 on Build GRCh38
rs766352624 Chr.3: 142784889-142784889 on Build GRCh38
rs768156525 Chr.3: 142784771-142784771 on Build GRCh38
rs367550915 Chr.3: 142784880-142784880 on Build GRCh38
rs764103703 Chr.3: 142784831-142784831 on Build GRCh38
rs193920786 Chr.3: 142784713-142784713 on Build GRCh38
rs772923487 Chr.3: 142784859-142784859 on Build GRCh38

View Full Product Details