Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 17760732-17760993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF13A
Entrez Gene ID
63971
Gene Name
kinesin family member 13A
Gene Aliases
RBKIN, bA500C11.2
UniGene
Hs.94499
Gene Chromosome Location
Chr.6: 17759183-17987623 on Build GRCh38
Transcripts
NM_001105568.2 XM_017011188.1 XM_017011192.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7373777 c.5245C>T p.R1749W Chr.6: 17760862-17760862 on Build GRCh38
5450219 c.5246G>A p.R1749Q Chr.6: 17760861-17760861 on Build GRCh38
1329200 c.5237C>T p.T1746I Chr.6: 17760870-17760870 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765365649 Chr.6: 17760844-17760844 on Build GRCh38
rs114131556 Chr.6: 17760800-17760800 on Build GRCh38
rs944976795 Chr.6: 17760945-17760945 on Build GRCh38
rs867005050 Chr.6: 17760854-17760854 on Build GRCh38
rs774025095 Chr.6: 17760925-17760925 on Build GRCh38
rs774119207 Chr.6: 17760927-17760927 on Build GRCh38
rs186263133 Chr.6: 17760839-17760839 on Build GRCh38
rs758642501 Chr.6: 17760861-17760861 on Build GRCh38
rs181523053 Chr.6: 17760762-17760762 on Build GRCh38
rs761558475 Chr.6: 17760816-17760816 on Build GRCh38
rs769193146 Chr.6: 17760788-17760788 on Build GRCh38
rs544363587 Chr.6: 17760821-17760821 on Build GRCh38
rs765895602 Chr.6: 17760914-17760914 on Build GRCh38
rs954120520 Chr.6: 17760934-17760934 on Build GRCh38
rs747205208 Chr.6: 17760885-17760885 on Build GRCh38
rs373335964 Chr.6: 17760903-17760903 on Build GRCh38
rs767235308 Chr.6: 17760820-17760820 on Build GRCh38
rs560727466 Chr.6: 17760915-17760915 on Build GRCh38
rs146219124 Chr.6: 17760810-17760810 on Build GRCh38
rs572807998 Chr.6: 17760761-17760761 on Build GRCh38
rs768268381 Chr.6: 17760811-17760811 on Build GRCh38
rs766055442 Chr.6: 17760830-17760830 on Build GRCh38
rs372472253 Chr.6: 17760842-17760842 on Build GRCh38
rs760595549 Chr.6: 17760825-17760825 on Build GRCh38
rs763910687 Chr.6: 17760890-17760890 on Build GRCh38
rs757710087 Chr.6: 17760895-17760895 on Build GRCh38
rs778021266 Chr.6: 17760881-17760881 on Build GRCh38
rs377476321 Chr.6: 17760958-17760958 on Build GRCh38
rs961818707 Chr.6: 17760900-17760900 on Build GRCh38
rs912132273 Chr.6: 17760930-17760930 on Build GRCh38
rs770129319 Chr.6: 17760922-17760922 on Build GRCh38
rs1015773709 Chr.6: 17760873-17760873 on Build GRCh38
rs376253065 Chr.6: 17760860-17760860 on Build GRCh38
rs1026991689 Chr.6: 17760933-17760933 on Build GRCh38
rs920721319 Chr.6: 17760864-17760864 on Build GRCh38
rs527982023 Chr.6: 17760963-17760963 on Build GRCh38
rs753497348 Chr.6: 17760926-17760927 on Build GRCh38
rs773674531 Chr.6: 17760815-17760815 on Build GRCh38
rs1057194794 Chr.6: 17760836-17760836 on Build GRCh38

View Full Product Details