Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52291356-52291581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT81
Entrez Gene ID
3887
Gene Name
keratin 81
Gene Aliases
HB1, Hb-1, KRTHB1, MLN137, ghHkb1, hHAKB2-1
UniGene
Hs.658118
Gene Chromosome Location
Chr.12: 52285913-52291515 on Build GRCh38
Transcripts
NM_002281.3 NM_001320198.1 XM_005268866.4 XM_017019296.1

Gene Symbol
KRT86
Entrez Gene ID
3892
Gene Name
keratin 86
Gene Aliases
HB6, Hb1, K86, KRTHB1, KRTHB6, MNX
UniGene
Hs.278658
Gene Chromosome Location
Chr.12: 52270507-52309163 on Build GRCh38
Transcripts
NM_002281.3 NM_001320198.1 XM_005268866.4 XM_017019296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4696909 c.62G>A p.R21Q Chr.12: 52291404-52291404 on Build GRCh38
6990692 c.60G>C p.P20P Chr.12: 52291406-52291406 on Build GRCh38
3871856 c.91C>T p.P31S Chr.12: 52291375-52291375 on Build GRCh38
2010290 c.71G>A p.R24H Chr.12: 52291395-52291395 on Build GRCh38
1362456 c.54C>T p.C18C Chr.12: 52291412-52291412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754915803 Chr.12: 52291440-52291440 on Build GRCh38
rs772904497 Chr.12: 52291479-52291479 on Build GRCh38
rs1035279501 Chr.12: 52291555-52291555 on Build GRCh38
rs778690347 Chr.12: 52291496-52291496 on Build GRCh38
rs759416656 Chr.12: 52291493-52291493 on Build GRCh38
rs749936395 Chr.12: 52291428-52291428 on Build GRCh38
rs745959240 Chr.12: 52291464-52291464 on Build GRCh38
rs756076197 Chr.12: 52291400-52291400 on Build GRCh38
rs202023317 Chr.12: 52291465-52291465 on Build GRCh38
rs747238469 Chr.12: 52291376-52291376 on Build GRCh38
rs770423410 Chr.12: 52291459-52291459 on Build GRCh38
rs770430502 Chr.12: 52291412-52291412 on Build GRCh38
rs767116184 Chr.12: 52291425-52291425 on Build GRCh38
rs776029360 Chr.12: 52291389-52291389 on Build GRCh38
rs761394571 Chr.12: 52291419-52291419 on Build GRCh38
rs763968162 Chr.12: 52291513-52291513 on Build GRCh38
rs767061164 Chr.12: 52291399-52291399 on Build GRCh38
rs768939809 Chr.12: 52291417-52291417 on Build GRCh38
rs769155748 Chr.12: 52291473-52291473 on Build GRCh38
rs747238851 Chr.12: 52291407-52291407 on Build GRCh38
rs546202958 Chr.12: 52291448-52291448 on Build GRCh38
rs189745410 Chr.12: 52291506-52291506 on Build GRCh38
rs149101320 Chr.12: 52291418-52291418 on Build GRCh38
rs994093592 Chr.12: 52291532-52291532 on Build GRCh38
rs771258593 Chr.12: 52291409-52291409 on Build GRCh38
rs780179904 Chr.12: 52291469-52291469 on Build GRCh38
rs770985846 Chr.12: 52291377-52291377 on Build GRCh38
rs758210072 Chr.12: 52291508-52291508 on Build GRCh38
rs763504601 Chr.12: 52291415-52291415 on Build GRCh38
rs765859692 Chr.12: 52291433-52291433 on Build GRCh38
rs773246999 Chr.12: 52291374-52291374 on Build GRCh38
rs765794740 Chr.12: 52291380-52291380 on Build GRCh38
rs549397349 Chr.12: 52291396-52291396 on Build GRCh38
rs202176338 Chr.12: 52291379-52291379 on Build GRCh38
rs555244465 Chr.12: 52291411-52291411 on Build GRCh38
rs776185671 Chr.12: 52291489-52291489 on Build GRCh38
rs753675735 Chr.12: 52291405-52291405 on Build GRCh38
rs79897879 Chr.12: 52291429-52291429 on Build GRCh38
rs763161208 Chr.12: 52291390-52291390 on Build GRCh38
rs148003777 Chr.12: 52291478-52291478 on Build GRCh38
rs1053032079 Chr.12: 52291446-52291446 on Build GRCh38
rs547619585 Chr.12: 52291551-52291551 on Build GRCh38
rs202205489 Chr.12: 52291404-52291404 on Build GRCh38
rs781499301 Chr.12: 52291456-52291456 on Build GRCh38
rs368423298 Chr.12: 52291474-52291474 on Build GRCh38
rs765911343 Chr.12: 52291486-52291486 on Build GRCh38
rs750743058 Chr.12: 52291517-52291517 on Build GRCh38
rs558774301 Chr.12: 52291395-52291395 on Build GRCh38
rs765185794 Chr.12: 52291500-52291500 on Build GRCh38
rs200940564 Chr.12: 52291512-52291512 on Build GRCh38
rs748558354 Chr.12: 52291403-52291403 on Build GRCh38
rs75660643 Chr.12: 52291406-52291406 on Build GRCh38
rs753782485 Chr.12: 52291438-52291438 on Build GRCh38
rs897177661 Chr.12: 52291521-52291521 on Build GRCh38
rs760314261 Chr.12: 52291484-52291484 on Build GRCh38
rs527487684 Chr.12: 52291534-52291534 on Build GRCh38
rs757588620 Chr.12: 52291455-52291455 on Build GRCh38
rs183915325 Chr.12: 52291444-52291444 on Build GRCh38
rs936080123 Chr.12: 52291439-52291439 on Build GRCh38
rs889907683 Chr.12: 52291504-52291504 on Build GRCh38

View Full Product Details