Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128693589-128693860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SET
Entrez Gene ID
6418
Gene Name
SET nuclear proto-oncogene
Gene Aliases
2PP2A, I2PP2A, IGAAD, IPP2A2, PHAPII, TAF-I, TAF-IBETA
UniGene
Hs.436687
Gene Chromosome Location
Chr.9: 128683432-128696396 on Build GRCh38
Transcripts
NM_001122821.1 NM_001248000.1 NM_001248001.1 NM_003011.3 XM_017015013.1 XM_017015014.1 XM_017015015.1 XM_017015016.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1292800 c.680C>T p.P227L Chr.9: 128693786-128693786 on Build GRCh38
6330037 c.477G>A p.S159S Chr.9: 128693655-128693655 on Build GRCh38
1265319 c.675T>C p.I225I Chr.9: 128693781-128693781 on Build GRCh38
6329583 c.563C>T p.A188V Chr.9: 128693741-128693741 on Build GRCh38
384861 c.630T>A p.D210E Chr.9: 128693775-128693775 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576392697 Chr.9: 128693664-128693664 on Build GRCh38
rs150832256 Chr.9: 128693663-128693663 on Build GRCh38
rs4836619 Chr.9: 128693755-128693755 on Build GRCh38
rs142648600 Chr.9: 128693654-128693654 on Build GRCh38
rs75427242 Chr.9: 128693754-128693754 on Build GRCh38
rs139428399 Chr.9: 128693651-128693651 on Build GRCh38
rs746575818 Chr.9: 128693816-128693816 on Build GRCh38
rs764082920 Chr.9: 128693655-128693655 on Build GRCh38
rs200600519 Chr.9: 128693712-128693712 on Build GRCh38
rs757228804 Chr.9: 128693818-128693818 on Build GRCh38
rs139252094 Chr.9: 128693736-128693736 on Build GRCh38
rs189358973 Chr.9: 128693834-128693834 on Build GRCh38
rs997815554 Chr.9: 128693631-128693631 on Build GRCh38
rs765701223 Chr.9: 128693646-128693646 on Build GRCh38
rs756532220 Chr.9: 128693718-128693718 on Build GRCh38
rs376525055 Chr.9: 128693751-128693751 on Build GRCh38
rs4836618 Chr.9: 128693624-128693624 on Build GRCh38
rs745644219 Chr.9: 128693835-128693835 on Build GRCh38
rs770136079 Chr.9: 128693630-128693630 on Build GRCh38
rs750786150 Chr.9: 128693715-128693715 on Build GRCh38
rs757886265 Chr.9: 128693796-128693796 on Build GRCh38
rs777172615 Chr.9: 128693799-128693799 on Build GRCh38

View Full Product Details