Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 63915484-63915713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINB10
Entrez Gene ID
5273
Gene Name
serpin family B member 10
Gene Aliases
PI-10, PI10
UniGene
Hs.158339
Gene Chromosome Location
Chr.18: 63907969-63936111 on Build GRCh38
Transcripts
NM_005024.2 XM_011526027.1 XM_017025793.1 XM_017025794.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7172216 c.10C>G p.L4V Chr.18: 63915520-63915520 on Build GRCh38
5388374 c.61G>A p.E21K Chr.18: 63915571-63915571 on Build GRCh38
247257 c.151G>T p.A51S Chr.18: 63915661-63915661 on Build GRCh38
5146186 c.76delA p.N27fs*13 Chr.18: 63915586-63915586 on Build GRCh38
4383189 c.88T>A p.F30I Chr.18: 63915598-63915598 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767132814 Chr.18: 63915538-63915538 on Build GRCh38
rs756322789 Chr.18: 63915650-63915650 on Build GRCh38
rs200832026 Chr.18: 63915684-63915684 on Build GRCh38
rs753186880 Chr.18: 63915560-63915560 on Build GRCh38
rs920260389 Chr.18: 63915556-63915556 on Build GRCh38
rs764668923 Chr.18: 63915644-63915644 on Build GRCh38
rs17072097 Chr.18: 63915517-63915517 on Build GRCh38
rs372835886 Chr.18: 63915541-63915541 on Build GRCh38
rs8097425 Chr.18: 63915633-63915633 on Build GRCh38
rs149980901 Chr.18: 63915581-63915581 on Build GRCh38
rs757455327 Chr.18: 63915576-63915576 on Build GRCh38
rs373691683 Chr.18: 63915614-63915614 on Build GRCh38
rs775153160 Chr.18: 63915594-63915594 on Build GRCh38
rs778008585 Chr.18: 63915541-63915542 on Build GRCh38
rs768478267 Chr.18: 63915667-63915667 on Build GRCh38
rs138905204 Chr.18: 63915640-63915640 on Build GRCh38
rs765566181 Chr.18: 63915632-63915632 on Build GRCh38
rs897823320 Chr.18: 63915677-63915677 on Build GRCh38
rs1040122490 Chr.18: 63915553-63915553 on Build GRCh38
rs758974901 Chr.18: 63915563-63915563 on Build GRCh38
rs138539635 Chr.18: 63915557-63915557 on Build GRCh38
rs749061784 Chr.18: 63915654-63915654 on Build GRCh38
rs747495550 Chr.18: 63915593-63915595 on Build GRCh38
rs751103435 Chr.18: 63915525-63915525 on Build GRCh38
rs35743864 Chr.18: 63915582-63915582 on Build GRCh38
rs760254643 Chr.18: 63915631-63915631 on Build GRCh38
rs913965653 Chr.18: 63915592-63915593 on Build GRCh38
rs944528730 Chr.18: 63915608-63915608 on Build GRCh38
rs758788449 Chr.18: 63915538-63915539 on Build GRCh38
rs767003165 Chr.18: 63915627-63915627 on Build GRCh38
rs35980016 Chr.18: 63915645-63915645 on Build GRCh38
rs757788389 Chr.18: 63915629-63915629 on Build GRCh38
rs750081547 Chr.18: 63915544-63915544 on Build GRCh38
rs768605151 Chr.18: 63915604-63915604 on Build GRCh38
rs931747815 Chr.18: 63915577-63915577 on Build GRCh38
rs377040803 Chr.18: 63915545-63915545 on Build GRCh38
rs762813126 Chr.18: 63915518-63915518 on Build GRCh38
rs779851948 Chr.18: 63915653-63915653 on Build GRCh38
rs1001608296 Chr.18: 63915674-63915674 on Build GRCh38
rs761279299 Chr.18: 63915622-63915622 on Build GRCh38
rs754955613 Chr.18: 63915583-63915583 on Build GRCh38
rs751534966 Chr.18: 63915624-63915624 on Build GRCh38
rs759346717 Chr.18: 63915515-63915515 on Build GRCh38
rs747065892 Chr.18: 63915574-63915574 on Build GRCh38
rs769547584 Chr.18: 63915516-63915516 on Build GRCh38
rs578228782 Chr.18: 63915602-63915602 on Build GRCh38
rs746037318 Chr.18: 63915647-63915647 on Build GRCh38
rs777956377 Chr.18: 63915564-63915564 on Build GRCh38
rs375113713 Chr.18: 63915646-63915646 on Build GRCh38
rs773245289 Chr.18: 63915680-63915680 on Build GRCh38
rs761407423 Chr.18: 63915532-63915532 on Build GRCh38
rs763528128 Chr.18: 63915520-63915520 on Build GRCh38

View Full Product Details