Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 49610863-49611110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHAT
Entrez Gene ID
1103
Gene Name
choline O-acetyltransferase
Gene Aliases
CHOACTASE, CMS1A, CMS1A2, CMS6
UniGene
Hs.302002
Gene Chromosome Location
Chr.10: 49609095-49665104 on Build GRCh38
Transcripts
NM_020984.3 NM_003055.2

Gene Symbol
SLC18A3
Entrez Gene ID
6572
Gene Name
solute carrier family 18 member A3
Gene Aliases
VACHT
UniGene
Hs.654374
Gene Chromosome Location
Chr.10: 49610301-49612720 on Build GRCh38
Transcripts
NM_020984.3 NM_003055.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5550781 c.283G>A p.A95T Chr.10: 49611023-49611023 on Build GRCh38
390424 c.370A>T p.I124F Chr.10: 49611110-49611110 on Build GRCh38
1505368 c.312G>A p.A104A Chr.10: 49611052-49611052 on Build GRCh38
6066088 c.196_197GG>TT p.G66L Chr.10: 49610936-49610937 on Build GRCh38
1561310 c.346C>A p.P116T Chr.10: 49611086-49611086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774410461 Chr.10: 49611089-49611089 on Build GRCh38
rs773724578 Chr.10: 49611066-49611066 on Build GRCh38
rs368999296 Chr.10: 49610882-49610882 on Build GRCh38
rs757220675 Chr.10: 49611030-49611030 on Build GRCh38
rs780898977 Chr.10: 49610887-49610887 on Build GRCh38
rs746944100 Chr.10: 49610936-49610936 on Build GRCh38
rs570262856 Chr.10: 49610907-49610907 on Build GRCh38
rs748792717 Chr.10: 49610906-49610906 on Build GRCh38
rs375068582 Chr.10: 49610949-49610949 on Build GRCh38
rs34959050 Chr.10: 49611069-49611069 on Build GRCh38
rs765344370 Chr.10: 49610945-49610945 on Build GRCh38
rs770222638 Chr.10: 49611062-49611062 on Build GRCh38
rs758567871 Chr.10: 49610948-49610948 on Build GRCh38
rs368610030 Chr.10: 49611076-49611076 on Build GRCh38
rs868440563 Chr.10: 49610903-49610903 on Build GRCh38
rs778997596 Chr.10: 49611036-49611036 on Build GRCh38
rs77672173 Chr.10: 49610923-49610923 on Build GRCh38
rs771775048 Chr.10: 49610926-49610926 on Build GRCh38
rs763102159 Chr.10: 49611004-49611004 on Build GRCh38
rs1018008774 Chr.10: 49611069-49611069 on Build GRCh38
rs371337298 Chr.10: 49611022-49611022 on Build GRCh38
rs778513207 Chr.10: 49610979-49610979 on Build GRCh38
rs748546701 Chr.10: 49611061-49611061 on Build GRCh38
rs749277704 Chr.10: 49610974-49610974 on Build GRCh38
rs200894047 Chr.10: 49611090-49611090 on Build GRCh38
rs748499497 Chr.10: 49610972-49610984 on Build GRCh38
rs752369107 Chr.10: 49610965-49610965 on Build GRCh38
rs769579877 Chr.10: 49610895-49610895 on Build GRCh38
rs756806169 Chr.10: 49610977-49610977 on Build GRCh38
rs375693441 Chr.10: 49611039-49611039 on Build GRCh38
rs747324702 Chr.10: 49610984-49610984 on Build GRCh38
rs761880188 Chr.10: 49610942-49610942 on Build GRCh38
rs150436609 Chr.10: 49610989-49610989 on Build GRCh38
rs771080063 Chr.10: 49611087-49611087 on Build GRCh38
rs774115054 Chr.10: 49610921-49610921 on Build GRCh38
rs538442914 Chr.10: 49611042-49611042 on Build GRCh38
rs550120542 Chr.10: 49610899-49610899 on Build GRCh38
rs765759606 Chr.10: 49610995-49610995 on Build GRCh38
rs552847289 Chr.10: 49610973-49610973 on Build GRCh38
rs750621434 Chr.10: 49610947-49610947 on Build GRCh38
rs779570062 Chr.10: 49611051-49611051 on Build GRCh38
rs775218452 Chr.10: 49610982-49610982 on Build GRCh38
rs8178982 Chr.10: 49610959-49610959 on Build GRCh38
rs751873022 Chr.10: 49611011-49611011 on Build GRCh38
rs367805043 Chr.10: 49611014-49611014 on Build GRCh38
rs571025751 Chr.10: 49611009-49611009 on Build GRCh38
rs779806837 Chr.10: 49610968-49610968 on Build GRCh38
rs771555626 Chr.10: 49610981-49610981 on Build GRCh38
rs917702300 Chr.10: 49610992-49610992 on Build GRCh38
rs755992962 Chr.10: 49610968-49610968 on Build GRCh38
rs369137682 Chr.10: 49610969-49610969 on Build GRCh38
rs71500278 Chr.10: 49611025-49611025 on Build GRCh38
rs770631008 Chr.10: 49610914-49610914 on Build GRCh38
rs773239597 Chr.10: 49611001-49611001 on Build GRCh38
rs568658869 Chr.10: 49611068-49611068 on Build GRCh38
rs138145399 Chr.10: 49610991-49610991 on Build GRCh38
rs955025798 Chr.10: 49610915-49610915 on Build GRCh38
rs556754739 Chr.10: 49611050-49611050 on Build GRCh38
rs775674834 Chr.10: 49610933-49610933 on Build GRCh38
rs747998317 Chr.10: 49610888-49610888 on Build GRCh38
rs547801205 Chr.10: 49610940-49610940 on Build GRCh38
rs763994314 Chr.10: 49611021-49611021 on Build GRCh38
rs768880483 Chr.10: 49611058-49611058 on Build GRCh38
rs764453365 Chr.10: 49610937-49610937 on Build GRCh38
rs199561778 Chr.10: 49611052-49611052 on Build GRCh38
rs776937045 Chr.10: 49611060-49611060 on Build GRCh38
rs919585742 Chr.10: 49610978-49610978 on Build GRCh38
rs1016793644 Chr.10: 49610891-49610891 on Build GRCh38
rs777566600 Chr.10: 49610972-49610972 on Build GRCh38
rs1044883672 Chr.10: 49611017-49611017 on Build GRCh38

View Full Product Details