Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58440684-58440957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VAPB
Entrez Gene ID
9217
Gene Name
VAMP associated protein B and C
Gene Aliases
ALS8, VAMP-B, VAP-B
UniGene
Hs.182625
Gene Chromosome Location
Chr.20: 58389119-58451101 on Build GRCh38
Transcripts
NM_001195677.1 NM_004738.4 NR_036633.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7417399 c.433G>A p.A145T Chr.20: 58440943-58440943 on Build GRCh38
3841376 c.441G>C p.K147N Chr.20: 58440951-58440951 on Build GRCh38
478333 c.419T>C p.I140T Chr.20: 58440929-58440929 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762421026 Chr.20: 58440856-58440856 on Build GRCh38
rs778667587 Chr.20: 58440910-58440910 on Build GRCh38
rs980844441 Chr.20: 58440889-58440889 on Build GRCh38
rs951196318 Chr.20: 58440739-58440739 on Build GRCh38
rs748973669 Chr.20: 58440836-58440836 on Build GRCh38
rs766739004 Chr.20: 58440872-58440872 on Build GRCh38
rs147666916 Chr.20: 58440779-58440779 on Build GRCh38
rs779305153 Chr.20: 58440868-58440868 on Build GRCh38
rs373280837 Chr.20: 58440727-58440727 on Build GRCh38
rs369315612 Chr.20: 58440904-58440904 on Build GRCh38
rs768273548 Chr.20: 58440867-58440867 on Build GRCh38
rs767598860 Chr.20: 58440891-58440891 on Build GRCh38
rs773652590 Chr.20: 58440862-58440862 on Build GRCh38
rs755183003 Chr.20: 58440879-58440879 on Build GRCh38
rs752709224 Chr.20: 58440895-58440895 on Build GRCh38
rs779385532 Chr.20: 58440725-58440725 on Build GRCh38
rs193128804 Chr.20: 58440812-58440812 on Build GRCh38
rs765593198 Chr.20: 58440857-58440857 on Build GRCh38
rs184682435 Chr.20: 58440873-58440873 on Build GRCh38
rs991956338 Chr.20: 58440814-58440814 on Build GRCh38
rs995533772 Chr.20: 58440716-58440716 on Build GRCh38
rs367775205 Chr.20: 58440870-58440870 on Build GRCh38
rs983607139 Chr.20: 58440813-58440813 on Build GRCh38
rs12625788 Chr.20: 58440722-58440722 on Build GRCh38

View Full Product Details