Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45877258-45877525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSWIM3
Entrez Gene ID
140831
Gene Name
zinc finger SWIM-type containing 3
Gene Aliases
C20orf164, PPP1R174
UniGene
Hs.292135
Gene Chromosome Location
Chr.20: 45857581-45879131 on Build GRCh38
Transcripts
NM_080752.3 NR_037628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5755586 c.715G>T p.V239L Chr.20: 45877273-45877273 on Build GRCh38
724435 c.726G>T p.K242N Chr.20: 45877284-45877284 on Build GRCh38
5600949 c.877C>G p.P293A Chr.20: 45877453-45877453 on Build GRCh38
6834417 c.922T>C p.Y308H Chr.20: 45877480-45877480 on Build GRCh38
3405151 c.905G>A p.R302H Chr.20: 45877463-45877463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1030265728 Chr.20: 45877467-45877467 on Build GRCh38
rs367699895 Chr.20: 45877492-45877492 on Build GRCh38
rs772357659 Chr.20: 45877422-45877422 on Build GRCh38
rs748737779 Chr.20: 45877489-45877489 on Build GRCh38
rs747725796 Chr.20: 45877357-45877357 on Build GRCh38
rs779247942 Chr.20: 45877331-45877331 on Build GRCh38
rs750810755 Chr.20: 45877448-45877448 on Build GRCh38
rs762315736 Chr.20: 45877435-45877435 on Build GRCh38
rs565625786 Chr.20: 45877364-45877364 on Build GRCh38
rs1010976619 Chr.20: 45877307-45877307 on Build GRCh38
rs757680101 Chr.20: 45877326-45877326 on Build GRCh38
rs767239922 Chr.20: 45877462-45877462 on Build GRCh38
rs775530105 Chr.20: 45877426-45877426 on Build GRCh38
rs769156098 Chr.20: 45877369-45877369 on Build GRCh38
rs986027175 Chr.20: 45877499-45877499 on Build GRCh38
rs764070928 Chr.20: 45877430-45877430 on Build GRCh38
rs146261883 Chr.20: 45877484-45877484 on Build GRCh38
rs749706288 Chr.20: 45877321-45877321 on Build GRCh38
rs745702737 Chr.20: 45877497-45877497 on Build GRCh38
rs773734126 Chr.20: 45877442-45877444 on Build GRCh38
rs569141770 Chr.20: 45877463-45877463 on Build GRCh38
rs779491530 Chr.20: 45877381-45877381 on Build GRCh38
rs780660086 Chr.20: 45877356-45877356 on Build GRCh38
rs764705608 Chr.20: 45877298-45877298 on Build GRCh38
rs888434366 Chr.20: 45877439-45877439 on Build GRCh38
rs754412817 Chr.20: 45877432-45877432 on Build GRCh38
rs141886372 Chr.20: 45877325-45877325 on Build GRCh38
rs777906861 Chr.20: 45877380-45877380 on Build GRCh38
rs760618511 Chr.20: 45877297-45877297 on Build GRCh38
rs759343926 Chr.20: 45877416-45877416 on Build GRCh38
rs534738492 Chr.20: 45877365-45877365 on Build GRCh38
rs754674476 Chr.20: 45877344-45877344 on Build GRCh38
rs35928298 Chr.20: 45877429-45877429 on Build GRCh38
rs755706420 Chr.20: 45877474-45877474 on Build GRCh38
rs375951724 Chr.20: 45877397-45877397 on Build GRCh38
rs752831495 Chr.20: 45877294-45877294 on Build GRCh38
rs536649474 Chr.20: 45877493-45877493 on Build GRCh38
rs866283655 Chr.20: 45877475-45877475 on Build GRCh38
rs141978692 Chr.20: 45877459-45877459 on Build GRCh38
rs772263510 Chr.20: 45877444-45877444 on Build GRCh38
rs767565586 Chr.20: 45877289-45877289 on Build GRCh38
rs2903808 Chr.20: 45877334-45877334 on Build GRCh38

View Full Product Details