Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 214728811-214729052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BARD1
Entrez Gene ID
580
Gene Name
BRCA1 associated RING domain 1
Gene Aliases
-
UniGene
Hs.591642
Gene Chromosome Location
Chr.2: 214725645-214809711 on Build GRCh38
Transcripts
NM_000465.3 NM_001282543.1 NM_001282545.1 NM_001282548.1 NM_001282549.1 NR_104212.1 NR_104215.1 NR_104216.1 XM_017004613.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1614308 c.2074A>T p.I692F Chr.2: 214728936-214728936 on Build GRCh38
6915084 c.2027A>T p.Y676F Chr.2: 214728983-214728983 on Build GRCh38
5456921 c.2171C>T p.A724V Chr.2: 214728839-214728839 on Build GRCh38
6971889 c.2159T>C p.V720A Chr.2: 214728851-214728851 on Build GRCh38
324796 c.2021G>T p.G674V Chr.2: 214728989-214728989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs876658687 Chr.2: 214728900-214728900 on Build GRCh38
rs759021562 Chr.2: 214728997-214728997 on Build GRCh38
rs771852699 Chr.2: 214728855-214728855 on Build GRCh38
rs878854005 Chr.2: 214728869-214728870 on Build GRCh38
rs750597867 Chr.2: 214728949-214728949 on Build GRCh38
rs730881423 Chr.2: 214728915-214728915 on Build GRCh38
rs368488821 Chr.2: 214728956-214728956 on Build GRCh38
rs876658286 Chr.2: 214728834-214728834 on Build GRCh38
rs864622716 Chr.2: 214728893-214728893 on Build GRCh38
rs587782415 Chr.2: 214728906-214728906 on Build GRCh38
rs864622380 Chr.2: 214728953-214728953 on Build GRCh38
rs746790711 Chr.2: 214728981-214728981 on Build GRCh38
rs111367604 Chr.2: 214728927-214728927 on Build GRCh38
rs779808630 Chr.2: 214728841-214728841 on Build GRCh38
rs876658260 Chr.2: 214729010-214729010 on Build GRCh38
rs749485640 Chr.2: 214728966-214728966 on Build GRCh38
rs747466434 Chr.2: 214728835-214728835 on Build GRCh38
rs150121935 Chr.2: 214728881-214728881 on Build GRCh38
rs139620052 Chr.2: 214728928-214728928 on Build GRCh38
rs143331809 Chr.2: 214728838-214728838 on Build GRCh38
rs786202805 Chr.2: 214728847-214728847 on Build GRCh38
rs786203188 Chr.2: 214728852-214728852 on Build GRCh38
rs546077003 Chr.2: 214728873-214728873 on Build GRCh38
rs730881424 Chr.2: 214728831-214728831 on Build GRCh38
rs764477334 Chr.2: 214728899-214728899 on Build GRCh38
rs149262370 Chr.2: 214728894-214728894 on Build GRCh38
rs1045454971 Chr.2: 214728891-214728891 on Build GRCh38
rs786203617 Chr.2: 214728995-214728995 on Build GRCh38
rs770917842 Chr.2: 214728983-214728983 on Build GRCh38
rs878854006 Chr.2: 214728864-214728864 on Build GRCh38
rs1036838204 Chr.2: 214728939-214728939 on Build GRCh38
rs554708247 Chr.2: 214728849-214728849 on Build GRCh38
rs775039040 Chr.2: 214729002-214729002 on Build GRCh38
rs1040231633 Chr.2: 214728885-214728885 on Build GRCh38
rs876658193 Chr.2: 214728872-214728872 on Build GRCh38
rs771766430 Chr.2: 214728977-214728977 on Build GRCh38
rs876658526 Chr.2: 214728860-214728860 on Build GRCh38
rs564838936 Chr.2: 214728879-214728879 on Build GRCh38
rs868183675 Chr.2: 214728862-214728862 on Build GRCh38
rs573443019 Chr.2: 214728979-214728979 on Build GRCh38
rs187240320 Chr.2: 214729019-214729019 on Build GRCh38
rs780161956 Chr.2: 214728959-214728959 on Build GRCh38
rs587780027 Chr.2: 214729020-214729020 on Build GRCh38
rs201873551 Chr.2: 214728832-214728832 on Build GRCh38
rs863224671 Chr.2: 214728945-214728945 on Build GRCh38
rs786203474 Chr.2: 214728884-214728884 on Build GRCh38
rs769302736 Chr.2: 214729001-214729001 on Build GRCh38
rs587782348 Chr.2: 214728909-214728909 on Build GRCh38
rs752114855 Chr.2: 214728905-214728905 on Build GRCh38
rs587781316 Chr.2: 214728934-214728934 on Build GRCh38
rs587782555 Chr.2: 214728935-214728935 on Build GRCh38
rs786203811 Chr.2: 214728861-214728862 on Build GRCh38
rs111284953 Chr.2: 214728926-214728926 on Build GRCh38
rs587782662 Chr.2: 214728839-214728839 on Build GRCh38
rs786201573 Chr.2: 214728919-214728919 on Build GRCh38
rs776515437 Chr.2: 214728992-214728992 on Build GRCh38
rs759046999 Chr.2: 214728874-214728874 on Build GRCh38
rs137888190 Chr.2: 214728850-214728850 on Build GRCh38
rs765890845 Chr.2: 214728866-214728866 on Build GRCh38
rs773166130 Chr.2: 214728857-214728857 on Build GRCh38
rs786203497 Chr.2: 214728871-214728871 on Build GRCh38
rs781133871 Chr.2: 214728873-214728874 on Build GRCh38
rs372160908 Chr.2: 214728887-214728887 on Build GRCh38
rs762601855 Chr.2: 214729009-214729009 on Build GRCh38
rs199758343 Chr.2: 214729018-214729018 on Build GRCh38
rs760541330 Chr.2: 214728865-214728865 on Build GRCh38
rs876659210 Chr.2: 214728913-214728913 on Build GRCh38

View Full Product Details