Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 26489060-26489317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCKAR
Entrez Gene ID
886
Gene Name
cholecystokinin A receptor
Gene Aliases
CCK-A, CCK1-R, CCK1R, CCKRA
UniGene
Hs.129
Gene Chromosome Location
Chr.4: 26481396-26490420 on Build GRCh38
Transcripts
NM_000730.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1247584 c.323T>C p.I108T Chr.4: 26489274-26489274 on Build GRCh38
3133681 c.332G>A p.S111N Chr.4: 26489265-26489265 on Build GRCh38
5528308 c.316G>A p.D106N Chr.4: 26489281-26489281 on Build GRCh38
3133682 c.302C>A p.P101H Chr.4: 26489295-26489295 on Build GRCh38
4973233 c.358T>G p.F120V Chr.4: 26489239-26489239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762124885 Chr.4: 26489284-26489284 on Build GRCh38
rs966827165 Chr.4: 26489268-26489268 on Build GRCh38
rs895051466 Chr.4: 26489149-26489149 on Build GRCh38
rs1024759767 Chr.4: 26489165-26489165 on Build GRCh38
rs770227531 Chr.4: 26489083-26489083 on Build GRCh38
rs747566096 Chr.4: 26489219-26489219 on Build GRCh38
rs949307306 Chr.4: 26489151-26489151 on Build GRCh38
rs200512105 Chr.4: 26489255-26489255 on Build GRCh38
rs201057238 Chr.4: 26489205-26489205 on Build GRCh38
rs749274561 Chr.4: 26489188-26489188 on Build GRCh38
rs917867671 Chr.4: 26489143-26489143 on Build GRCh38
rs751834504 Chr.4: 26489192-26489192 on Build GRCh38
rs966954990 Chr.4: 26489181-26489181 on Build GRCh38
rs998147624 Chr.4: 26489238-26489238 on Build GRCh38
rs764169394 Chr.4: 26489191-26489191 on Build GRCh38
rs770573001 Chr.4: 26489189-26489189 on Build GRCh38
rs776087260 Chr.4: 26489264-26489264 on Build GRCh38
rs866356134 Chr.4: 26489268-26489269 on Build GRCh38
rs200290624 Chr.4: 26489198-26489198 on Build GRCh38
rs200314183 Chr.4: 26489295-26489295 on Build GRCh38
rs1013477982 Chr.4: 26489158-26489158 on Build GRCh38
rs1006318355 Chr.4: 26489118-26489118 on Build GRCh38
rs1036882880 Chr.4: 26489161-26489161 on Build GRCh38
rs533978182 Chr.4: 26489122-26489122 on Build GRCh38
rs754898946 Chr.4: 26489203-26489203 on Build GRCh38
rs746653160 Chr.4: 26489243-26489243 on Build GRCh38
rs771644173 Chr.4: 26489223-26489223 on Build GRCh38
rs754847511 Chr.4: 26489296-26489296 on Build GRCh38
rs772638309 Chr.4: 26489234-26489234 on Build GRCh38
rs184763738 Chr.4: 26489233-26489233 on Build GRCh38
rs759897337 Chr.4: 26489250-26489250 on Build GRCh38
rs767859493 Chr.4: 26489289-26489289 on Build GRCh38
rs572324768 Chr.4: 26489141-26489141 on Build GRCh38
rs764199682 Chr.4: 26489269-26489269 on Build GRCh38
rs779011641 Chr.4: 26489209-26489209 on Build GRCh38
rs770226902 Chr.4: 26489260-26489260 on Build GRCh38
rs555106345 Chr.4: 26489089-26489089 on Build GRCh38
rs540944060 Chr.4: 26489159-26489159 on Build GRCh38
rs367754706 Chr.4: 26489202-26489202 on Build GRCh38
rs977495548 Chr.4: 26489197-26489197 on Build GRCh38
rs778289728 Chr.4: 26489240-26489240 on Build GRCh38
rs937698633 Chr.4: 26489102-26489102 on Build GRCh38
rs74385997 Chr.4: 26489292-26489292 on Build GRCh38
rs1047941899 Chr.4: 26489088-26489088 on Build GRCh38
rs776973940 Chr.4: 26489245-26489245 on Build GRCh38
rs201475596 Chr.4: 26489291-26489291 on Build GRCh38
rs202206580 Chr.4: 26489244-26489244 on Build GRCh38
rs757457884 Chr.4: 26489195-26489195 on Build GRCh38
rs751636808 Chr.4: 26489272-26489272 on Build GRCh38

View Full Product Details