Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42068652-42068911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G4D
Entrez Gene ID
283748
Gene Name
phospholipase A2 group IVD
Gene Aliases
cPLA2delta
UniGene
Hs.380225
Gene Chromosome Location
Chr.15: 42067562-42094559 on Build GRCh38
Transcripts
NM_178034.3 XM_011521467.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4753714 c.2302C>A p.L768M Chr.15: 42068870-42068870 on Build GRCh38
7114144 c.2370G>C p.Q790H Chr.15: 42068802-42068802 on Build GRCh38
5849168 c.2313G>A p.M771I Chr.15: 42068859-42068859 on Build GRCh38
3816053 c.2351T>C p.L784P Chr.15: 42068821-42068821 on Build GRCh38
3501203 c.2323G>A p.E775K Chr.15: 42068849-42068849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773518016 Chr.15: 42068685-42068685 on Build GRCh38
rs759722214 Chr.15: 42068711-42068711 on Build GRCh38
rs776950765 Chr.15: 42068884-42068884 on Build GRCh38
rs746895776 Chr.15: 42068762-42068762 on Build GRCh38
rs377556063 Chr.15: 42068795-42068795 on Build GRCh38
rs373566512 Chr.15: 42068728-42068728 on Build GRCh38
rs767863748 Chr.15: 42068678-42068678 on Build GRCh38
rs780094261 Chr.15: 42068754-42068754 on Build GRCh38
rs771370348 Chr.15: 42068838-42068838 on Build GRCh38
rs746403466 Chr.15: 42068857-42068857 on Build GRCh38
rs1041751991 Chr.15: 42068677-42068677 on Build GRCh38
rs773573161 Chr.15: 42068785-42068785 on Build GRCh38
rs750052 Chr.15: 42068824-42068824 on Build GRCh38
rs767685181 Chr.15: 42068712-42068712 on Build GRCh38
rs767740297 Chr.15: 42068800-42068800 on Build GRCh38
rs777540857 Chr.15: 42068834-42068834 on Build GRCh38
rs752007123 Chr.15: 42068702-42068702 on Build GRCh38
rs774542401 Chr.15: 42068852-42068852 on Build GRCh38
rs768194154 Chr.15: 42068763-42068763 on Build GRCh38
rs371234748 Chr.15: 42068806-42068806 on Build GRCh38
rs759704219 Chr.15: 42068710-42068710 on Build GRCh38
rs180765637 Chr.15: 42068679-42068679 on Build GRCh38
rs983765161 Chr.15: 42068768-42068768 on Build GRCh38
rs756368840 Chr.15: 42068723-42068723 on Build GRCh38
rs140469782 Chr.15: 42068820-42068820 on Build GRCh38
rs771189883 Chr.15: 42068792-42068792 on Build GRCh38
rs150838987 Chr.15: 42068837-42068837 on Build GRCh38
rs763443223 Chr.15: 42068787-42068787 on Build GRCh38
rs573716409 Chr.15: 42068808-42068808 on Build GRCh38
rs776133237 Chr.15: 42068867-42068867 on Build GRCh38
rs764475029 Chr.15: 42068809-42068809 on Build GRCh38
rs147198080 Chr.15: 42068814-42068814 on Build GRCh38
rs760891913 Chr.15: 42068872-42068872 on Build GRCh38
rs780640309 Chr.15: 42068764-42068764 on Build GRCh38
rs747591819 Chr.15: 42068826-42068826 on Build GRCh38
rs557149489 Chr.15: 42068765-42068765 on Build GRCh38
rs200341650 Chr.15: 42068892-42068892 on Build GRCh38
rs372553469 Chr.15: 42068766-42068766 on Build GRCh38
rs774414741 Chr.15: 42068700-42068701 on Build GRCh38
rs764244132 Chr.15: 42068881-42068881 on Build GRCh38
rs968717385 Chr.15: 42068739-42068739 on Build GRCh38
rs527618735 Chr.15: 42068861-42068861 on Build GRCh38
rs766219776 Chr.15: 42068891-42068891 on Build GRCh38
rs780695430 Chr.15: 42068825-42068825 on Build GRCh38
rs753012259 Chr.15: 42068721-42068721 on Build GRCh38
rs375867370 Chr.15: 42068807-42068807 on Build GRCh38
rs767614655 Chr.15: 42068809-42068809 on Build GRCh38
rs558372548 Chr.15: 42068692-42068692 on Build GRCh38
rs769291984 Chr.15: 42068833-42068833 on Build GRCh38
rs370314190 Chr.15: 42068740-42068740 on Build GRCh38
rs750051 Chr.15: 42068752-42068752 on Build GRCh38
rs202006301 Chr.15: 42068686-42068686 on Build GRCh38
rs1022988094 Chr.15: 42068693-42068693 on Build GRCh38
rs1012944786 Chr.15: 42068688-42068688 on Build GRCh38
rs761901333 Chr.15: 42068889-42068889 on Build GRCh38
rs201203477 Chr.15: 42068731-42068731 on Build GRCh38
rs759933682 Chr.15: 42068796-42068796 on Build GRCh38

View Full Product Details