Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 39191652-39191887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK5
Entrez Gene ID
8645
Gene Name
potassium two pore domain channel subfamily K member 5
Gene Aliases
K2p5.1, KCNK5b, TASK-2, TASK2
UniGene
Hs.444448
Gene Chromosome Location
Chr.6: 39188971-39229475 on Build GRCh38
Transcripts
NM_003740.3 XM_005249456.1 XM_006715235.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3777639 c.701G>T p.G234V Chr.6: 39191689-39191689 on Build GRCh38
3077605 c.679G>A p.V227M Chr.6: 39191711-39191711 on Build GRCh38
3715533 c.682G>C p.E228Q Chr.6: 39191708-39191708 on Build GRCh38
4391290 c.716C>T p.S239F Chr.6: 39191674-39191674 on Build GRCh38
7308017 c.695A>C p.Y232S Chr.6: 39191695-39191695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017757180 Chr.6: 39191712-39191712 on Build GRCh38
rs763936853 Chr.6: 39191721-39191721 on Build GRCh38
rs990953818 Chr.6: 39191849-39191849 on Build GRCh38
rs750384806 Chr.6: 39191733-39191733 on Build GRCh38
rs376599617 Chr.6: 39191741-39191741 on Build GRCh38
rs748910127 Chr.6: 39191759-39191759 on Build GRCh38
rs186749083 Chr.6: 39191799-39191799 on Build GRCh38
rs377240292 Chr.6: 39191761-39191761 on Build GRCh38
rs779431095 Chr.6: 39191751-39191751 on Build GRCh38
rs771542549 Chr.6: 39191766-39191766 on Build GRCh38
rs916313292 Chr.6: 39191862-39191862 on Build GRCh38
rs781160511 Chr.6: 39191730-39191730 on Build GRCh38
rs759708414 Chr.6: 39191687-39191687 on Build GRCh38
rs999370355 Chr.6: 39191771-39191771 on Build GRCh38
rs765252884 Chr.6: 39191696-39191696 on Build GRCh38
rs746730962 Chr.6: 39191786-39191787 on Build GRCh38
rs755614036 Chr.6: 39191742-39191742 on Build GRCh38
rs764343563 Chr.6: 39191801-39191801 on Build GRCh38
rs762146868 Chr.6: 39191749-39191749 on Build GRCh38
rs751336553 Chr.6: 39191802-39191802 on Build GRCh38
rs757159016 Chr.6: 39191729-39191729 on Build GRCh38
rs938149442 Chr.6: 39191863-39191863 on Build GRCh38
rs528721149 Chr.6: 39191867-39191867 on Build GRCh38
rs758564719 Chr.6: 39191719-39191719 on Build GRCh38
rs377076675 Chr.6: 39191800-39191800 on Build GRCh38
rs189125008 Chr.6: 39191812-39191812 on Build GRCh38
rs770323318 Chr.6: 39191787-39191788 on Build GRCh38
rs752898883 Chr.6: 39191714-39191714 on Build GRCh38
rs769855584 Chr.6: 39191790-39191790 on Build GRCh38
rs751424318 Chr.6: 39191724-39191724 on Build GRCh38
rs778274242 Chr.6: 39191762-39191762 on Build GRCh38
rs1038564215 Chr.6: 39191809-39191809 on Build GRCh38
rs777228499 Chr.6: 39191783-39191783 on Build GRCh38

View Full Product Details