Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 7866766-7867029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FASTKD3
Entrez Gene ID
79072
Gene Name
FAST kinase domains 3
Gene Aliases
-
UniGene
Hs.653162
Gene Chromosome Location
Chr.5: 7859159-7869037 on Build GRCh38
Transcripts
NM_024091.3 NR_036553.1 NR_073608.1 XM_006714498.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7111261 c.1126G>C p.E376Q Chr.5: 7866958-7866958 on Build GRCh38
7111260 c.1289G>C p.R430T Chr.5: 7866795-7866795 on Build GRCh38
1252030 c.1207T>A p.F403I Chr.5: 7866877-7866877 on Build GRCh38
1252029 c.1221G>A p.Q407Q Chr.5: 7866863-7866863 on Build GRCh38
3074750 c.1105G>T p.E369* Chr.5: 7866979-7866979 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77130393 Chr.5: 7866914-7866914 on Build GRCh38
rs759102652 Chr.5: 7867006-7867006 on Build GRCh38
rs750213168 Chr.5: 7866868-7866868 on Build GRCh38
rs138946599 Chr.5: 7866828-7866828 on Build GRCh38
rs749691419 Chr.5: 7866825-7866825 on Build GRCh38
rs145140718 Chr.5: 7866816-7866816 on Build GRCh38
rs773592462 Chr.5: 7866989-7866989 on Build GRCh38
rs920032596 Chr.5: 7866892-7866892 on Build GRCh38
rs148214151 Chr.5: 7866889-7866889 on Build GRCh38
rs746313705 Chr.5: 7866833-7866833 on Build GRCh38
rs201039302 Chr.5: 7866976-7866976 on Build GRCh38
rs762935003 Chr.5: 7866869-7866869 on Build GRCh38
rs764801414 Chr.5: 7866804-7866804 on Build GRCh38
rs369728205 Chr.5: 7866956-7866956 on Build GRCh38
rs143045532 Chr.5: 7866863-7866863 on Build GRCh38
rs779187340 Chr.5: 7866805-7866805 on Build GRCh38
rs772706206 Chr.5: 7866838-7866838 on Build GRCh38
rs145776485 Chr.5: 7866792-7866792 on Build GRCh38
rs752235876 Chr.5: 7866917-7866917 on Build GRCh38
rs747144604 Chr.5: 7866960-7866960 on Build GRCh38
rs376555432 Chr.5: 7866934-7866934 on Build GRCh38
rs865816654 Chr.5: 7866847-7866847 on Build GRCh38
rs776679614 Chr.5: 7866855-7866855 on Build GRCh38
rs162030 Chr.5: 7866854-7866854 on Build GRCh38
rs756001427 Chr.5: 7867003-7867004 on Build GRCh38
rs147382123 Chr.5: 7866962-7866962 on Build GRCh38
rs1055100528 Chr.5: 7866950-7866950 on Build GRCh38
rs776914343 Chr.5: 7866967-7866967 on Build GRCh38
rs747546762 Chr.5: 7866834-7866834 on Build GRCh38
rs780480840 Chr.5: 7866795-7866795 on Build GRCh38
rs775771469 Chr.5: 7866845-7866845 on Build GRCh38
rs368966263 Chr.5: 7866803-7866803 on Build GRCh38
rs773035415 Chr.5: 7866985-7866985 on Build GRCh38
rs772755203 Chr.5: 7866945-7866945 on Build GRCh38
rs1001132397 Chr.5: 7866995-7866995 on Build GRCh38
rs139622048 Chr.5: 7867002-7867002 on Build GRCh38
rs748218014 Chr.5: 7866920-7866920 on Build GRCh38
rs770103693 Chr.5: 7866922-7866922 on Build GRCh38
rs147051108 Chr.5: 7866793-7866793 on Build GRCh38
rs771155052 Chr.5: 7866826-7866826 on Build GRCh38
rs951173878 Chr.5: 7866958-7866958 on Build GRCh38
rs769989253 Chr.5: 7866979-7866979 on Build GRCh38
rs141358751 Chr.5: 7866806-7866806 on Build GRCh38
rs746225663 Chr.5: 7866943-7866943 on Build GRCh38
rs780674322 Chr.5: 7866817-7866817 on Build GRCh38
rs766277691 Chr.5: 7866871-7866871 on Build GRCh38
rs201384822 Chr.5: 7866990-7866990 on Build GRCh38
rs570525633 Chr.5: 7866897-7866897 on Build GRCh38

View Full Product Details