Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 125083835-125084108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC12A8
Entrez Gene ID
84561
Gene Name
solute carrier family 12 member 8
Gene Aliases
CCC9
UniGene
Hs.658514
Gene Chromosome Location
Chr.3: 125082636-125212765 on Build GRCh38
Transcripts
NM_001195483.1 NM_024628.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5491710 c.1522C>T p.R508W Chr.3: 125083916-125083916 on Build GRCh38
1037632 c.2193C>T p.F731F Chr.3: 125083956-125083956 on Build GRCh38
5713551 c.1999_2000insC p.Q667fs*12 Chr.3: 125084035-125084036 on Build GRCh38
5638254 c.1402C>A p.Q468K Chr.3: 125084036-125084036 on Build GRCh38
3058064 c.2139C>G p.H713Q Chr.3: 125083896-125083896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761507923 Chr.3: 125084019-125084019 on Build GRCh38
rs756421607 Chr.3: 125083871-125083871 on Build GRCh38
rs75686093 Chr.3: 125084070-125084070 on Build GRCh38
rs2981482 Chr.3: 125084044-125084044 on Build GRCh38
rs772457404 Chr.3: 125083920-125083920 on Build GRCh38
rs754135704 Chr.3: 125083956-125083956 on Build GRCh38
rs35116542 Chr.3: 125083941-125083941 on Build GRCh38
rs201194975 Chr.3: 125084017-125084017 on Build GRCh38
rs373256263 Chr.3: 125083991-125083991 on Build GRCh38
rs752293540 Chr.3: 125083946-125083946 on Build GRCh38
rs567281553 Chr.3: 125083977-125083977 on Build GRCh38
rs375996151 Chr.3: 125083949-125083949 on Build GRCh38
rs767132548 Chr.3: 125084021-125084021 on Build GRCh38
rs200353603 Chr.3: 125084041-125084041 on Build GRCh38
rs765064853 Chr.3: 125083943-125083943 on Build GRCh38
rs780552343 Chr.3: 125083888-125083888 on Build GRCh38
rs756474667 Chr.3: 125083951-125083951 on Build GRCh38
rs1031121023 Chr.3: 125083868-125083868 on Build GRCh38
rs370088988 Chr.3: 125083968-125083968 on Build GRCh38
rs762328693 Chr.3: 125083947-125083947 on Build GRCh38
rs773520894 Chr.3: 125083922-125083922 on Build GRCh38
rs762538191 Chr.3: 125084014-125084014 on Build GRCh38
rs745620565 Chr.3: 125084007-125084007 on Build GRCh38
rs769415902 Chr.3: 125084066-125084066 on Build GRCh38
rs780460449 Chr.3: 125083955-125083955 on Build GRCh38
rs763526253 Chr.3: 125084016-125084016 on Build GRCh38
rs376393434 Chr.3: 125084013-125084013 on Build GRCh38
rs755252842 Chr.3: 125084036-125084036 on Build GRCh38
rs535514603 Chr.3: 125084054-125084054 on Build GRCh38
rs373580339 Chr.3: 125084039-125084039 on Build GRCh38
rs774877324 Chr.3: 125083948-125083948 on Build GRCh38
rs2981483 Chr.3: 125084037-125084037 on Build GRCh38
rs747305033 Chr.3: 125083982-125083982 on Build GRCh38
rs368445439 Chr.3: 125083893-125083893 on Build GRCh38
rs748284533 Chr.3: 125083970-125083970 on Build GRCh38
rs769035505 Chr.3: 125083912-125083912 on Build GRCh38
rs749289617 Chr.3: 125084078-125084078 on Build GRCh38
rs773896213 Chr.3: 125084082-125084082 on Build GRCh38
rs1019588916 Chr.3: 125083897-125083897 on Build GRCh38
rs200565283 Chr.3: 125083923-125083923 on Build GRCh38
rs771546073 Chr.3: 125083915-125083915 on Build GRCh38
rs755555375 Chr.3: 125083965-125083965 on Build GRCh38
rs781232638 Chr.3: 125084045-125084045 on Build GRCh38
rs771266464 Chr.3: 125083987-125083987 on Build GRCh38
rs199682098 Chr.3: 125084048-125084048 on Build GRCh38
rs535475842 Chr.3: 125084077-125084077 on Build GRCh38
rs754266251 Chr.3: 125084029-125084029 on Build GRCh38
rs768564423 Chr.3: 125084081-125084081 on Build GRCh38
rs776564193 Chr.3: 125083942-125083942 on Build GRCh38
rs748152373 Chr.3: 125083916-125083916 on Build GRCh38
rs769800533 Chr.3: 125084012-125084012 on Build GRCh38

View Full Product Details