Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 116847295-116847562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD21
Entrez Gene ID
5885
Gene Name
RAD21 cohesin complex component
Gene Aliases
CDLS4, HR21, HRAD21, MCD1, NXP1, SCC1, hHR21
UniGene
Hs.81848
Gene Chromosome Location
Chr.8: 116845934-116874866 on Build GRCh38
Transcripts
NM_006265.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3781368 c.1878A>C p.P626P Chr.8: 116847518-116847518 on Build GRCh38
5463690 c.1871C>T p.P624L Chr.8: 116847525-116847525 on Build GRCh38
6023545 c.1824_1847del24 p.I609_P616del Chr.8: 116847549-116847572 on Build GRCh38
1095321 c.1850A>G p.Y617C Chr.8: 116847546-116847546 on Build GRCh38
6112171 c.1880G>A p.R627K Chr.8: 116847516-116847516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775266057 Chr.8: 116847532-116847532 on Build GRCh38
rs146026669 Chr.8: 116847498-116847498 on Build GRCh38
rs769825200 Chr.8: 116847508-116847508 on Build GRCh38
rs960641181 Chr.8: 116847321-116847321 on Build GRCh38
rs917289753 Chr.8: 116847376-116847376 on Build GRCh38
rs763059424 Chr.8: 116847533-116847533 on Build GRCh38
rs776417390 Chr.8: 116847483-116847483 on Build GRCh38
rs764524271 Chr.8: 116847449-116847450 on Build GRCh38
rs747754527 Chr.8: 116847463-116847463 on Build GRCh38
rs542080108 Chr.8: 116847391-116847391 on Build GRCh38
rs967777337 Chr.8: 116847389-116847389 on Build GRCh38
rs771689646 Chr.8: 116847467-116847467 on Build GRCh38
rs182038781 Chr.8: 116847350-116847350 on Build GRCh38
rs764118613 Chr.8: 116847538-116847538 on Build GRCh38
rs563358787 Chr.8: 116847415-116847415 on Build GRCh38
rs114862132 Chr.8: 116847469-116847469 on Build GRCh38
rs530788755 Chr.8: 116847460-116847460 on Build GRCh38
rs200117381 Chr.8: 116847482-116847482 on Build GRCh38
rs186493289 Chr.8: 116847475-116847475 on Build GRCh38

View Full Product Details