Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29664701-29664958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPN
Entrez Gene ID
6693
Gene Name
sialophorin
Gene Aliases
CD43, GALGP, GPL115, LSN
UniGene
Hs.632188
Gene Chromosome Location
Chr.16: 29662950-29670507 on Build GRCh38
Transcripts
NM_001030288.2 NM_003123.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5493415 c.1045C>T p.R349W Chr.16: 29664773-29664773 on Build GRCh38
117619 c.1166C>A p.P389H Chr.16: 29664894-29664894 on Build GRCh38
5586675 c.1065C>T p.S355S Chr.16: 29664793-29664793 on Build GRCh38
4564294 c.1112_1113GG>AA p.G371E Chr.16: 29664840-29664841 on Build GRCh38
4522240 c.1152G>A p.V384V Chr.16: 29664880-29664880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763785411 Chr.16: 29664891-29664891 on Build GRCh38
rs772706070 Chr.16: 29664722-29664722 on Build GRCh38
rs745743575 Chr.16: 29664919-29664919 on Build GRCh38
rs780661381 Chr.16: 29664792-29664792 on Build GRCh38
rs202146424 Chr.16: 29664909-29664909 on Build GRCh38
rs767056889 Chr.16: 29664912-29664912 on Build GRCh38
rs777920495 Chr.16: 29664914-29664914 on Build GRCh38
rs752148140 Chr.16: 29664887-29664887 on Build GRCh38
rs751510138 Chr.16: 29664773-29664773 on Build GRCh38
rs766236829 Chr.16: 29664771-29664771 on Build GRCh38
rs868248003 Chr.16: 29664767-29664767 on Build GRCh38
rs759371471 Chr.16: 29664861-29664861 on Build GRCh38
rs775515010 Chr.16: 29664932-29664932 on Build GRCh38
rs549616294 Chr.16: 29664917-29664917 on Build GRCh38
rs745460559 Chr.16: 29664908-29664908 on Build GRCh38
rs769243541 Chr.16: 29664798-29664798 on Build GRCh38
rs781107267 Chr.16: 29664814-29664814 on Build GRCh38
rs753569700 Chr.16: 29664892-29664892 on Build GRCh38
rs769903361 Chr.16: 29664827-29664827 on Build GRCh38
rs36084120 Chr.16: 29664820-29664820 on Build GRCh38
rs763807172 Chr.16: 29664743-29664743 on Build GRCh38
rs749483155 Chr.16: 29664916-29664916 on Build GRCh38
rs777639519 Chr.16: 29664788-29664788 on Build GRCh38
rs750754867 Chr.16: 29664789-29664789 on Build GRCh38
rs770733556 Chr.16: 29664851-29664851 on Build GRCh38
rs574440536 Chr.16: 29664774-29664774 on Build GRCh38
rs369917979 Chr.16: 29664841-29664841 on Build GRCh38
rs373798010 Chr.16: 29664905-29664905 on Build GRCh38
rs369665246 Chr.16: 29664731-29664731 on Build GRCh38
rs556240980 Chr.16: 29664750-29664750 on Build GRCh38
rs914647648 Chr.16: 29664799-29664799 on Build GRCh38
rs770099258 Chr.16: 29664741-29664741 on Build GRCh38
rs781756615 Chr.16: 29664910-29664910 on Build GRCh38
rs1002896688 Chr.16: 29664751-29664751 on Build GRCh38
rs564293552 Chr.16: 29664889-29664889 on Build GRCh38
rs766907270 Chr.16: 29664782-29664782 on Build GRCh38
rs372182301 Chr.16: 29664721-29664721 on Build GRCh38
rs866250284 Chr.16: 29664884-29664884 on Build GRCh38
rs541956371 Chr.16: 29664783-29664783 on Build GRCh38
rs747500313 Chr.16: 29664796-29664796 on Build GRCh38
rs200041567 Chr.16: 29664738-29664738 on Build GRCh38
rs776147972 Chr.16: 29664729-29664729 on Build GRCh38
rs763730594 Chr.16: 29664938-29664938 on Build GRCh38
rs776869439 Chr.16: 29664739-29664739 on Build GRCh38
rs767548713 Chr.16: 29664883-29664883 on Build GRCh38
rs568122397 Chr.16: 29664937-29664937 on Build GRCh38
rs200681097 Chr.16: 29664740-29664740 on Build GRCh38
rs771842135 Chr.16: 29664927-29664927 on Build GRCh38
rs545710589 Chr.16: 29664858-29664858 on Build GRCh38

View Full Product Details