Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 17395101-17395372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC8
Entrez Gene ID
6833
Gene Name
ATP binding cassette subfamily C member 8
Gene Aliases
ABC36, HHF1, HI, HRINS, MRP8, PHHI, SUR, SUR1, SUR1delta2, TNDM2
UniGene
Hs.54470
Gene Chromosome Location
Chr.11: 17392885-17476870 on Build GRCh38
Transcripts
NM_000352.4 NM_001287174.1 XM_011520331.2 XM_017018197.1 XM_017018198.1 XM_017018199.1 XM_017018200.1 XM_017018201.1 XM_017018202.1 XM_017018204.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6420228 c.4392G>T p.K1464N Chr.11: 17395191-17395191 on Build GRCh38
3808873 c.4308-1G>A p.? Chr.11: 17395276-17395276 on Build GRCh38
6408368 c.4319A>G p.D1440G Chr.11: 17395264-17395264 on Build GRCh38
6588440 c.4339G>A p.D1447N Chr.11: 17395244-17395244 on Build GRCh38
3445808 c.4354G>T p.E1452* Chr.11: 17395229-17395229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374703754 Chr.11: 17395241-17395241 on Build GRCh38
rs72559717 Chr.11: 17395214-17395214 on Build GRCh38
rs780047943 Chr.11: 17395298-17395298 on Build GRCh38
rs758410083 Chr.11: 17395151-17395151 on Build GRCh38
rs971604271 Chr.11: 17395207-17395207 on Build GRCh38
rs761043838 Chr.11: 17395275-17395275 on Build GRCh38
rs758844607 Chr.11: 17395261-17395261 on Build GRCh38
rs1033994566 Chr.11: 17395235-17395235 on Build GRCh38
rs564543196 Chr.11: 17395142-17395142 on Build GRCh38
rs1057516404 Chr.11: 17395230-17395230 on Build GRCh38
rs764389246 Chr.11: 17395294-17395294 on Build GRCh38
rs912484225 Chr.11: 17395322-17395322 on Build GRCh38
rs72559716 Chr.11: 17395172-17395172 on Build GRCh38
rs1014389339 Chr.11: 17395194-17395194 on Build GRCh38
rs886041391 Chr.11: 17395277-17395277 on Build GRCh38
rs750266577 Chr.11: 17395295-17395295 on Build GRCh38
rs918457756 Chr.11: 17395262-17395262 on Build GRCh38
rs774418147 Chr.11: 17395187-17395187 on Build GRCh38
rs739690 Chr.11: 17395331-17395331 on Build GRCh38
rs562680077 Chr.11: 17395258-17395258 on Build GRCh38
rs544133684 Chr.11: 17395223-17395223 on Build GRCh38
rs754932948 Chr.11: 17395316-17395316 on Build GRCh38
rs751397035 Chr.11: 17395309-17395309 on Build GRCh38
rs769435360 Chr.11: 17395224-17395224 on Build GRCh38
rs1025154500 Chr.11: 17395311-17395311 on Build GRCh38
rs754903921 Chr.11: 17395163-17395163 on Build GRCh38
rs752809991 Chr.11: 17395269-17395269 on Build GRCh38
rs61880303 Chr.11: 17395334-17395334 on Build GRCh38
rs986564120 Chr.11: 17395318-17395318 on Build GRCh38
rs758090008 Chr.11: 17395296-17395296 on Build GRCh38
rs868347815 Chr.11: 17395220-17395220 on Build GRCh38
rs767436777 Chr.11: 17395267-17395267 on Build GRCh38
rs763277685 Chr.11: 17395233-17395233 on Build GRCh38
rs1039460669 Chr.11: 17395336-17395336 on Build GRCh38
rs193922403 Chr.11: 17395215-17395215 on Build GRCh38
rs748800266 Chr.11: 17395123-17395123 on Build GRCh38
rs973834445 Chr.11: 17395255-17395255 on Build GRCh38
rs369810811 Chr.11: 17395173-17395173 on Build GRCh38
rs778909433 Chr.11: 17395135-17395135 on Build GRCh38
rs759663539 Chr.11: 17395259-17395259 on Build GRCh38
rs961820480 Chr.11: 17395321-17395321 on Build GRCh38
rs1027223698 Chr.11: 17395337-17395337 on Build GRCh38

View Full Product Details