Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 61203051-61203294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP34
Entrez Gene ID
9736
Gene Name
ubiquitin specific peptidase 34
Gene Aliases
-
UniGene
Hs.644708
Gene Chromosome Location
Chr.2: 61187455-61470714 on Build GRCh38
Transcripts
NM_014709.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4996244 c.9418G>A p.D3140N Chr.2: 61203230-61203230 on Build GRCh38
1668963 c.9405T>G p.D3135E Chr.2: 61203243-61203243 on Build GRCh38
6818578 c.9484T>G p.F3162V Chr.2: 61203164-61203164 on Build GRCh38
5449734 c.9472A>G p.N3158D Chr.2: 61203176-61203176 on Build GRCh38
4926476 c.9385-2A>C p.? Chr.2: 61203265-61203265 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043996835 Chr.2: 61203101-61203101 on Build GRCh38
rs766760350 Chr.2: 61203209-61203209 on Build GRCh38
rs775092866 Chr.2: 61203117-61203117 on Build GRCh38
rs376116349 Chr.2: 61203223-61203223 on Build GRCh38
rs773459747 Chr.2: 61203094-61203094 on Build GRCh38
rs762497896 Chr.2: 61203122-61203122 on Build GRCh38
rs371936848 Chr.2: 61203179-61203179 on Build GRCh38
rs748198636 Chr.2: 61203256-61203256 on Build GRCh38
rs748600417 Chr.2: 61203090-61203090 on Build GRCh38
rs199725690 Chr.2: 61203250-61203250 on Build GRCh38
rs946757449 Chr.2: 61203082-61203082 on Build GRCh38
rs772561443 Chr.2: 61203092-61203092 on Build GRCh38
rs755178059 Chr.2: 61203240-61203240 on Build GRCh38
rs761048500 Chr.2: 61203109-61203109 on Build GRCh38
rs769358590 Chr.2: 61203115-61203115 on Build GRCh38
rs763578885 Chr.2: 61203129-61203129 on Build GRCh38
rs761672388 Chr.2: 61203153-61203153 on Build GRCh38
rs767442129 Chr.2: 61203165-61203165 on Build GRCh38
rs369411892 Chr.2: 61203131-61203131 on Build GRCh38

View Full Product Details