Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159714364-159714621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRP
Entrez Gene ID
1401
Gene Name
C-reactive protein, pentraxin-related
Gene Aliases
PTX1
UniGene
Hs.709456
Gene Chromosome Location
Chr.1: 159712289-159714623 on Build GRCh38
Transcripts
NM_000567.2 XM_011509207.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4411318 c.6G>T p.E2D Chr.1: 159714480-159714480 on Build GRCh38
6508015 c.12G>T p.L4L Chr.1: 159714474-159714474 on Build GRCh38
4674776 c.17_18delGT p.C6fs*74 Chr.1: 159714468-159714469 on Build GRCh38
6327397 c.4G>A p.E2K Chr.1: 159714482-159714482 on Build GRCh38
898055 c.54C>T p.G18G Chr.1: 159714432-159714432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773661507 Chr.1: 159714516-159714516 on Build GRCh38
rs767647839 Chr.1: 159714465-159714465 on Build GRCh38
rs35086834 Chr.1: 159714512-159714512 on Build GRCh38
rs200304858 Chr.1: 159714492-159714492 on Build GRCh38
rs543234081 Chr.1: 159714575-159714575 on Build GRCh38
rs766929411 Chr.1: 159714456-159714456 on Build GRCh38
rs770910391 Chr.1: 159714427-159714427 on Build GRCh38
rs751689372 Chr.1: 159714385-159714385 on Build GRCh38
rs756339613 Chr.1: 159714476-159714476 on Build GRCh38
rs867726017 Chr.1: 159714449-159714449 on Build GRCh38
rs1417938 Chr.1: 159714396-159714396 on Build GRCh38
rs769569016 Chr.1: 159714397-159714397 on Build GRCh38
rs540472216 Chr.1: 159714433-159714433 on Build GRCh38
rs756387151 Chr.1: 159714548-159714548 on Build GRCh38
rs749117623 Chr.1: 159714485-159714485 on Build GRCh38
rs775198789 Chr.1: 159714503-159714503 on Build GRCh38
rs778888874 Chr.1: 159714489-159714489 on Build GRCh38
rs199953854 Chr.1: 159714455-159714455 on Build GRCh38
rs745409002 Chr.1: 159714432-159714432 on Build GRCh38
rs868078724 Chr.1: 159714533-159714533 on Build GRCh38
rs369841237 Chr.1: 159714517-159714517 on Build GRCh38
rs779657474 Chr.1: 159714418-159714418 on Build GRCh38
rs868258270 Chr.1: 159714543-159714543 on Build GRCh38
rs376990385 Chr.1: 159714491-159714491 on Build GRCh38
rs774915577 Chr.1: 159714532-159714532 on Build GRCh38
rs760470889 Chr.1: 159714444-159714444 on Build GRCh38
rs112774042 Chr.1: 159714423-159714423 on Build GRCh38
rs765684497 Chr.1: 159714446-159714446 on Build GRCh38
rs375033806 Chr.1: 159714431-159714431 on Build GRCh38
rs777480636 Chr.1: 159714484-159714484 on Build GRCh38
rs766691549 Chr.1: 159714528-159714528 on Build GRCh38
rs3093064 Chr.1: 159714591-159714591 on Build GRCh38
rs773715039 Chr.1: 159714451-159714451 on Build GRCh38
rs755051450 Chr.1: 159714463-159714463 on Build GRCh38
rs934783601 Chr.1: 159714409-159714409 on Build GRCh38
rs949854273 Chr.1: 159714429-159714429 on Build GRCh38
rs757095336 Chr.1: 159714488-159714488 on Build GRCh38
rs113469793 Chr.1: 159714424-159714424 on Build GRCh38
rs972486072 Chr.1: 159714404-159714404 on Build GRCh38
rs781450959 Chr.1: 159714392-159714392 on Build GRCh38
rs564967781 Chr.1: 159714474-159714474 on Build GRCh38
rs370080304 Chr.1: 159714595-159714595 on Build GRCh38
rs752054019 Chr.1: 159714457-159714457 on Build GRCh38
rs777598980 Chr.1: 159714401-159714401 on Build GRCh38
rs142597715 Chr.1: 159714389-159714389 on Build GRCh38
rs746591738 Chr.1: 159714509-159714509 on Build GRCh38
rs775254047 Chr.1: 159714443-159714443 on Build GRCh38

View Full Product Details